Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24092708-24093217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435647 c.530-8T>G p.? Chr.16: 24092783-24092783 on Build GRCh38
3507912 c.668G>A p.W223* Chr.16: 24092929-24092929 on Build GRCh38
3507906 c.610C>T p.P204S Chr.16: 24092871-24092871 on Build GRCh38
4059460 c.532A>C p.R178R Chr.16: 24092793-24092793 on Build GRCh38
5700728 c.611C>A p.P204H Chr.16: 24092872-24092872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029252769 Chr.16: 24092794-24092794 on Build GRCh38
rs939384008 Chr.16: 24093002-24093002 on Build GRCh38
rs880824 Chr.16: 24093007-24093007 on Build GRCh38
rs575165010 Chr.16: 24092733-24092733 on Build GRCh38
rs569862207 Chr.16: 24093139-24093140 on Build GRCh38
rs779094238 Chr.16: 24092796-24092796 on Build GRCh38
rs748510627 Chr.16: 24092862-24092862 on Build GRCh38
rs202027145 Chr.16: 24092964-24092964 on Build GRCh38
rs372031319 Chr.16: 24092975-24092975 on Build GRCh38
rs3729898 Chr.16: 24093001-24093001 on Build GRCh38
rs890180397 Chr.16: 24093138-24093138 on Build GRCh38
rs1052049037 Chr.16: 24093098-24093098 on Build GRCh38
rs185384836 Chr.16: 24093191-24093191 on Build GRCh38
rs781505280 Chr.16: 24093192-24093192 on Build GRCh38
rs8048554 Chr.16: 24093055-24093055 on Build GRCh38
rs777343122 Chr.16: 24092855-24092855 on Build GRCh38
rs916562354 Chr.16: 24093022-24093022 on Build GRCh38
rs758345369 Chr.16: 24092832-24092832 on Build GRCh38
rs752461659 Chr.16: 24092772-24092772 on Build GRCh38
rs3729897 Chr.16: 24092999-24092999 on Build GRCh38
rs759011396 Chr.16: 24092953-24092953 on Build GRCh38
rs199950702 Chr.16: 24092967-24092967 on Build GRCh38
rs770291872 Chr.16: 24092863-24092863 on Build GRCh38
rs781543122 Chr.16: 24092950-24092950 on Build GRCh38
rs569484350 Chr.16: 24093078-24093078 on Build GRCh38
rs758670242 Chr.16: 24092991-24092991 on Build GRCh38
rs747105951 Chr.16: 24092847-24092847 on Build GRCh38
rs917317004 Chr.16: 24092894-24092894 on Build GRCh38
rs550961132 Chr.16: 24093158-24093158 on Build GRCh38
rs997865853 Chr.16: 24093121-24093121 on Build GRCh38
rs3729896 Chr.16: 24092867-24092867 on Build GRCh38
rs1049563527 Chr.16: 24093050-24093050 on Build GRCh38
rs763881337 Chr.16: 24092978-24092978 on Build GRCh38
rs940942417 Chr.16: 24093063-24093063 on Build GRCh38
rs759210165 Chr.16: 24092782-24092782 on Build GRCh38
rs531707938 Chr.16: 24093113-24093113 on Build GRCh38
rs543897172 Chr.16: 24092846-24092846 on Build GRCh38
rs773891890 Chr.16: 24092901-24092901 on Build GRCh38
rs936038722 Chr.16: 24093004-24093004 on Build GRCh38
rs754963571 Chr.16: 24092759-24092759 on Build GRCh38
rs551455890 Chr.16: 24093156-24093156 on Build GRCh38
rs762695587 Chr.16: 24092897-24092897 on Build GRCh38
rs755974100 Chr.16: 24092783-24092783 on Build GRCh38
rs750571880 Chr.16: 24092986-24092986 on Build GRCh38
rs948066479 Chr.16: 24093025-24093025 on Build GRCh38
rs573366163 Chr.16: 24092811-24092811 on Build GRCh38
rs751409221 Chr.16: 24092755-24092755 on Build GRCh38
rs769253047 Chr.16: 24092849-24092849 on Build GRCh38
rs563181974 Chr.16: 24093182-24093182 on Build GRCh38
rs767494458 Chr.16: 24092771-24092771 on Build GRCh38
rs746115990 Chr.16: 24092810-24092810 on Build GRCh38
rs1005322850 Chr.16: 24092813-24092813 on Build GRCh38
rs752290250 Chr.16: 24092958-24092958 on Build GRCh38

View Full Product Details