Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58692467-58692981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51C
Entrez Gene ID
5889
Gene Name
RAD51 paralog C
Gene Aliases
BROVCA3, FANCO, R51H3, RAD51L2
UniGene
Hs.412587
Gene Chromosome Location
Chr.17: 58692538-58734342 on Build GRCh38
Transcripts
NM_002876.3 NM_058216.2 NR_103872.1 NR_103873.1 XM_006722001.3 XM_006722002.3 XM_006722004.3 XM_006722005.3 XM_011525094.2 XM_017024914.1 XM_017024916.1 XM_017024917.1 XM_017024918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6982829 c.37G>A p.D13N Chr.17: 58692680-58692680 on Build GRCh38
4719984 c.61C>A p.P21T Chr.17: 58692704-58692704 on Build GRCh38
1263728 c.12G>A p.K4K Chr.17: 58692655-58692655 on Build GRCh38
1479795 c.61C>T p.P21S Chr.17: 58692704-58692704 on Build GRCh38
7131474 c.90G>T p.A30A Chr.17: 58692733-58692733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934163061 Chr.17: 58692850-58692850 on Build GRCh38
rs533565734 Chr.17: 58692871-58692871 on Build GRCh38
rs575897767 Chr.17: 58692587-58692587 on Build GRCh38
rs777210433 Chr.17: 58692822-58692822 on Build GRCh38
rs746120658 Chr.17: 58692798-58692798 on Build GRCh38
rs757116652 Chr.17: 58692716-58692716 on Build GRCh38
rs982832032 Chr.17: 58692582-58692582 on Build GRCh38
rs764457287 Chr.17: 58692638-58692638 on Build GRCh38
rs757128712 Chr.17: 58692789-58692789 on Build GRCh38
rs565398047 Chr.17: 58692703-58692703 on Build GRCh38
rs760830928 Chr.17: 58692723-58692742 on Build GRCh38
rs1003448484 Chr.17: 58692549-58692549 on Build GRCh38
rs893449142 Chr.17: 58692542-58692542 on Build GRCh38
rs976614805 Chr.17: 58692820-58692820 on Build GRCh38
rs779468229 Chr.17: 58692608-58692608 on Build GRCh38
rs375603530 Chr.17: 58692615-58692615 on Build GRCh38
rs945177565 Chr.17: 58692842-58692842 on Build GRCh38
rs1043707871 Chr.17: 58692906-58692906 on Build GRCh38
rs45574736 Chr.17: 58692905-58692905 on Build GRCh38
rs786203249 Chr.17: 58692730-58692730 on Build GRCh38
rs774528162 Chr.17: 58692630-58692630 on Build GRCh38
rs777585467 Chr.17: 58692658-58692658 on Build GRCh38
rs864622460 Chr.17: 58692721-58692721 on Build GRCh38
rs371608994 Chr.17: 58692778-58692778 on Build GRCh38
rs760603345 Chr.17: 58692947-58692947 on Build GRCh38
rs749150256 Chr.17: 58692715-58692715 on Build GRCh38
rs878855180 Chr.17: 58692713-58692713 on Build GRCh38
rs754498936 Chr.17: 58692696-58692696 on Build GRCh38
rs75889871 Chr.17: 58692523-58692523 on Build GRCh38
rs952662506 Chr.17: 58692577-58692577 on Build GRCh38
rs28910276 Chr.17: 58692677-58692677 on Build GRCh38
rs778780988 Chr.17: 58692795-58692795 on Build GRCh38
rs894759401 Chr.17: 58692901-58692901 on Build GRCh38
rs762060755 Chr.17: 58692689-58692689 on Build GRCh38
rs587781383 Chr.17: 58692777-58692777 on Build GRCh38
rs777554369 Chr.17: 58692714-58692714 on Build GRCh38
rs751117852 Chr.17: 58692652-58692652 on Build GRCh38
rs958870284 Chr.17: 58692553-58692553 on Build GRCh38
rs550757849 Chr.17: 58692745-58692745 on Build GRCh38
rs876659683 Chr.17: 58692729-58692729 on Build GRCh38
rs1055305858 Chr.17: 58692578-58692593 on Build GRCh38
rs774649783 Chr.17: 58692596-58692596 on Build GRCh38
rs1039870300 Chr.17: 58692532-58692532 on Build GRCh38
rs753249247 Chr.17: 58692637-58692637 on Build GRCh38
rs536596205 Chr.17: 58692791-58692791 on Build GRCh38
rs929669006 Chr.17: 58692525-58692525 on Build GRCh38
rs376403182 Chr.17: 58692650-58692650 on Build GRCh38
rs876660695 Chr.17: 58692709-58692709 on Build GRCh38
rs786203944 Chr.17: 58692702-58692702 on Build GRCh38
rs201523760 Chr.17: 58692657-58692657 on Build GRCh38
rs779332818 Chr.17: 58692718-58692718 on Build GRCh38
rs113383404 Chr.17: 58692531-58692531 on Build GRCh38
rs587782511 Chr.17: 58692705-58692705 on Build GRCh38
rs543872977 Chr.17: 58692894-58692894 on Build GRCh38
rs938025916 Chr.17: 58692933-58692933 on Build GRCh38
rs1053439653 Chr.17: 58692635-58692635 on Build GRCh38
rs879254131 Chr.17: 58692764-58692764 on Build GRCh38
rs786204086 Chr.17: 58692761-58692761 on Build GRCh38
rs12946397 Chr.17: 58692618-58692618 on Build GRCh38
rs1000113630 Chr.17: 58692732-58692732 on Build GRCh38
rs921435798 Chr.17: 58692644-58692644 on Build GRCh38
rs764394130 Chr.17: 58692678-58692678 on Build GRCh38
rs892464609 Chr.17: 58692562-58692562 on Build GRCh38
rs759669075 Chr.17: 58692631-58692631 on Build GRCh38
rs763292447 Chr.17: 58692806-58692806 on Build GRCh38
rs762230288 Chr.17: 58692744-58692744 on Build GRCh38
rs786201775 Chr.17: 58692722-58692722 on Build GRCh38
rs758029117 Chr.17: 58692647-58692647 on Build GRCh38
rs1057517641 Chr.17: 58692790-58692790 on Build GRCh38
rs1009246645 Chr.17: 58692543-58692543 on Build GRCh38
rs749498443 Chr.17: 58692659-58692659 on Build GRCh38
rs775871420 Chr.17: 58692673-58692673 on Build GRCh38
rs773998134 Chr.17: 58692749-58692749 on Build GRCh38
rs748589580 Chr.17: 58692828-58692828 on Build GRCh38
rs932368286 Chr.17: 58692530-58692530 on Build GRCh38
rs1057372852 Chr.17: 58692900-58692900 on Build GRCh38
rs766958594 Chr.17: 58692771-58692771 on Build GRCh38
rs572500997 Chr.17: 58692519-58692519 on Build GRCh38
rs765425309 Chr.17: 58692595-58692595 on Build GRCh38
rs878855177 Chr.17: 58692790-58692795 on Build GRCh38
rs876658796 Chr.17: 58692642-58692642 on Build GRCh38
rs16943176 Chr.17: 58692526-58692526 on Build GRCh38
rs566163602 Chr.17: 58692863-58692863 on Build GRCh38
rs761536774 Chr.17: 58692826-58692827 on Build GRCh38
rs780116924 Chr.17: 58692803-58692803 on Build GRCh38
rs759149207 Chr.17: 58692758-58692758 on Build GRCh38
rs939036084 Chr.17: 58692899-58692899 on Build GRCh38
rs768612583 Chr.17: 58692817-58692817 on Build GRCh38
rs768182004 Chr.17: 58692636-58692636 on Build GRCh38
rs577305218 Chr.17: 58692908-58692908 on Build GRCh38
rs542951292 Chr.17: 58692599-58692599 on Build GRCh38
rs892567748 Chr.17: 58692663-58692663 on Build GRCh38
rs943856771 Chr.17: 58692520-58692520 on Build GRCh38
rs545213879 Chr.17: 58692541-58692541 on Build GRCh38
rs750069703 Chr.17: 58692846-58692865 on Build GRCh38
rs587781309 Chr.17: 58692723-58692723 on Build GRCh38
rs369854656 Chr.17: 58692805-58692805 on Build GRCh38
rs1012969444 Chr.17: 58692568-58692568 on Build GRCh38
rs769255656 Chr.17: 58692736-58692736 on Build GRCh38
rs773517999 Chr.17: 58692837-58692837 on Build GRCh38
rs769053886 Chr.17: 58692646-58692646 on Build GRCh38
rs757856869 Chr.17: 58692607-58692607 on Build GRCh38
rs759759863 Chr.17: 58692662-58692662 on Build GRCh38
rs777004225 Chr.17: 58692681-58692681 on Build GRCh38
rs375889604 Chr.17: 58692625-58692625 on Build GRCh38
rs576540617 Chr.17: 58692892-58692892 on Build GRCh38
rs28363298 Chr.17: 58692590-58692590 on Build GRCh38
rs576364420 Chr.17: 58692838-58692838 on Build GRCh38
rs796780656 Chr.17: 58692878-58692878 on Build GRCh38
rs765072353 Chr.17: 58692837-58692856 on Build GRCh38
rs778493896 Chr.17: 58692605-58692605 on Build GRCh38
rs768020569 Chr.17: 58692601-58692601 on Build GRCh38
rs910155232 Chr.17: 58692529-58692529 on Build GRCh38
rs752608224 Chr.17: 58692704-58692704 on Build GRCh38
rs753709131 Chr.17: 58692788-58692788 on Build GRCh38
rs1010017149 Chr.17: 58692563-58692563 on Build GRCh38
rs770175762 Chr.17: 58692834-58692834 on Build GRCh38
rs770111989 Chr.17: 58692706-58692706 on Build GRCh38
rs568912602 Chr.17: 58692784-58692784 on Build GRCh38
rs587782698 Chr.17: 58692667-58692667 on Build GRCh38
rs776552532 Chr.17: 58692743-58692744 on Build GRCh38
rs563690436 Chr.17: 58692571-58692571 on Build GRCh38
rs536114335 Chr.17: 58692493-58692493 on Build GRCh38
rs547142453 Chr.17: 58692695-58692695 on Build GRCh38
rs941631413 Chr.17: 58692621-58692621 on Build GRCh38
rs587780840 Chr.17: 58692740-58692741 on Build GRCh38
rs558097188 Chr.17: 58692886-58692886 on Build GRCh38
rs876660577 Chr.17: 58692793-58692793 on Build GRCh38
rs775482687 Chr.17: 58692715-58692716 on Build GRCh38
rs1055706551 Chr.17: 58692935-58692935 on Build GRCh38
rs899583055 Chr.17: 58692528-58692528 on Build GRCh38
rs1016111699 Chr.17: 58692948-58692948 on Build GRCh38
rs864622151 Chr.17: 58692641-58692641 on Build GRCh38
rs181444396 Chr.17: 58692620-58692620 on Build GRCh38
rs746978518 Chr.17: 58692610-58692610 on Build GRCh38
rs587781441 Chr.17: 58692735-58692735 on Build GRCh38
rs876658482 Chr.17: 58692639-58692639 on Build GRCh38
rs746026526 Chr.17: 58692720-58692720 on Build GRCh38
rs750074739 Chr.17: 58692606-58692606 on Build GRCh38
rs766058636 Chr.17: 58692691-58692691 on Build GRCh38
rs1035896435 Chr.17: 58692552-58692552 on Build GRCh38
rs748248444 Chr.17: 58692676-58692676 on Build GRCh38
rs1027059784 Chr.17: 58692775-58692775 on Build GRCh38
rs554552471 Chr.17: 58692495-58692495 on Build GRCh38
rs876659581 Chr.17: 58692712-58692712 on Build GRCh38
rs774685897 Chr.17: 58692661-58692661 on Build GRCh38
rs28363299 Chr.17: 58692591-58692591 on Build GRCh38
rs876658798 Chr.17: 58692679-58692679 on Build GRCh38
rs730881936 Chr.17: 58692672-58692672 on Build GRCh38
rs189059313 Chr.17: 58692527-58692527 on Build GRCh38
rs908469236 Chr.17: 58692514-58692514 on Build GRCh38
rs786202265 Chr.17: 58692757-58692757 on Build GRCh38
rs762043712 Chr.17: 58692593-58692593 on Build GRCh38
rs747492326 Chr.17: 58692734-58692734 on Build GRCh38
rs917018276 Chr.17: 58692597-58692597 on Build GRCh38
rs888522190 Chr.17: 58692909-58692909 on Build GRCh38
rs1042346993 Chr.17: 58692544-58692544 on Build GRCh38
rs903457948 Chr.17: 58692548-58692548 on Build GRCh38
rs771332058 Chr.17: 58692660-58692660 on Build GRCh38
rs764003475 Chr.17: 58692953-58692953 on Build GRCh38
rs730881937 Chr.17: 58692675-58692675 on Build GRCh38
rs115414895 Chr.17: 58692733-58692733 on Build GRCh38
rs373116323 Chr.17: 58692603-58692603 on Build GRCh38
rs377297129 Chr.17: 58692800-58692800 on Build GRCh38
rs587782528 Chr.17: 58692740-58692740 on Build GRCh38
rs750045091 Chr.17: 58692643-58692643 on Build GRCh38
rs730881942 Chr.17: 58692736-58692736 on Build GRCh38
rs927006535 Chr.17: 58692505-58692505 on Build GRCh38
rs781166242 Chr.17: 58692655-58692655 on Build GRCh38
rs538205403 Chr.17: 58692929-58692929 on Build GRCh38
rs747962411 Chr.17: 58692619-58692619 on Build GRCh38
rs752117032 Chr.17: 58692598-58692598 on Build GRCh38
rs901144205 Chr.17: 58692959-58692959 on Build GRCh38

View Full Product Details