Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132658094-132658601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL13
Entrez Gene ID
3596
Gene Name
interleukin 13
Gene Aliases
IL-13, P600
UniGene
Hs.845
Gene Chromosome Location
Chr.5: 132658173-132661109 on Build GRCh38
Transcripts
NM_002188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4456453 c.100C>T p.P34S Chr.5: 132658286-132658286 on Build GRCh38
6325135 c.59C>T p.T20I Chr.5: 132658245-132658245 on Build GRCh38
1242373 c.47C>T p.A16V Chr.5: 132658233-132658233 on Build GRCh38
6684869 c.78T>A p.T26T Chr.5: 132658264-132658264 on Build GRCh38
4530009 c.165G>A p.Q55Q Chr.5: 132658351-132658351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773714921 Chr.5: 132658334-132658334 on Build GRCh38
rs866145689 Chr.5: 132658302-132658302 on Build GRCh38
rs903576385 Chr.5: 132658351-132658351 on Build GRCh38
rs185375084 Chr.5: 132658473-132658473 on Build GRCh38
rs1022009733 Chr.5: 132658140-132658140 on Build GRCh38
rs879943037 Chr.5: 132658313-132658313 on Build GRCh38
rs919980028 Chr.5: 132658240-132658240 on Build GRCh38
rs754964536 Chr.5: 132658251-132658251 on Build GRCh38
rs777533503 Chr.5: 132658279-132658279 on Build GRCh38
rs538901488 Chr.5: 132658567-132658567 on Build GRCh38
rs768184182 Chr.5: 132658142-132658142 on Build GRCh38
rs778963746 Chr.5: 132658191-132658191 on Build GRCh38
rs947830837 Chr.5: 132658325-132658325 on Build GRCh38
rs771833459 Chr.5: 132658205-132658205 on Build GRCh38
rs1011100276 Chr.5: 132658525-132658526 on Build GRCh38
rs911055382 Chr.5: 132658446-132658446 on Build GRCh38
rs762306581 Chr.5: 132658225-132658225 on Build GRCh38
rs757510998 Chr.5: 132658141-132658141 on Build GRCh38
rs531039668 Chr.5: 132658451-132658451 on Build GRCh38
rs955794889 Chr.5: 132658208-132658208 on Build GRCh38
rs928620851 Chr.5: 132658493-132658493 on Build GRCh38
rs1026495086 Chr.5: 132658255-132658255 on Build GRCh38
rs71583493 Chr.5: 132658470-132658470 on Build GRCh38
rs78181870 Chr.5: 132658248-132658248 on Build GRCh38
rs56035208 Chr.5: 132658321-132658321 on Build GRCh38
rs200852476 Chr.5: 132658162-132658162 on Build GRCh38
rs202220644 Chr.5: 132658288-132658288 on Build GRCh38
rs199845178 Chr.5: 132658370-132658370 on Build GRCh38
rs200571678 Chr.5: 132658311-132658311 on Build GRCh38
rs756326109 Chr.5: 132658156-132658156 on Build GRCh38
rs201172066 Chr.5: 132658369-132658369 on Build GRCh38
rs1047967775 Chr.5: 132658482-132658482 on Build GRCh38
rs867567803 Chr.5: 132658185-132658185 on Build GRCh38
rs202009492 Chr.5: 132658178-132658178 on Build GRCh38
rs769152159 Chr.5: 132658309-132658309 on Build GRCh38
rs1037778711 Chr.5: 132658455-132658455 on Build GRCh38
rs201578359 Chr.5: 132658127-132658127 on Build GRCh38
rs200542257 Chr.5: 132658361-132658361 on Build GRCh38
rs190132828 Chr.5: 132658491-132658491 on Build GRCh38
rs1038146791 Chr.5: 132658203-132658203 on Build GRCh38
rs143149119 Chr.5: 132658575-132658575 on Build GRCh38
rs199924106 Chr.5: 132658234-132658234 on Build GRCh38
rs112896241 Chr.5: 132658412-132658412 on Build GRCh38
rs201053365 Chr.5: 132658241-132658241 on Build GRCh38
rs992079803 Chr.5: 132658531-132658531 on Build GRCh38
rs1008430709 Chr.5: 132658432-132658432 on Build GRCh38
rs951529860 Chr.5: 132658486-132658486 on Build GRCh38
rs374069088 Chr.5: 132658308-132658308 on Build GRCh38
rs200581554 Chr.5: 132658126-132658126 on Build GRCh38
rs758908496 Chr.5: 132658222-132658222 on Build GRCh38
rs200726947 Chr.5: 132658397-132658397 on Build GRCh38
rs55733734 Chr.5: 132658246-132658246 on Build GRCh38
rs768364050 Chr.5: 132658332-132658332 on Build GRCh38
rs202011628 Chr.5: 132658315-132658315 on Build GRCh38
rs148621685 Chr.5: 132658552-132658552 on Build GRCh38
rs375964270 Chr.5: 132658161-132658161 on Build GRCh38
rs376870298 Chr.5: 132658233-132658233 on Build GRCh38
rs536445464 Chr.5: 132658534-132658534 on Build GRCh38
rs200638839 Chr.5: 132658294-132658294 on Build GRCh38
rs767987922 Chr.5: 132658229-132658229 on Build GRCh38
rs939290474 Chr.5: 132658530-132658530 on Build GRCh38
rs967742677 Chr.5: 132658153-132658153 on Build GRCh38
rs566756777 Chr.5: 132658566-132658566 on Build GRCh38
rs895295697 Chr.5: 132658494-132658494 on Build GRCh38
rs200186367 Chr.5: 132658261-132658261 on Build GRCh38
rs758258492 Chr.5: 132658274-132658274 on Build GRCh38
rs201723465 Chr.5: 132658408-132658408 on Build GRCh38
rs201424420 Chr.5: 132658273-132658273 on Build GRCh38
rs1027091694 Chr.5: 132658481-132658481 on Build GRCh38
rs750926575 Chr.5: 132658147-132658147 on Build GRCh38
rs985326631 Chr.5: 132658439-132658439 on Build GRCh38
rs34255686 Chr.5: 132658339-132658339 on Build GRCh38
rs765230043 Chr.5: 132658373-132658373 on Build GRCh38
rs943472993 Chr.5: 132658449-132658449 on Build GRCh38
rs746947963 Chr.5: 132658213-132658213 on Build GRCh38
rs770488630 Chr.5: 132658217-132658217 on Build GRCh38
rs199899897 Chr.5: 132658159-132658159 on Build GRCh38
rs201121980 Chr.5: 132658206-132658206 on Build GRCh38
rs752977001 Chr.5: 132658380-132658380 on Build GRCh38
rs974128475 Chr.5: 132658238-132658238 on Build GRCh38
rs529456706 Chr.5: 132658224-132658224 on Build GRCh38
rs202050622 Chr.5: 132658220-132658220 on Build GRCh38
rs76471112 Chr.5: 132658175-132658175 on Build GRCh38
rs889168347 Chr.5: 132658186-132658186 on Build GRCh38
rs914913440 Chr.5: 132658352-132658352 on Build GRCh38
rs1018440953 Chr.5: 132658459-132658459 on Build GRCh38
rs746751863 Chr.5: 132658287-132658287 on Build GRCh38
rs200168712 Chr.5: 132658194-132658194 on Build GRCh38
rs974185468 Chr.5: 132658476-132658476 on Build GRCh38

View Full Product Details