Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181238988-181239496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3854473 c.594G>A p.V198V Chr.5: 181239109-181239109 on Build GRCh38
5868351 c.628G>A p.G210R Chr.5: 181239075-181239075 on Build GRCh38
7276515 c.525+6C>T p.? Chr.5: 181239481-181239481 on Build GRCh38
3721942 c.616C>T p.L206F Chr.5: 181239087-181239087 on Build GRCh38
1241218 c.635A>G p.K212R Chr.5: 181239068-181239068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565875576 Chr.5: 181239271-181239271 on Build GRCh38
rs764649817 Chr.5: 181239452-181239452 on Build GRCh38
rs758928876 Chr.5: 181239379-181239379 on Build GRCh38
rs781523091 Chr.5: 181239379-181239379 on Build GRCh38
rs376003338 Chr.5: 181239050-181239050 on Build GRCh38
rs182302817 Chr.5: 181239331-181239331 on Build GRCh38
rs369594764 Chr.5: 181239192-181239192 on Build GRCh38
rs755576010 Chr.5: 181239374-181239374 on Build GRCh38
rs763867674 Chr.5: 181239354-181239354 on Build GRCh38
rs11540196 Chr.5: 181239074-181239074 on Build GRCh38
rs975156882 Chr.5: 181239284-181239284 on Build GRCh38
rs79933748 Chr.5: 181239191-181239191 on Build GRCh38
rs770891403 Chr.5: 181239399-181239399 on Build GRCh38
rs774171279 Chr.5: 181239339-181239339 on Build GRCh38
rs775362656 Chr.5: 181239432-181239432 on Build GRCh38
rs769240032 Chr.5: 181239422-181239423 on Build GRCh38
rs765295307 Chr.5: 181239055-181239055 on Build GRCh38
rs751361972 Chr.5: 181239025-181239025 on Build GRCh38
rs777487890 Chr.5: 181239045-181239045 on Build GRCh38
rs543316137 Chr.5: 181239315-181239315 on Build GRCh38
rs1017966183 Chr.5: 181239242-181239242 on Build GRCh38
rs768888291 Chr.5: 181239183-181239183 on Build GRCh38
rs146971141 Chr.5: 181239351-181239351 on Build GRCh38
rs141896951 Chr.5: 181239310-181239310 on Build GRCh38
rs751331029 Chr.5: 181239106-181239106 on Build GRCh38
rs200700750 Chr.5: 181239374-181239375 on Build GRCh38
rs767419404 Chr.5: 181239345-181239345 on Build GRCh38
rs371463954 Chr.5: 181239466-181239466 on Build GRCh38
rs951236310 Chr.5: 181239268-181239268 on Build GRCh38
rs760581130 Chr.5: 181239434-181239434 on Build GRCh38
rs754736780 Chr.5: 181239112-181239112 on Build GRCh38
rs755901696 Chr.5: 181239133-181239133 on Build GRCh38
rs769445153 Chr.5: 181239038-181239038 on Build GRCh38
rs962596910 Chr.5: 181239254-181239254 on Build GRCh38
rs748825736 Chr.5: 181239145-181239145 on Build GRCh38
rs754611347 Chr.5: 181239026-181239026 on Build GRCh38
rs569641699 Chr.5: 181239240-181239240 on Build GRCh38
rs200305657 Chr.5: 181239165-181239165 on Build GRCh38
rs376487405 Chr.5: 181239163-181239163 on Build GRCh38
rs752388576 Chr.5: 181239125-181239125 on Build GRCh38
rs777429707 Chr.5: 181239377-181239380 on Build GRCh38
rs991538374 Chr.5: 181239443-181239443 on Build GRCh38
rs2287716 Chr.5: 181239276-181239276 on Build GRCh38
rs532745028 Chr.5: 181239356-181239356 on Build GRCh38
rs1017113956 Chr.5: 181239275-181239275 on Build GRCh38
rs371509460 Chr.5: 181239105-181239105 on Build GRCh38
rs397843845 Chr.5: 181239070-181239070 on Build GRCh38
rs995426115 Chr.5: 181239262-181239262 on Build GRCh38
rs781666486 Chr.5: 181239469-181239469 on Build GRCh38
rs548945137 Chr.5: 181239269-181239269 on Build GRCh38
rs112495919 Chr.5: 181239047-181239047 on Build GRCh38
rs529953435 Chr.5: 181239235-181239235 on Build GRCh38
rs202217694 Chr.5: 181239432-181239433 on Build GRCh38
rs747485823 Chr.5: 181239413-181239423 on Build GRCh38
rs189395828 Chr.5: 181239293-181239293 on Build GRCh38
rs1025446814 Chr.5: 181239267-181239267 on Build GRCh38
rs762926985 Chr.5: 181239097-181239097 on Build GRCh38
rs918441073 Chr.5: 181239023-181239023 on Build GRCh38
rs559645553 Chr.5: 181239330-181239330 on Build GRCh38
rs887399635 Chr.5: 181239470-181239470 on Build GRCh38
rs552692366 Chr.5: 181239287-181239287 on Build GRCh38
rs747443435 Chr.5: 181239366-181239366 on Build GRCh38
rs780426977 Chr.5: 181239168-181239168 on Build GRCh38
rs774527161 Chr.5: 181239416-181239416 on Build GRCh38
rs777390691 Chr.5: 181239137-181239137 on Build GRCh38
rs77347265 Chr.5: 181239049-181239049 on Build GRCh38
rs756729180 Chr.5: 181239361-181239361 on Build GRCh38
rs11540199 Chr.5: 181239114-181239114 on Build GRCh38
rs767580705 Chr.5: 181239429-181239429 on Build GRCh38
rs558117559 Chr.5: 181239381-181239381 on Build GRCh38
rs778215165 Chr.5: 181239381-181239382 on Build GRCh38
rs776849584 Chr.5: 181239053-181239053 on Build GRCh38
rs374490266 Chr.5: 181239383-181239383 on Build GRCh38
rs753003321 Chr.5: 181239379-181239380 on Build GRCh38
rs138108713 Chr.5: 181239382-181239382 on Build GRCh38
rs1039346833 Chr.5: 181239338-181239338 on Build GRCh38
rs780741578 Chr.5: 181239031-181239031 on Build GRCh38
rs373846678 Chr.5: 181239188-181239188 on Build GRCh38
rs11540189 Chr.5: 181239093-181239093 on Build GRCh38
rs867462850 Chr.5: 181239455-181239455 on Build GRCh38
rs747597266 Chr.5: 181239035-181239035 on Build GRCh38
rs368728504 Chr.5: 181239185-181239185 on Build GRCh38
rs540735589 Chr.5: 181239349-181239349 on Build GRCh38
rs41285569 Chr.5: 181239294-181239294 on Build GRCh38
rs748470910 Chr.5: 181239471-181239471 on Build GRCh38
rs778541368 Chr.5: 181239365-181239365 on Build GRCh38
rs73365992 Chr.5: 181239241-181239241 on Build GRCh38
rs780842393 Chr.5: 181239113-181239113 on Build GRCh38
rs759579473 Chr.5: 181239343-181239343 on Build GRCh38
rs114405829 Chr.5: 181239024-181239024 on Build GRCh38
rs768853411 Chr.5: 181239052-181239052 on Build GRCh38
rs763793690 Chr.5: 181239448-181239448 on Build GRCh38
rs946627414 Chr.5: 181239325-181239325 on Build GRCh38
rs568155413 Chr.5: 181239187-181239187 on Build GRCh38
rs749337757 Chr.5: 181239384-181239384 on Build GRCh38
rs750541644 Chr.5: 181239060-181239060 on Build GRCh38
rs772588134 Chr.5: 181239155-181239155 on Build GRCh38
rs748964248 Chr.5: 181239458-181239459 on Build GRCh38
rs747338652 Chr.5: 181239182-181239182 on Build GRCh38
rs377062567 Chr.5: 181239454-181239454 on Build GRCh38
rs759383736 Chr.5: 181239420-181239420 on Build GRCh38
rs143666944 Chr.5: 181239054-181239054 on Build GRCh38
rs746376973 Chr.5: 181239046-181239046 on Build GRCh38
rs866607916 Chr.5: 181239475-181239475 on Build GRCh38
rs755223375 Chr.5: 181239337-181239337 on Build GRCh38
rs529132882 Chr.5: 181239436-181239436 on Build GRCh38
rs767334605 Chr.5: 181239258-181239258 on Build GRCh38
rs534758591 Chr.5: 181239274-181239274 on Build GRCh38
rs199652309 Chr.5: 181239201-181239201 on Build GRCh38

View Full Product Details