Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44150877-44151375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCK
Entrez Gene ID
2645
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
UniGene
Hs.1270
Gene Chromosome Location
Chr.7: 44143213-44189439 on Build GRCh38
Transcripts
NM_000162.3 NM_033507.1 NM_033508.1 XM_017011966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950536 c.447C>A p.F149L Chr.7: 44150989-44150989 on Build GRCh38
1253129 c.481G>A p.D161N Chr.7: 44150961-44150961 on Build GRCh38
5734714 c.400_401delTT p.F134fs*28 Chr.7: 44151041-44151042 on Build GRCh38
232394 c.472G>A p.E158K Chr.7: 44150970-44150970 on Build GRCh38
6669961 c.390C>T p.C130C Chr.7: 44151052-44151052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978589911 Chr.7: 44151307-44151307 on Build GRCh38
rs1057194666 Chr.7: 44151064-44151064 on Build GRCh38
rs1020030261 Chr.7: 44151117-44151117 on Build GRCh38
rs193922302 Chr.7: 44150956-44150956 on Build GRCh38
rs958680608 Chr.7: 44151317-44151317 on Build GRCh38
rs767286670 Chr.7: 44151081-44151081 on Build GRCh38
rs193922295 Chr.7: 44151046-44151046 on Build GRCh38
rs749296295 Chr.7: 44150971-44150971 on Build GRCh38
rs115206967 Chr.7: 44150898-44150898 on Build GRCh38
rs765976007 Chr.7: 44150931-44150931 on Build GRCh38
rs193922296 Chr.7: 44150999-44150999 on Build GRCh38
rs540705994 Chr.7: 44151198-44151198 on Build GRCh38
rs1010584811 Chr.7: 44151235-44151235 on Build GRCh38
rs972127518 Chr.7: 44151196-44151196 on Build GRCh38
rs559377449 Chr.7: 44151199-44151199 on Build GRCh38
rs199875329 Chr.7: 44150923-44150923 on Build GRCh38
rs773281783 Chr.7: 44151004-44151004 on Build GRCh38
rs780241676 Chr.7: 44151114-44151114 on Build GRCh38
rs762419802 Chr.7: 44151045-44151045 on Build GRCh38
rs104894010 Chr.7: 44151048-44151048 on Build GRCh38
rs1019561083 Chr.7: 44150912-44150912 on Build GRCh38
rs150779253 Chr.7: 44151006-44151006 on Build GRCh38
rs999792578 Chr.7: 44151354-44151354 on Build GRCh38
rs892624195 Chr.7: 44151310-44151310 on Build GRCh38
rs911719951 Chr.7: 44151165-44151165 on Build GRCh38
rs757270147 Chr.7: 44151095-44151095 on Build GRCh38
rs759072800 Chr.7: 44151069-44151069 on Build GRCh38
rs926855799 Chr.7: 44151293-44151293 on Build GRCh38
rs775853823 Chr.7: 44150914-44150914 on Build GRCh38
rs35262363 Chr.7: 44151244-44151244 on Build GRCh38
rs541584107 Chr.7: 44151294-44151294 on Build GRCh38
rs191255582 Chr.7: 44151093-44151093 on Build GRCh38
rs764477022 Chr.7: 44151126-44151126 on Build GRCh38
rs987279901 Chr.7: 44151147-44151147 on Build GRCh38
rs74852379 Chr.7: 44150942-44150942 on Build GRCh38
rs193922300 Chr.7: 44150982-44150982 on Build GRCh38
rs537746109 Chr.7: 44151113-44151113 on Build GRCh38
rs193922299 Chr.7: 44150989-44150989 on Build GRCh38
rs1036483919 Chr.7: 44151050-44151050 on Build GRCh38
rs193922297 Chr.7: 44150990-44150990 on Build GRCh38
rs563145848 Chr.7: 44151333-44151333 on Build GRCh38
rs980787653 Chr.7: 44151252-44151252 on Build GRCh38
rs866750443 Chr.7: 44150995-44150995 on Build GRCh38
rs368929137 Chr.7: 44151170-44151170 on Build GRCh38
rs536833815 Chr.7: 44151040-44151040 on Build GRCh38
rs375966659 Chr.7: 44151116-44151116 on Build GRCh38
rs773670581 Chr.7: 44151127-44151127 on Build GRCh38
rs961945339 Chr.7: 44151194-44151194 on Build GRCh38
rs767171010 Chr.7: 44150945-44150945 on Build GRCh38
rs750519458 Chr.7: 44151111-44151111 on Build GRCh38
rs142668032 Chr.7: 44150930-44150930 on Build GRCh38
rs372213715 Chr.7: 44151101-44151101 on Build GRCh38
rs367789645 Chr.7: 44150914-44150914 on Build GRCh38
rs1045329968 Chr.7: 44151090-44151090 on Build GRCh38
rs968389257 Chr.7: 44150932-44150932 on Build GRCh38
rs151002306 Chr.7: 44150948-44150948 on Build GRCh38
rs953434805 Chr.7: 44151142-44151142 on Build GRCh38
rs112337604 Chr.7: 44151262-44151262 on Build GRCh38
rs930603664 Chr.7: 44151228-44151228 on Build GRCh38
rs372405219 Chr.7: 44151088-44151088 on Build GRCh38
rs779091112 Chr.7: 44150979-44150979 on Build GRCh38
rs777868052 Chr.7: 44150950-44150950 on Build GRCh38
rs756053003 Chr.7: 44150949-44150949 on Build GRCh38
rs193922301 Chr.7: 44150976-44150976 on Build GRCh38
rs193922293 Chr.7: 44151109-44151110 on Build GRCh38
rs761128140 Chr.7: 44150920-44150920 on Build GRCh38
rs761998125 Chr.7: 44151309-44151309 on Build GRCh38
rs754705291 Chr.7: 44150944-44150944 on Build GRCh38
rs1057036978 Chr.7: 44151110-44151110 on Build GRCh38
rs140890965 Chr.7: 44151119-44151119 on Build GRCh38
rs772650102 Chr.7: 44150926-44150926 on Build GRCh38
rs574496101 Chr.7: 44151202-44151202 on Build GRCh38
rs193922294 Chr.7: 44151122-44151123 on Build GRCh38
rs1041435517 Chr.7: 44151330-44151330 on Build GRCh38
rs747295070 Chr.7: 44151115-44151115 on Build GRCh38
rs368137186 Chr.7: 44151024-44151024 on Build GRCh38
rs746026875 Chr.7: 44150992-44150992 on Build GRCh38
rs755961174 Chr.7: 44151089-44151089 on Build GRCh38
rs879728079 Chr.7: 44151253-44151253 on Build GRCh38
rs757281286 Chr.7: 44150974-44150974 on Build GRCh38
rs367636249 Chr.7: 44151120-44151120 on Build GRCh38
rs774649114 Chr.7: 44151141-44151141 on Build GRCh38
rs992566551 Chr.7: 44151334-44151334 on Build GRCh38
rs377106269 Chr.7: 44151054-44151054 on Build GRCh38
rs549915375 Chr.7: 44151280-44151280 on Build GRCh38
rs999846681 Chr.7: 44151133-44151133 on Build GRCh38
rs945588246 Chr.7: 44151316-44151316 on Build GRCh38
rs920707738 Chr.7: 44151221-44151221 on Build GRCh38
rs139139350 Chr.7: 44151046-44151046 on Build GRCh38

View Full Product Details