Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65967505-65968010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265275 c.1586A>G p.Y529C Chr.7: 65967798-65967798 on Build GRCh38
6337082 c.1653+7A>G p.? Chr.7: 65967724-65967724 on Build GRCh38
1209225 c.1534G>A p.G512R Chr.7: 65967850-65967850 on Build GRCh38
3882052 c.1646T>C p.F549S Chr.7: 65967738-65967738 on Build GRCh38
6236796 c.1481C>T p.P494L Chr.7: 65967903-65967903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031410008 Chr.7: 65967585-65967585 on Build GRCh38
rs768292111 Chr.7: 65967731-65967731 on Build GRCh38
rs767814273 Chr.7: 65967938-65967938 on Build GRCh38
rs778095106 Chr.7: 65967713-65967713 on Build GRCh38
rs386714059 Chr.7: 65967580-65967582 on Build GRCh38
rs368053364 Chr.7: 65967764-65967764 on Build GRCh38
rs1880556 Chr.7: 65967557-65967557 on Build GRCh38
rs558826073 Chr.7: 65967565-65967565 on Build GRCh38
rs981387734 Chr.7: 65967854-65967854 on Build GRCh38
rs758492902 Chr.7: 65967791-65967793 on Build GRCh38
rs943088549 Chr.7: 65967943-65967943 on Build GRCh38
rs775509284 Chr.7: 65967948-65967948 on Build GRCh38
rs778161730 Chr.7: 65967857-65967858 on Build GRCh38
rs201361157 Chr.7: 65967931-65967931 on Build GRCh38
rs1051326180 Chr.7: 65967980-65967980 on Build GRCh38
rs747192427 Chr.7: 65967940-65967941 on Build GRCh38
rs915725855 Chr.7: 65967662-65967662 on Build GRCh38
rs779693654 Chr.7: 65967722-65967722 on Build GRCh38
rs377519272 Chr.7: 65967767-65967767 on Build GRCh38
rs752845837 Chr.7: 65967851-65967851 on Build GRCh38
rs534226705 Chr.7: 65967602-65967602 on Build GRCh38
rs766339888 Chr.7: 65967797-65967797 on Build GRCh38
rs774380038 Chr.7: 65967655-65967655 on Build GRCh38
rs760449944 Chr.7: 65967887-65967887 on Build GRCh38
rs772768221 Chr.7: 65967917-65967917 on Build GRCh38
rs794726973 Chr.7: 65967850-65967850 on Build GRCh38
rs776242697 Chr.7: 65967740-65967740 on Build GRCh38
rs1048828900 Chr.7: 65967573-65967573 on Build GRCh38
rs753886152 Chr.7: 65967969-65967969 on Build GRCh38
rs121918179 Chr.7: 65967863-65967863 on Build GRCh38
rs746721071 Chr.7: 65967730-65967730 on Build GRCh38
rs778349139 Chr.7: 65967856-65967856 on Build GRCh38
rs201928248 Chr.7: 65967857-65967857 on Build GRCh38
rs756694911 Chr.7: 65967703-65967703 on Build GRCh38
rs1880555 Chr.7: 65967580-65967580 on Build GRCh38
rs1029855609 Chr.7: 65967634-65967634 on Build GRCh38
rs548880426 Chr.7: 65967902-65967902 on Build GRCh38
rs540344035 Chr.7: 65967633-65967633 on Build GRCh38
rs777838133 Chr.7: 65967925-65967925 on Build GRCh38
rs756256226 Chr.7: 65967760-65967761 on Build GRCh38
rs767826344 Chr.7: 65967831-65967831 on Build GRCh38
rs759704615 Chr.7: 65967665-65967665 on Build GRCh38
rs121918178 Chr.7: 65967900-65967900 on Build GRCh38
rs187795195 Chr.7: 65967564-65967564 on Build GRCh38
rs527753075 Chr.7: 65967930-65967930 on Build GRCh38
rs748181124 Chr.7: 65967686-65967687 on Build GRCh38
rs546478173 Chr.7: 65967582-65967582 on Build GRCh38
rs1017273259 Chr.7: 65967568-65967568 on Build GRCh38
rs62470934 Chr.7: 65967975-65967975 on Build GRCh38
rs749695052 Chr.7: 65967714-65967714 on Build GRCh38
rs768343264 Chr.7: 65967873-65967873 on Build GRCh38
rs570476045 Chr.7: 65967959-65967959 on Build GRCh38
rs762859080 Chr.7: 65967779-65967779 on Build GRCh38
rs761204529 Chr.7: 65967749-65967749 on Build GRCh38
rs746198220 Chr.7: 65967869-65967869 on Build GRCh38
rs554682109 Chr.7: 65967595-65967595 on Build GRCh38
rs763054536 Chr.7: 65967920-65967920 on Build GRCh38
rs929372840 Chr.7: 65967721-65967721 on Build GRCh38
rs143677154 Chr.7: 65967641-65967641 on Build GRCh38
rs759880257 Chr.7: 65967823-65967823 on Build GRCh38
rs751397640 Chr.7: 65967802-65967802 on Build GRCh38
rs786205674 Chr.7: 65967798-65967798 on Build GRCh38
rs897468305 Chr.7: 65967536-65967536 on Build GRCh38
rs955769770 Chr.7: 65967590-65967590 on Build GRCh38
rs771219944 Chr.7: 65967715-65967715 on Build GRCh38
rs770302273 Chr.7: 65967556-65967556 on Build GRCh38
rs1045445639 Chr.7: 65967533-65967533 on Build GRCh38
rs546138392 Chr.7: 65967654-65967654 on Build GRCh38
rs552187538 Chr.7: 65967977-65967977 on Build GRCh38
rs759358076 Chr.7: 65967935-65967935 on Build GRCh38
rs747719992 Chr.7: 65967903-65967903 on Build GRCh38
rs929148497 Chr.7: 65967535-65967535 on Build GRCh38
rs192156180 Chr.7: 65967658-65967658 on Build GRCh38
rs143495981 Chr.7: 65967904-65967904 on Build GRCh38
rs1039860919 Chr.7: 65967958-65967958 on Build GRCh38
rs199694121 Chr.7: 65967852-65967852 on Build GRCh38
rs1018825272 Chr.7: 65967601-65967601 on Build GRCh38
rs560162692 Chr.7: 65967637-65967637 on Build GRCh38
rs973385123 Chr.7: 65967678-65967678 on Build GRCh38
rs767730315 Chr.7: 65967799-65967799 on Build GRCh38
rs998364215 Chr.7: 65967608-65967608 on Build GRCh38
rs769788461 Chr.7: 65967761-65967761 on Build GRCh38

View Full Product Details