Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22098926-22099425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_011545533.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs766318717 Chr.X: 22099258-22099259 on Build GRCh38
rs763042990 Chr.X: 22099019-22099019 on Build GRCh38
rs770491624 Chr.X: 22099081-22099081 on Build GRCh38
rs761575825 Chr.X: 22099096-22099096 on Build GRCh38
rs369359960 Chr.X: 22098978-22098978 on Build GRCh38
rs746847960 Chr.X: 22099098-22099098 on Build GRCh38
rs886041853 Chr.X: 22099013-22099013 on Build GRCh38
rs886039583 Chr.X: 22099152-22099152 on Build GRCh38
rs190284402 Chr.X: 22099208-22099208 on Build GRCh38
rs1052355917 Chr.X: 22099346-22099346 on Build GRCh38
rs757539874 Chr.X: 22099009-22099009 on Build GRCh38
rs766310950 Chr.X: 22099020-22099020 on Build GRCh38
rs375062555 Chr.X: 22098968-22098968 on Build GRCh38
rs371953854 Chr.X: 22098967-22098967 on Build GRCh38
rs775973349 Chr.X: 22099080-22099080 on Build GRCh38
rs909020299 Chr.X: 22099210-22099210 on Build GRCh38
rs766616716 Chr.X: 22099134-22099134 on Build GRCh38
rs182865564 Chr.X: 22099256-22099256 on Build GRCh38
rs5951691 Chr.X: 22098970-22098970 on Build GRCh38
rs768400503 Chr.X: 22099099-22099099 on Build GRCh38
rs760451270 Chr.X: 22099008-22099008 on Build GRCh38
rs916341020 Chr.X: 22099254-22099254 on Build GRCh38
rs930073324 Chr.X: 22099190-22099190 on Build GRCh38
rs185862803 Chr.X: 22099151-22099151 on Build GRCh38
rs763094620 Chr.X: 22099117-22099117 on Build GRCh38
rs765422660 Chr.X: 22099291-22099291 on Build GRCh38
rs866640200 Chr.X: 22099103-22099103 on Build GRCh38
rs373261521 Chr.X: 22099029-22099029 on Build GRCh38
rs773436322 Chr.X: 22099114-22099114 on Build GRCh38
rs759634976 Chr.X: 22099158-22099158 on Build GRCh38
rs376461141 Chr.X: 22099097-22099097 on Build GRCh38
rs775216821 Chr.X: 22099002-22099002 on Build GRCh38
rs1021660256 Chr.X: 22099011-22099011 on Build GRCh38
rs201383931 Chr.X: 22099054-22099054 on Build GRCh38
rs774372335 Chr.X: 22098976-22098976 on Build GRCh38
rs745382329 Chr.X: 22099093-22099093 on Build GRCh38
rs751465972 Chr.X: 22099321-22099321 on Build GRCh38
rs3833907 Chr.X: 22099263-22099264 on Build GRCh38
rs1057517792 Chr.X: 22099118-22099119 on Build GRCh38
rs756512186 Chr.X: 22099061-22099061 on Build GRCh38
rs1032503119 Chr.X: 22098969-22098969 on Build GRCh38
rs750613071 Chr.X: 22099017-22099017 on Build GRCh38
rs149793221 Chr.X: 22099086-22099086 on Build GRCh38
rs146073934 Chr.X: 22099179-22099179 on Build GRCh38
rs771907880 Chr.X: 22099000-22099000 on Build GRCh38
rs774349246 Chr.X: 22099149-22099149 on Build GRCh38
rs781713539 Chr.X: 22099045-22099045 on Build GRCh38
rs1046680131 Chr.X: 22099281-22099281 on Build GRCh38
rs139749399 Chr.X: 22099068-22099068 on Build GRCh38
rs199965602 Chr.X: 22099043-22099043 on Build GRCh38
rs777541394 Chr.X: 22098965-22098965 on Build GRCh38
rs45541842 Chr.X: 22099364-22099364 on Build GRCh38

View Full Product Details