Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8121193-8121728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES7
Entrez Gene ID
84667
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
SCDO4, bHLHb37
UniGene
Hs.434828
Gene Chromosome Location
Chr.17: 8120590-8126032 on Build GRCh38
Transcripts
NM_001165967.1 NM_032580.3 XM_011524041.2 XM_017025232.1

SNP Detail

SNP ID Chromosome Location
rs541012018 Chr.17: 8121425-8121425 on Build GRCh38
rs966299468 Chr.17: 8121525-8121525 on Build GRCh38
rs911422224 Chr.17: 8121649-8121649 on Build GRCh38
rs985652294 Chr.17: 8121625-8121625 on Build GRCh38
rs1036854618 Chr.17: 8121577-8121577 on Build GRCh38
rs751740177 Chr.17: 8121457-8121457 on Build GRCh38
rs888207014 Chr.17: 8121373-8121373 on Build GRCh38
rs1024243984 Chr.17: 8121453-8121453 on Build GRCh38
rs1014616261 Chr.17: 8121446-8121446 on Build GRCh38
rs978268609 Chr.17: 8121563-8121563 on Build GRCh38
rs559461603 Chr.17: 8121455-8121455 on Build GRCh38
rs536990518 Chr.17: 8121602-8121602 on Build GRCh38
rs1005351109 Chr.17: 8121414-8121414 on Build GRCh38
rs531263972 Chr.17: 8121436-8121436 on Build GRCh38
rs550425522 Chr.17: 8121707-8121707 on Build GRCh38
rs1001485691 Chr.17: 8121639-8121639 on Build GRCh38
rs896956362 Chr.17: 8121582-8121582 on Build GRCh38
rs922393458 Chr.17: 8121589-8121589 on Build GRCh38
rs1038368865 Chr.17: 8121677-8121677 on Build GRCh38
rs1047540918 Chr.17: 8121478-8121478 on Build GRCh38
rs1010298012 Chr.17: 8121681-8121681 on Build GRCh38
rs897238266 Chr.17: 8121445-8121445 on Build GRCh38
rs773751563 Chr.17: 8121277-8121277 on Build GRCh38
rs890054537 Chr.17: 8121234-8121234 on Build GRCh38
rs571199056 Chr.17: 8121438-8121438 on Build GRCh38
rs1020899015 Chr.17: 8121524-8121524 on Build GRCh38
rs552824640 Chr.17: 8121437-8121437 on Build GRCh38
rs957489418 Chr.17: 8121517-8121517 on Build GRCh38
rs8076629 Chr.17: 8121490-8121490 on Build GRCh38
rs1033980399 Chr.17: 8121511-8121511 on Build GRCh38
rs535914789 Chr.17: 8121559-8121559 on Build GRCh38
rs966418854 Chr.17: 8121561-8121561 on Build GRCh38
rs565453136 Chr.17: 8121536-8121536 on Build GRCh38
rs1001497070 Chr.17: 8121485-8121485 on Build GRCh38
rs955697892 Chr.17: 8121623-8121623 on Build GRCh38
rs1003392417 Chr.17: 8121509-8121509 on Build GRCh38
rs929981617 Chr.17: 8121697-8121697 on Build GRCh38
rs922223596 Chr.17: 8121551-8121551 on Build GRCh38
rs905790545 Chr.17: 8121628-8121628 on Build GRCh38
rs941306526 Chr.17: 8121399-8121399 on Build GRCh38
rs1049478935 Chr.17: 8121359-8121359 on Build GRCh38
rs575817248 Chr.17: 8121583-8121583 on Build GRCh38
rs988929677 Chr.17: 8121518-8121518 on Build GRCh38
rs905830986 Chr.17: 8121465-8121465 on Build GRCh38
rs932326227 Chr.17: 8121249-8121249 on Build GRCh38
rs387906978 Chr.17: 8121693-8121693 on Build GRCh38
rs1010525340 Chr.17: 8121541-8121541 on Build GRCh38
rs759681801 Chr.17: 8121685-8121685 on Build GRCh38
rs766566795 Chr.17: 8121456-8121456 on Build GRCh38
rs558811781 Chr.17: 8121673-8121673 on Build GRCh38
rs114049508 Chr.17: 8121440-8121440 on Build GRCh38
rs8076664 Chr.17: 8121560-8121560 on Build GRCh38

View Full Product Details