Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31269351-31269882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-C
Entrez Gene ID
3107
Gene Name
major histocompatibility complex, class I, C
Gene Aliases
D6S204, HLA-JY3, HLAC, HLC-C, MHC, PSORS1
UniGene
Hs.656020
Gene Chromosome Location
Chr.6: 31268749-31272136 on Build GRCh38
Transcripts
NM_001243042.1 NM_002117.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352838 c.1043C>T p.A348V Chr.6: 31269498-31269498 on Build GRCh38
4160664 c.1049G>C p.C350S Chr.6: 31269385-31269385 on Build GRCh38
7352839 c.1154C>T p.A385V Chr.6: 31269498-31269498 on Build GRCh38
6173671 c.1093G>A p.E365K Chr.6: 31269359-31269359 on Build GRCh38
6173670 c.1186G>A p.E396K Chr.6: 31269359-31269359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770655323 Chr.6: 31269400-31269400 on Build GRCh38
rs68094471 Chr.6: 31269546-31269546 on Build GRCh38
rs748648607 Chr.6: 31269566-31269566 on Build GRCh38
rs765405213 Chr.6: 31269586-31269586 on Build GRCh38
rs72558157 Chr.6: 31269794-31269794 on Build GRCh38
rs281860590 Chr.6: 31269407-31269407 on Build GRCh38
rs281860585 Chr.6: 31269808-31269808 on Build GRCh38
rs68037221 Chr.6: 31269672-31269672 on Build GRCh38
rs777991288 Chr.6: 31269397-31269398 on Build GRCh38
rs200405836 Chr.6: 31269751-31269751 on Build GRCh38
rs750626667 Chr.6: 31269450-31269450 on Build GRCh38
rs45473495 Chr.6: 31269849-31269849 on Build GRCh38
rs988161108 Chr.6: 31269493-31269493 on Build GRCh38
rs527842025 Chr.6: 31269440-31269440 on Build GRCh38
rs45437892 Chr.6: 31269765-31269765 on Build GRCh38
rs72558161 Chr.6: 31269680-31269680 on Build GRCh38
rs767661877 Chr.6: 31269435-31269435 on Build GRCh38
rs386698957 Chr.6: 31269576-31269577 on Build GRCh38
rs9264596 Chr.6: 31269453-31269453 on Build GRCh38
rs1055337554 Chr.6: 31269391-31269391 on Build GRCh38
rs386698959 Chr.6: 31269684-31269686 on Build GRCh38
rs567530604 Chr.6: 31269535-31269535 on Build GRCh38
rs41552718 Chr.6: 31269482-31269482 on Build GRCh38
rs3177824 Chr.6: 31269492-31269492 on Build GRCh38
rs1015598888 Chr.6: 31269694-31269694 on Build GRCh38
rs67827555 Chr.6: 31269576-31269576 on Build GRCh38
rs9264607 Chr.6: 31269828-31269828 on Build GRCh38
rs9264603 Chr.6: 31269661-31269661 on Build GRCh38
rs755677995 Chr.6: 31269545-31269545 on Build GRCh38
rs776451724 Chr.6: 31269405-31269405 on Build GRCh38
rs66620546 Chr.6: 31269686-31269686 on Build GRCh38
rs767852739 Chr.6: 31269481-31269481 on Build GRCh38
rs56010430 Chr.6: 31269628-31269628 on Build GRCh38
rs893840148 Chr.6: 31269837-31269837 on Build GRCh38
rs41550315 Chr.6: 31269487-31269487 on Build GRCh38
rs72558163 Chr.6: 31269565-31269565 on Build GRCh38
rs981934687 Chr.6: 31269812-31269812 on Build GRCh38
rs66459704 Chr.6: 31269660-31269660 on Build GRCh38
rs984501849 Chr.6: 31269638-31269638 on Build GRCh38
rs35708511 Chr.6: 31269385-31269385 on Build GRCh38
rs370407485 Chr.6: 31269431-31269431 on Build GRCh38
rs281860591 Chr.6: 31269398-31269398 on Build GRCh38
rs3819287 Chr.6: 31269786-31269786 on Build GRCh38
rs777872223 Chr.6: 31269458-31269458 on Build GRCh38
rs981317281 Chr.6: 31269747-31269747 on Build GRCh38
rs9264601 Chr.6: 31269577-31269577 on Build GRCh38
rs781101853 Chr.6: 31269469-31269469 on Build GRCh38
rs76294545 Chr.6: 31269507-31269507 on Build GRCh38
rs996234154 Chr.6: 31269591-31269591 on Build GRCh38
rs139702282 Chr.6: 31269495-31269495 on Build GRCh38
rs543182678 Chr.6: 31269603-31269603 on Build GRCh38
rs879632567 Chr.6: 31269497-31269497 on Build GRCh38
rs1046785428 Chr.6: 31269606-31269606 on Build GRCh38
rs2523609 Chr.6: 31269478-31269478 on Build GRCh38
rs72558162 Chr.6: 31269580-31269580 on Build GRCh38
rs17881458 Chr.6: 31269460-31269460 on Build GRCh38
rs9279069 Chr.6: 31269799-31269800 on Build GRCh38
rs895081373 Chr.6: 31269402-31269402 on Build GRCh38
rs776913998 Chr.6: 31269584-31269584 on Build GRCh38
rs751872755 Chr.6: 31269426-31269426 on Build GRCh38
rs751554648 Chr.6: 31269506-31269506 on Build GRCh38
rs61759946 Chr.6: 31269382-31269382 on Build GRCh38
rs757120520 Chr.6: 31269468-31269468 on Build GRCh38
rs45618340 Chr.6: 31269853-31269853 on Build GRCh38
rs17879162 Chr.6: 31269480-31269480 on Build GRCh38
rs41541318 Chr.6: 31269859-31269859 on Build GRCh38
rs281860589 Chr.6: 31269423-31269423 on Build GRCh38
rs9264597 Chr.6: 31269456-31269456 on Build GRCh38
rs72558159 Chr.6: 31269736-31269736 on Build GRCh38
rs746845038 Chr.6: 31269399-31269399 on Build GRCh38
rs769529783 Chr.6: 31269474-31269474 on Build GRCh38
rs1053614364 Chr.6: 31269832-31269832 on Build GRCh38
rs372183108 Chr.6: 31269801-31269802 on Build GRCh38
rs9264606 Chr.6: 31269815-31269815 on Build GRCh38
rs45508494 Chr.6: 31269809-31269809 on Build GRCh38
rs748818590 Chr.6: 31269475-31269475 on Build GRCh38
rs928524570 Chr.6: 31269770-31269770 on Build GRCh38
rs772202592 Chr.6: 31269392-31269392 on Build GRCh38
rs281860586 Chr.6: 31269807-31269807 on Build GRCh38
rs753990733 Chr.6: 31269454-31269454 on Build GRCh38
rs45492599 Chr.6: 31269768-31269768 on Build GRCh38
rs72502556 Chr.6: 31269684-31269684 on Build GRCh38
rs9264602 Chr.6: 31269629-31269629 on Build GRCh38
rs545140350 Chr.6: 31269416-31269416 on Build GRCh38
rs386698958 Chr.6: 31269660-31269661 on Build GRCh38
rs542273765 Chr.6: 31269422-31269422 on Build GRCh38
rs769176621 Chr.6: 31269413-31269413 on Build GRCh38
rs9281299 Chr.6: 31269750-31269751 on Build GRCh38
rs550578214 Chr.6: 31269473-31269473 on Build GRCh38
rs868510131 Chr.6: 31269488-31269488 on Build GRCh38
rs111308207 Chr.6: 31269491-31269491 on Build GRCh38
rs45498195 Chr.6: 31269777-31269777 on Build GRCh38
rs748730545 Chr.6: 31269550-31269550 on Build GRCh38
rs907750566 Chr.6: 31269394-31269394 on Build GRCh38
rs750476831 Chr.6: 31269377-31269377 on Build GRCh38
rs75141084 Chr.6: 31269749-31269750 on Build GRCh38
rs781416483 Chr.6: 31269376-31269376 on Build GRCh38
rs41562921 Chr.6: 31269477-31269477 on Build GRCh38
rs937308184 Chr.6: 31269774-31269774 on Build GRCh38
rs999492224 Chr.6: 31269654-31269654 on Build GRCh38
rs999710500 Chr.6: 31269509-31269509 on Build GRCh38
rs281860587 Chr.6: 31269799-31269800 on Build GRCh38
rs568718965 Chr.6: 31269512-31269512 on Build GRCh38
rs41559915 Chr.6: 31269498-31269498 on Build GRCh38
rs66772001 Chr.6: 31269556-31269556 on Build GRCh38
rs537747274 Chr.6: 31269788-31269788 on Build GRCh38
rs373826500 Chr.6: 31269807-31269808 on Build GRCh38
rs386698956 Chr.6: 31269478-31269480 on Build GRCh38
rs76798670 Chr.6: 31269455-31269455 on Build GRCh38
rs754110840 Chr.6: 31269485-31269485 on Build GRCh38
rs281860588 Chr.6: 31269494-31269494 on Build GRCh38

View Full Product Details