Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108867683-108868224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
UniGene
Hs.494738
Gene Chromosome Location
Chr.9: 108867520-108934332 on Build GRCh38
Transcripts
NM_001318360.1 NM_003640.4 XM_011519136.1 XM_011519137.1

SNP Detail

SNP ID Chromosome Location
rs140005128 Chr.9: 108867842-108867842 on Build GRCh38
rs958727998 Chr.9: 108867840-108867840 on Build GRCh38
rs564290288 Chr.9: 108867746-108867746 on Build GRCh38
rs914551358 Chr.9: 108868138-108868138 on Build GRCh38
rs1020176903 Chr.9: 108867972-108867972 on Build GRCh38
rs900568479 Chr.9: 108867885-108867885 on Build GRCh38
rs189897190 Chr.9: 108867834-108867834 on Build GRCh38
rs908107439 Chr.9: 108868055-108868055 on Build GRCh38
rs781024809 Chr.9: 108868195-108868195 on Build GRCh38
rs969586518 Chr.9: 108867736-108867737 on Build GRCh38
rs386737465 Chr.9: 108867855-108867856 on Build GRCh38
rs1042139385 Chr.9: 108868197-108868197 on Build GRCh38
rs550189912 Chr.9: 108867818-108867818 on Build GRCh38
rs769032654 Chr.9: 108868178-108868178 on Build GRCh38
rs886063344 Chr.9: 108867763-108867763 on Build GRCh38
rs537888003 Chr.9: 108867838-108867838 on Build GRCh38
rs13299651 Chr.9: 108867855-108867855 on Build GRCh38
rs756637677 Chr.9: 108868031-108868031 on Build GRCh38
rs932034582 Chr.9: 108868117-108868117 on Build GRCh38
rs559551917 Chr.9: 108868053-108868053 on Build GRCh38
rs13299652 Chr.9: 108867856-108867856 on Build GRCh38
rs987535417 Chr.9: 108867985-108867985 on Build GRCh38
rs574030921 Chr.9: 108867949-108867949 on Build GRCh38
rs143906434 Chr.9: 108867901-108867901 on Build GRCh38
rs929226044 Chr.9: 108868066-108868066 on Build GRCh38
rs41305457 Chr.9: 108868044-108868044 on Build GRCh38
rs954794186 Chr.9: 108867946-108867946 on Build GRCh38
rs895450123 Chr.9: 108868112-108868112 on Build GRCh38
rs539404183 Chr.9: 108867924-108867924 on Build GRCh38
rs76994656 Chr.9: 108867868-108867868 on Build GRCh38
rs777849094 Chr.9: 108867800-108867800 on Build GRCh38
rs746907451 Chr.9: 108867991-108867991 on Build GRCh38
rs1044117179 Chr.9: 108868159-108868159 on Build GRCh38
rs1051568111 Chr.9: 108867820-108867820 on Build GRCh38
rs922129728 Chr.9: 108868092-108868092 on Build GRCh38
rs933409985 Chr.9: 108867813-108867813 on Build GRCh38
rs539476728 Chr.9: 108868138-108868139 on Build GRCh38
rs944908633 Chr.9: 108867872-108867872 on Build GRCh38
rs958533883 Chr.9: 108867936-108867936 on Build GRCh38
rs1010069999 Chr.9: 108867967-108867967 on Build GRCh38
rs1010144022 Chr.9: 108867789-108867789 on Build GRCh38
rs749719194 Chr.9: 108868124-108868124 on Build GRCh38
rs913905319 Chr.9: 108867944-108867944 on Build GRCh38
rs1026615138 Chr.9: 108868045-108868045 on Build GRCh38
rs936677116 Chr.9: 108867786-108867786 on Build GRCh38
rs1054473445 Chr.9: 108868122-108868122 on Build GRCh38
rs577931217 Chr.9: 108868061-108868061 on Build GRCh38
rs932099681 Chr.9: 108867889-108867889 on Build GRCh38
rs879924643 Chr.9: 108867861-108867861 on Build GRCh38
rs1054379768 Chr.9: 108867898-108867898 on Build GRCh38
rs996181236 Chr.9: 108867736-108867736 on Build GRCh38
rs971258024 Chr.9: 108867824-108867824 on Build GRCh38
rs1032592090 Chr.9: 108867931-108867931 on Build GRCh38
rs1025109186 Chr.9: 108867726-108867726 on Build GRCh38
rs565942455 Chr.9: 108867846-108867846 on Build GRCh38
rs976276357 Chr.9: 108868089-108868089 on Build GRCh38
rs560485412 Chr.9: 108867860-108867860 on Build GRCh38
rs951027770 Chr.9: 108868051-108868051 on Build GRCh38
rs66608307 Chr.9: 108867856-108867856 on Build GRCh38
rs1040975275 Chr.9: 108867875-108867875 on Build GRCh38
rs1034619639 Chr.9: 108868041-108868041 on Build GRCh38
rs893053253 Chr.9: 108867787-108867787 on Build GRCh38
rs4978371 Chr.9: 108867764-108867764 on Build GRCh38
rs988126392 Chr.9: 108867940-108867940 on Build GRCh38
rs913950140 Chr.9: 108867807-108867807 on Build GRCh38
rs958479111 Chr.9: 108868043-108868043 on Build GRCh38
rs907994989 Chr.9: 108867871-108867871 on Build GRCh38

View Full Product Details