Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73151783-73152295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2

SNP Detail

SNP ID Chromosome Location
rs147968642 Chr.9: 73151835-73151835 on Build GRCh38
rs1035416906 Chr.9: 73152229-73152229 on Build GRCh38
rs1050890262 Chr.9: 73151945-73151945 on Build GRCh38
rs182007166 Chr.9: 73151916-73151916 on Build GRCh38
rs763469951 Chr.9: 73151950-73151950 on Build GRCh38
rs117520090 Chr.9: 73152151-73152151 on Build GRCh38
rs1131072 Chr.9: 73151922-73151922 on Build GRCh38
rs772490050 Chr.9: 73151849-73151849 on Build GRCh38
rs573646499 Chr.9: 73152015-73152015 on Build GRCh38
rs1035363767 Chr.9: 73151898-73151898 on Build GRCh38
rs769267299 Chr.9: 73151987-73151987 on Build GRCh38
rs1054948062 Chr.9: 73152107-73152107 on Build GRCh38
rs995864037 Chr.9: 73152070-73152070 on Build GRCh38
rs1013699837 Chr.9: 73151820-73151820 on Build GRCh38
rs964810389 Chr.9: 73151880-73151880 on Build GRCh38
rs914228137 Chr.9: 73151938-73151938 on Build GRCh38
rs1025525993 Chr.9: 73152108-73152108 on Build GRCh38
rs1014059078 Chr.9: 73152215-73152215 on Build GRCh38
rs546144543 Chr.9: 73151866-73151866 on Build GRCh38
rs187211571 Chr.9: 73152041-73152041 on Build GRCh38
rs950737587 Chr.9: 73152242-73152242 on Build GRCh38
rs1043632953 Chr.9: 73151964-73151964 on Build GRCh38
rs960771819 Chr.9: 73151808-73151808 on Build GRCh38
rs529145380 Chr.9: 73151935-73151935 on Build GRCh38
rs555744050 Chr.9: 73152196-73152196 on Build GRCh38
rs935448947 Chr.9: 73152006-73152006 on Build GRCh38
rs190445887 Chr.9: 73152261-73152261 on Build GRCh38
rs542605716 Chr.9: 73151956-73151956 on Build GRCh38
rs923476755 Chr.9: 73151908-73151908 on Build GRCh38
rs78330792 Chr.9: 73151959-73151959 on Build GRCh38
rs891189806 Chr.9: 73152024-73152024 on Build GRCh38
rs369263411 Chr.9: 73152235-73152235 on Build GRCh38
rs945067591 Chr.9: 73151960-73151960 on Build GRCh38
rs909476953 Chr.9: 73151877-73151877 on Build GRCh38
rs542275125 Chr.9: 73152187-73152187 on Build GRCh38
rs974881240 Chr.9: 73151891-73151891 on Build GRCh38
rs958382940 Chr.9: 73151905-73151905 on Build GRCh38
rs898493985 Chr.9: 73152039-73152039 on Build GRCh38
rs1002637297 Chr.9: 73152177-73152177 on Build GRCh38
rs1040132646 Chr.9: 73152036-73152036 on Build GRCh38
rs1025938426 Chr.9: 73151811-73151811 on Build GRCh38
rs994544240 Chr.9: 73151975-73151975 on Build GRCh38
rs761442753 Chr.9: 73152034-73152034 on Build GRCh38
rs553880438 Chr.9: 73151946-73151946 on Build GRCh38
rs79551665 Chr.9: 73152241-73152241 on Build GRCh38

View Full Product Details