Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73158389-73158900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2 XM_011518609.1 XM_017014657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5023392 c.37_49del13 p.F13fs*9 Chr.9: 73158572-73158584 on Build GRCh38
6013496 c.146C>T p.P49L Chr.9: 73158741-73158741 on Build GRCh38
4959824 c.160G>A p.A54T Chr.9: 73158788-73158788 on Build GRCh38
4903239 c.49G>A p.E17K Chr.9: 73158584-73158584 on Build GRCh38
7182611 c.130C>T p.P44S Chr.9: 73158758-73158758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554203401 Chr.9: 73158442-73158442 on Build GRCh38
rs563971978 Chr.9: 73158562-73158562 on Build GRCh38
rs537232568 Chr.9: 73158433-73158433 on Build GRCh38
rs770284358 Chr.9: 73158620-73158620 on Build GRCh38
rs148187721 Chr.9: 73158766-73158766 on Build GRCh38
rs768486386 Chr.9: 73158774-73158774 on Build GRCh38
rs764812476 Chr.9: 73158836-73158836 on Build GRCh38
rs202158567 Chr.9: 73158487-73158487 on Build GRCh38
rs190143359 Chr.9: 73158838-73158838 on Build GRCh38
rs536707670 Chr.9: 73158481-73158481 on Build GRCh38
rs756453145 Chr.9: 73158831-73158831 on Build GRCh38
rs11557052 Chr.9: 73158755-73158755 on Build GRCh38
rs368717717 Chr.9: 73158757-73158757 on Build GRCh38
rs148532111 Chr.9: 73158694-73158694 on Build GRCh38
rs757423622 Chr.9: 73158656-73158656 on Build GRCh38
rs556547962 Chr.9: 73158839-73158839 on Build GRCh38
rs760100719 Chr.9: 73158809-73158809 on Build GRCh38
rs11557058 Chr.9: 73158771-73158771 on Build GRCh38
rs534541502 Chr.9: 73158415-73158415 on Build GRCh38
rs780540903 Chr.9: 73158596-73158596 on Build GRCh38
rs753992010 Chr.9: 73158652-73158652 on Build GRCh38
rs755464124 Chr.9: 73158754-73158754 on Build GRCh38
rs750986401 Chr.9: 73158661-73158661 on Build GRCh38
rs1043798068 Chr.9: 73158869-73158869 on Build GRCh38
rs769598511 Chr.9: 73158608-73158608 on Build GRCh38
rs780663466 Chr.9: 73158668-73158668 on Build GRCh38
rs761625248 Chr.9: 73158778-73158778 on Build GRCh38
rs753950463 Chr.9: 73158542-73158542 on Build GRCh38
rs756308938 Chr.9: 73158523-73158523 on Build GRCh38
rs200500450 Chr.9: 73158724-73158724 on Build GRCh38
rs751990285 Chr.9: 73158670-73158670 on Build GRCh38
rs374463316 Chr.9: 73158494-73158494 on Build GRCh38
rs916745255 Chr.9: 73158834-73158834 on Build GRCh38
rs767002428 Chr.9: 73158743-73158743 on Build GRCh38
rs571956937 Chr.9: 73158414-73158414 on Build GRCh38
rs373357116 Chr.9: 73158760-73158760 on Build GRCh38
rs1006343430 Chr.9: 73158690-73158690 on Build GRCh38
rs199772708 Chr.9: 73158727-73158727 on Build GRCh38
rs928127817 Chr.9: 73158444-73158444 on Build GRCh38
rs371854581 Chr.9: 73158558-73158558 on Build GRCh38
rs576441840 Chr.9: 73158851-73158851 on Build GRCh38
rs142849845 Chr.9: 73158725-73158725 on Build GRCh38
rs534445509 Chr.9: 73158761-73158761 on Build GRCh38
rs540141537 Chr.9: 73158641-73158641 on Build GRCh38
rs113684866 Chr.9: 73158602-73158602 on Build GRCh38
rs1043985644 Chr.9: 73158459-73158459 on Build GRCh38
rs200105739 Chr.9: 73158808-73158808 on Build GRCh38
rs768036900 Chr.9: 73158818-73158818 on Build GRCh38
rs367968614 Chr.9: 73158649-73158649 on Build GRCh38
rs571918496 Chr.9: 73158736-73158736 on Build GRCh38
rs573952148 Chr.9: 73158448-73158448 on Build GRCh38
rs766566059 Chr.9: 73158807-73158807 on Build GRCh38
rs1050986430 Chr.9: 73158666-73158666 on Build GRCh38
rs774101805 Chr.9: 73158497-73158497 on Build GRCh38
rs376171580 Chr.9: 73158732-73158732 on Build GRCh38
rs983690277 Chr.9: 73158431-73158431 on Build GRCh38
rs748825404 Chr.9: 73158612-73158612 on Build GRCh38
rs770534471 Chr.9: 73158699-73158699 on Build GRCh38
rs141175221 Chr.9: 73158791-73158791 on Build GRCh38
rs185723500 Chr.9: 73158499-73158499 on Build GRCh38
rs764200392 Chr.9: 73158541-73158541 on Build GRCh38
rs767215706 Chr.9: 73158506-73158506 on Build GRCh38
rs142511024 Chr.9: 73158773-73158773 on Build GRCh38
rs781682499 Chr.9: 73158756-73158756 on Build GRCh38
rs768435046 Chr.9: 73158730-73158730 on Build GRCh38
rs752976956 Chr.9: 73158519-73158519 on Build GRCh38
rs867127271 Chr.9: 73158767-73158767 on Build GRCh38
rs776256101 Chr.9: 73158775-73158775 on Build GRCh38
rs748890727 Chr.9: 73158697-73158697 on Build GRCh38
rs772814437 Chr.9: 73158492-73158492 on Build GRCh38
rs779015140 Chr.9: 73158526-73158527 on Build GRCh38
rs575367154 Chr.9: 73158493-73158493 on Build GRCh38
rs753037155 Chr.9: 73158829-73158829 on Build GRCh38
rs868517148 Chr.9: 73158584-73158584 on Build GRCh38
rs11557056 Chr.9: 73158590-73158590 on Build GRCh38
rs377206672 Chr.9: 73158765-73158765 on Build GRCh38
rs763037344 Chr.9: 73158741-73158741 on Build GRCh38
rs991026489 Chr.9: 73158823-73158823 on Build GRCh38
rs928210855 Chr.9: 73158797-73158797 on Build GRCh38
rs1055629791 Chr.9: 73158460-73158460 on Build GRCh38
rs755341198 Chr.9: 73158682-73158682 on Build GRCh38
rs758853777 Chr.9: 73158589-73158589 on Build GRCh38
rs759381580 Chr.9: 73158628-73158628 on Build GRCh38
rs1013751305 Chr.9: 73158632-73158632 on Build GRCh38
rs773421579 Chr.9: 73158795-73158795 on Build GRCh38
rs758797231 Chr.9: 73158662-73158662 on Build GRCh38
rs778266723 Chr.9: 73158716-73158716 on Build GRCh38
rs780032916 Chr.9: 73158772-73158772 on Build GRCh38
rs867179177 Chr.9: 73158855-73158855 on Build GRCh38
rs201041541 Chr.9: 73158611-73158611 on Build GRCh38
rs375641852 Chr.9: 73158640-73158640 on Build GRCh38
rs762950307 Chr.9: 73158799-73158799 on Build GRCh38
rs774417259 Chr.9: 73158627-73158627 on Build GRCh38
rs779515245 Chr.9: 73158853-73158853 on Build GRCh38
rs757226094 Chr.9: 73158549-73158549 on Build GRCh38
rs35304764 Chr.9: 73158733-73158733 on Build GRCh38
rs528896897 Chr.9: 73158659-73158659 on Build GRCh38
rs1055623365 Chr.9: 73158876-73158876 on Build GRCh38

View Full Product Details