Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73167358-73167872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2 XM_011518609.1 XM_017014657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
754258 c.838G>T p.E280* Chr.9: 73167532-73167532 on Build GRCh38
6582181 c.847C>T p.H283Y Chr.9: 73167541-73167541 on Build GRCh38
1462936 c.831C>A p.F277L Chr.9: 73167525-73167525 on Build GRCh38
28372 c.831C>T p.F277F Chr.9: 73167525-73167525 on Build GRCh38
4923039 c.816A>T p.T272T Chr.9: 73167510-73167510 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770628167 Chr.9: 73167453-73167453 on Build GRCh38
rs866942919 Chr.9: 73167436-73167436 on Build GRCh38
rs758683604 Chr.9: 73167635-73167635 on Build GRCh38
rs541971115 Chr.9: 73167460-73167460 on Build GRCh38
rs541915852 Chr.9: 73167437-73167437 on Build GRCh38
rs1981288 Chr.9: 73167739-73167739 on Build GRCh38
rs369473616 Chr.9: 73167477-73167477 on Build GRCh38
rs930678777 Chr.9: 73167443-73167443 on Build GRCh38
rs775172056 Chr.9: 73167580-73167580 on Build GRCh38
rs961456379 Chr.9: 73167648-73167648 on Build GRCh38
rs757299304 Chr.9: 73167583-73167583 on Build GRCh38
rs758692597 Chr.9: 73167594-73167594 on Build GRCh38
rs910005851 Chr.9: 73167383-73167383 on Build GRCh38
rs764084381 Chr.9: 73167486-73167486 on Build GRCh38
rs972736928 Chr.9: 73167406-73167406 on Build GRCh38
rs901580971 Chr.9: 73167839-73167839 on Build GRCh38
rs765559979 Chr.9: 73167494-73167494 on Build GRCh38
rs781568482 Chr.9: 73167821-73167821 on Build GRCh38
rs748513926 Chr.9: 73167559-73167559 on Build GRCh38
rs750743071 Chr.9: 73167626-73167626 on Build GRCh38
rs760215149 Chr.9: 73167582-73167582 on Build GRCh38
rs982156255 Chr.9: 73167599-73167599 on Build GRCh38
rs766261103 Chr.9: 73167451-73167451 on Build GRCh38
rs552147642 Chr.9: 73167598-73167598 on Build GRCh38
rs202142251 Chr.9: 73167470-73167470 on Build GRCh38
rs191023973 Chr.9: 73167442-73167442 on Build GRCh38
rs917264868 Chr.9: 73167670-73167670 on Build GRCh38
rs747350047 Chr.9: 73167678-73167678 on Build GRCh38
rs200843894 Chr.9: 73167531-73167531 on Build GRCh38
rs961915428 Chr.9: 73167609-73167609 on Build GRCh38
rs969194465 Chr.9: 73167597-73167597 on Build GRCh38
rs954316437 Chr.9: 73167613-73167613 on Build GRCh38
rs368918336 Chr.9: 73167574-73167574 on Build GRCh38
rs760658757 Chr.9: 73167480-73167480 on Build GRCh38
rs371942094 Chr.9: 73167468-73167468 on Build GRCh38
rs972787563 Chr.9: 73167651-73167651 on Build GRCh38
rs745488481 Chr.9: 73167577-73167577 on Build GRCh38
rs750696979 Chr.9: 73167606-73167606 on Build GRCh38
rs376537013 Chr.9: 73167504-73167504 on Build GRCh38
rs79581073 Chr.9: 73167822-73167822 on Build GRCh38
rs1342017 Chr.9: 73167594-73167594 on Build GRCh38
rs1013483429 Chr.9: 73167762-73167762 on Build GRCh38
rs893314851 Chr.9: 73167557-73167557 on Build GRCh38
rs944824171 Chr.9: 73167687-73167687 on Build GRCh38
rs375251312 Chr.9: 73167491-73167491 on Build GRCh38
rs927857421 Chr.9: 73167398-73167398 on Build GRCh38
rs372722753 Chr.9: 73167561-73167561 on Build GRCh38
rs141810071 Chr.9: 73167505-73167505 on Build GRCh38
rs771656662 Chr.9: 73167579-73167579 on Build GRCh38
rs760037657 Chr.9: 73167466-73167466 on Build GRCh38
rs376925893 Chr.9: 73167604-73167604 on Build GRCh38
rs554265596 Chr.9: 73167696-73167696 on Build GRCh38
rs150591447 Chr.9: 73167529-73167529 on Build GRCh38
rs781618965 Chr.9: 73167533-73167533 on Build GRCh38
rs934881175 Chr.9: 73167727-73167727 on Build GRCh38
rs961445045 Chr.9: 73167405-73167405 on Build GRCh38
rs924619104 Chr.9: 73167704-73167704 on Build GRCh38
rs73491666 Chr.9: 73167784-73167784 on Build GRCh38
rs971831043 Chr.9: 73167677-73167677 on Build GRCh38
rs752848971 Chr.9: 73167483-73167485 on Build GRCh38
rs369705098 Chr.9: 73167571-73167571 on Build GRCh38
rs1044582327 Chr.9: 73167766-73167766 on Build GRCh38
rs17058570 Chr.9: 73167639-73167639 on Build GRCh38
rs927242880 Chr.9: 73167733-73167733 on Build GRCh38
rs757438191 Chr.9: 73167810-73167811 on Build GRCh38
rs748458922 Chr.9: 73167452-73167452 on Build GRCh38
rs1040755709 Chr.9: 73167694-73167694 on Build GRCh38
rs372730961 Chr.9: 73167490-73167490 on Build GRCh38
rs376543717 Chr.9: 73167610-73167610 on Build GRCh38
rs998462522 Chr.9: 73167601-73167601 on Build GRCh38
rs181802457 Chr.9: 73167565-73167565 on Build GRCh38
rs1044674424 Chr.9: 73167479-73167479 on Build GRCh38
rs935927146 Chr.9: 73167736-73167736 on Build GRCh38
rs889243107 Chr.9: 73167482-73167482 on Build GRCh38

View Full Product Details