Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77201157-77201674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260379 c.145-4G>T p.? Chr.9: 77201361-77201361 on Build GRCh38
5027083 c.177T>C p.V59V Chr.9: 77201397-77201397 on Build GRCh38
5027085 c.177T>C p.V59V Chr.9: 77201397-77201397 on Build GRCh38
5027084 c.177T>C p.V59V Chr.9: 77201397-77201397 on Build GRCh38
6260377 c.145-4G>T p.? Chr.9: 77201361-77201361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764960108 Chr.9: 77201406-77201406 on Build GRCh38
rs770094234 Chr.9: 77201326-77201326 on Build GRCh38
rs971049211 Chr.9: 77201408-77201408 on Build GRCh38
rs374994178 Chr.9: 77201338-77201338 on Build GRCh38
rs201490407 Chr.9: 77201392-77201392 on Build GRCh38
rs907509619 Chr.9: 77201589-77201589 on Build GRCh38
rs527467903 Chr.9: 77201553-77201553 on Build GRCh38
rs192252329 Chr.9: 77201454-77201454 on Build GRCh38
rs1038051922 Chr.9: 77201612-77201612 on Build GRCh38
rs773748695 Chr.9: 77201342-77201342 on Build GRCh38
rs780864964 Chr.9: 77201441-77201441 on Build GRCh38
rs373944070 Chr.9: 77201323-77201323 on Build GRCh38
rs982854218 Chr.9: 77201428-77201428 on Build GRCh38
rs367761703 Chr.9: 77201537-77201537 on Build GRCh38
rs915481091 Chr.9: 77201491-77201491 on Build GRCh38
rs370010973 Chr.9: 77201457-77201457 on Build GRCh38
rs958465700 Chr.9: 77201250-77201250 on Build GRCh38
rs754661980 Chr.9: 77201346-77201346 on Build GRCh38
rs945435537 Chr.9: 77201595-77201595 on Build GRCh38
rs970746451 Chr.9: 77201369-77201369 on Build GRCh38
rs1031390957 Chr.9: 77201443-77201443 on Build GRCh38
rs1005179599 Chr.9: 77201275-77201275 on Build GRCh38
rs751820949 Chr.9: 77201489-77201490 on Build GRCh38
rs575039679 Chr.9: 77201524-77201524 on Build GRCh38
rs780764326 Chr.9: 77201318-77201318 on Build GRCh38
rs990160798 Chr.9: 77201484-77201484 on Build GRCh38
rs559400268 Chr.9: 77201603-77201603 on Build GRCh38
rs1003553884 Chr.9: 77201377-77201377 on Build GRCh38
rs767771206 Chr.9: 77201380-77201380 on Build GRCh38
rs986814380 Chr.9: 77201569-77201569 on Build GRCh38
rs373128010 Chr.9: 77201209-77201209 on Build GRCh38
rs751415681 Chr.9: 77201353-77201353 on Build GRCh38
rs573008708 Chr.9: 77201361-77201361 on Build GRCh38
rs921684024 Chr.9: 77201526-77201526 on Build GRCh38
rs142737744 Chr.9: 77201602-77201602 on Build GRCh38
rs1023712870 Chr.9: 77201348-77201348 on Build GRCh38
rs932752080 Chr.9: 77201566-77201566 on Build GRCh38
rs145306631 Chr.9: 77201386-77201386 on Build GRCh38
rs956992701 Chr.9: 77201479-77201479 on Build GRCh38
rs140390541 Chr.9: 77201546-77201546 on Build GRCh38
rs751101064 Chr.9: 77201430-77201430 on Build GRCh38
rs1057515645 Chr.9: 77201379-77201379 on Build GRCh38
rs1051830471 Chr.9: 77201179-77201179 on Build GRCh38
rs964238103 Chr.9: 77201494-77201494 on Build GRCh38
rs933025143 Chr.9: 77201570-77201570 on Build GRCh38
rs117478720 Chr.9: 77201573-77201573 on Build GRCh38
rs750344038 Chr.9: 77201420-77201420 on Build GRCh38
rs908829695 Chr.9: 77201480-77201480 on Build GRCh38
rs758410781 Chr.9: 77201421-77201421 on Build GRCh38
rs899027958 Chr.9: 77201306-77201306 on Build GRCh38
rs150034322 Chr.9: 77201370-77201370 on Build GRCh38
rs903129280 Chr.9: 77201613-77201613 on Build GRCh38
rs747806517 Chr.9: 77201449-77201449 on Build GRCh38
rs541823714 Chr.9: 77201436-77201436 on Build GRCh38
rs921409545 Chr.9: 77201515-77201515 on Build GRCh38
rs565212468 Chr.9: 77201512-77201512 on Build GRCh38
rs903095490 Chr.9: 77201192-77201192 on Build GRCh38
rs765929238 Chr.9: 77201429-77201429 on Build GRCh38
rs1012645934 Chr.9: 77201298-77201298 on Build GRCh38
rs1037559103 Chr.9: 77201302-77201302 on Build GRCh38
rs759700958 Chr.9: 77201376-77201376 on Build GRCh38
rs1012023425 Chr.9: 77201344-77201344 on Build GRCh38
rs780525040 Chr.9: 77201474-77201474 on Build GRCh38

View Full Product Details