Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127454811-127455309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001005374.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
752684 c.129G>T p.E43D Chr.9: 127455054-127455054 on Build GRCh38
1105321 c.123C>A p.L41L Chr.9: 127455048-127455048 on Build GRCh38
210996 c.95A>G p.D32G Chr.9: 127455020-127455020 on Build GRCh38
7310721 c.85G>T p.G29W Chr.9: 127455010-127455010 on Build GRCh38
4699343 c.129_129+1insTTTAC p.I44fs*19 Chr.9: 127455054-127455055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754374008 Chr.9: 127455000-127455000 on Build GRCh38
rs34004486 Chr.9: 127454845-127454846 on Build GRCh38
rs749592869 Chr.9: 127455096-127455096 on Build GRCh38
rs984897512 Chr.9: 127455119-127455119 on Build GRCh38
rs756364990 Chr.9: 127454990-127454990 on Build GRCh38
rs1042698351 Chr.9: 127455198-127455198 on Build GRCh38
rs989483317 Chr.9: 127455192-127455192 on Build GRCh38
rs761882657 Chr.9: 127455212-127455212 on Build GRCh38
rs779388263 Chr.9: 127455105-127455105 on Build GRCh38
rs749609283 Chr.9: 127455006-127455006 on Build GRCh38
rs554398886 Chr.9: 127455172-127455172 on Build GRCh38
rs116885615 Chr.9: 127455171-127455171 on Build GRCh38
rs750570495 Chr.9: 127455241-127455241 on Build GRCh38
rs1015490953 Chr.9: 127454960-127454960 on Build GRCh38
rs997504257 Chr.9: 127455266-127455266 on Build GRCh38
rs372244339 Chr.9: 127455065-127455065 on Build GRCh38
rs949045119 Chr.9: 127455197-127455197 on Build GRCh38
rs748752390 Chr.9: 127455022-127455022 on Build GRCh38
rs762731732 Chr.9: 127455047-127455047 on Build GRCh38
rs142325014 Chr.9: 127455238-127455238 on Build GRCh38
rs775125689 Chr.9: 127455045-127455045 on Build GRCh38
rs866014835 Chr.9: 127455082-127455082 on Build GRCh38
rs950999428 Chr.9: 127455288-127455288 on Build GRCh38
rs556723022 Chr.9: 127455232-127455232 on Build GRCh38
rs372437126 Chr.9: 127455139-127455139 on Build GRCh38
rs781381373 Chr.9: 127454973-127454973 on Build GRCh38
rs762785076 Chr.9: 127455027-127455027 on Build GRCh38
rs766476460 Chr.9: 127455074-127455074 on Build GRCh38
rs893196167 Chr.9: 127454868-127454868 on Build GRCh38
rs776291709 Chr.9: 127454870-127454870 on Build GRCh38
rs369250629 Chr.9: 127454977-127454977 on Build GRCh38
rs950905476 Chr.9: 127455054-127455054 on Build GRCh38
rs151069056 Chr.9: 127455079-127455079 on Build GRCh38
rs150784835 Chr.9: 127455019-127455019 on Build GRCh38
rs751429630 Chr.9: 127455068-127455068 on Build GRCh38
rs1018833853 Chr.9: 127455055-127455055 on Build GRCh38
rs764809777 Chr.9: 127455040-127455040 on Build GRCh38
rs1003745469 Chr.9: 127454902-127454902 on Build GRCh38
rs1054562138 Chr.9: 127454842-127454842 on Build GRCh38
rs769070078 Chr.9: 127455014-127455014 on Build GRCh38
rs948307525 Chr.9: 127454848-127454848 on Build GRCh38
rs759065503 Chr.9: 127455031-127455031 on Build GRCh38
rs1007634949 Chr.9: 127454875-127454875 on Build GRCh38
rs371533252 Chr.9: 127454957-127454957 on Build GRCh38
rs889604541 Chr.9: 127455216-127455216 on Build GRCh38
rs756278772 Chr.9: 127455088-127455088 on Build GRCh38
rs551183702 Chr.9: 127455028-127455028 on Build GRCh38
rs906704572 Chr.9: 127454888-127454888 on Build GRCh38
rs7851331 Chr.9: 127454968-127454968 on Build GRCh38
rs142846298 Chr.9: 127455011-127455011 on Build GRCh38
rs113268454 Chr.9: 127455173-127455173 on Build GRCh38
rs780358722 Chr.9: 127455094-127455094 on Build GRCh38
rs746055151 Chr.9: 127454984-127454984 on Build GRCh38
rs1026998853 Chr.9: 127455281-127455281 on Build GRCh38
rs936502202 Chr.9: 127455204-127455204 on Build GRCh38
rs199681048 Chr.9: 127455247-127455248 on Build GRCh38
rs767506004 Chr.9: 127455239-127455239 on Build GRCh38
rs553351622 Chr.9: 127455097-127455097 on Build GRCh38
rs113996142 Chr.9: 127455182-127455182 on Build GRCh38
rs923829746 Chr.9: 127455203-127455203 on Build GRCh38
rs570989650 Chr.9: 127455077-127455077 on Build GRCh38
rs563289493 Chr.9: 127454943-127454943 on Build GRCh38

View Full Product Details