Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127455235-127455735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001005374.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3779629 c.133C>G p.P45A Chr.9: 127455579-127455579 on Build GRCh38
1701754 c.134C>T p.P45L Chr.9: 127455580-127455580 on Build GRCh38
4163342 c.149C>A p.A50E Chr.9: 127455595-127455595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999143997 Chr.9: 127455492-127455492 on Build GRCh38
rs1036568768 Chr.9: 127455334-127455334 on Build GRCh38
rs775109275 Chr.9: 127455544-127455544 on Build GRCh38
rs755427350 Chr.9: 127455600-127455600 on Build GRCh38
rs559309350 Chr.9: 127455409-127455409 on Build GRCh38
rs562408597 Chr.9: 127455462-127455462 on Build GRCh38
rs779022904 Chr.9: 127455621-127455621 on Build GRCh38
rs531212208 Chr.9: 127455463-127455463 on Build GRCh38
rs761604510 Chr.9: 127455669-127455669 on Build GRCh38
rs959522626 Chr.9: 127455394-127455394 on Build GRCh38
rs546739304 Chr.9: 127455582-127455582 on Build GRCh38
rs377012683 Chr.9: 127455350-127455350 on Build GRCh38
rs551222373 Chr.9: 127455487-127455487 on Build GRCh38
rs761535879 Chr.9: 127455574-127455574 on Build GRCh38
rs778570971 Chr.9: 127455654-127455666 on Build GRCh38
rs769269709 Chr.9: 127455657-127455657 on Build GRCh38
rs778245782 Chr.9: 127455645-127455645 on Build GRCh38
rs769357543 Chr.9: 127455543-127455543 on Build GRCh38
rs997504257 Chr.9: 127455266-127455266 on Build GRCh38
rs752012848 Chr.9: 127455654-127455654 on Build GRCh38
rs748778245 Chr.9: 127455658-127455658 on Build GRCh38
rs773718248 Chr.9: 127455538-127455538 on Build GRCh38
rs754095363 Chr.9: 127455604-127455604 on Build GRCh38
rs758692752 Chr.9: 127455635-127455635 on Build GRCh38
rs950999428 Chr.9: 127455288-127455288 on Build GRCh38
rs541806458 Chr.9: 127455448-127455448 on Build GRCh38
rs752175786 Chr.9: 127455657-127455666 on Build GRCh38
rs933390412 Chr.9: 127455662-127455662 on Build GRCh38
rs768307214 Chr.9: 127455659-127455659 on Build GRCh38
rs756282129 Chr.9: 127455345-127455345 on Build GRCh38
rs756260380 Chr.9: 127455589-127455589 on Build GRCh38
rs886442023 Chr.9: 127455707-127455707 on Build GRCh38
rs755272225 Chr.9: 127455610-127455610 on Build GRCh38
rs571026765 Chr.9: 127455494-127455494 on Build GRCh38
rs762713959 Chr.9: 127455547-127455547 on Build GRCh38
rs757980176 Chr.9: 127455657-127455659 on Build GRCh38
rs957176698 Chr.9: 127455559-127455559 on Build GRCh38
rs897963329 Chr.9: 127455375-127455375 on Build GRCh38
rs923587436 Chr.9: 127455464-127455464 on Build GRCh38
rs939628894 Chr.9: 127455671-127455671 on Build GRCh38
rs754216773 Chr.9: 127455528-127455528 on Build GRCh38
rs970817544 Chr.9: 127455440-127455440 on Build GRCh38
rs763765654 Chr.9: 127455555-127455555 on Build GRCh38
rs942509436 Chr.9: 127455592-127455592 on Build GRCh38
rs779499213 Chr.9: 127455710-127455710 on Build GRCh38
rs533484572 Chr.9: 127455502-127455502 on Build GRCh38
rs1006412056 Chr.9: 127455324-127455324 on Build GRCh38
rs745307512 Chr.9: 127455542-127455542 on Build GRCh38
rs373156612 Chr.9: 127455630-127455630 on Build GRCh38
rs754578966 Chr.9: 127455651-127455652 on Build GRCh38
rs369384290 Chr.9: 127455570-127455570 on Build GRCh38
rs772577965 Chr.9: 127455533-127455533 on Build GRCh38
rs565955354 Chr.9: 127455426-127455426 on Build GRCh38
rs989390114 Chr.9: 127455518-127455518 on Build GRCh38
rs911066520 Chr.9: 127455534-127455534 on Build GRCh38
rs139695483 Chr.9: 127455580-127455580 on Build GRCh38
rs1048820114 Chr.9: 127455432-127455432 on Build GRCh38
rs936595016 Chr.9: 127455472-127455472 on Build GRCh38
rs1026998853 Chr.9: 127455281-127455281 on Build GRCh38
rs34598808 Chr.9: 127455300-127455301 on Build GRCh38
rs1047746185 Chr.9: 127455702-127455702 on Build GRCh38
rs777564486 Chr.9: 127455661-127455666 on Build GRCh38
rs977756345 Chr.9: 127455451-127455451 on Build GRCh38
rs773889029 Chr.9: 127455660-127455660 on Build GRCh38
rs375537151 Chr.9: 127455525-127455525 on Build GRCh38
rs545553301 Chr.9: 127455382-127455382 on Build GRCh38
rs753041879 Chr.9: 127455423-127455423 on Build GRCh38

View Full Product Details