Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38299059-38299540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1235538 c.124G>A p.V42I Chr.15: 38299464-38299464 on Build GRCh38
3386841 c.66G>T p.M22I Chr.15: 38299406-38299406 on Build GRCh38
433752 c.182G>A p.R61H Chr.15: 38299522-38299522 on Build GRCh38
107788 c.148C>T p.Q50* Chr.15: 38299488-38299488 on Build GRCh38
416510 c.185G>A p.G62E Chr.15: 38299525-38299525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775334473 Chr.15: 38299503-38299503 on Build GRCh38
rs781022987 Chr.15: 38299295-38299295 on Build GRCh38
rs780614363 Chr.15: 38299398-38299398 on Build GRCh38
rs781390048 Chr.15: 38299459-38299459 on Build GRCh38
rs962223261 Chr.15: 38299120-38299120 on Build GRCh38
rs761830220 Chr.15: 38299518-38299518 on Build GRCh38
rs142406055 Chr.15: 38299294-38299294 on Build GRCh38
rs986112492 Chr.15: 38299244-38299244 on Build GRCh38
rs1057517941 Chr.15: 38299386-38299386 on Build GRCh38
rs397517871 Chr.15: 38299517-38299517 on Build GRCh38
rs766826710 Chr.15: 38299149-38299149 on Build GRCh38
rs536022328 Chr.15: 38299090-38299090 on Build GRCh38
rs114699794 Chr.15: 38299171-38299171 on Build GRCh38
rs914544937 Chr.15: 38299258-38299258 on Build GRCh38
rs774071345 Chr.15: 38299325-38299325 on Build GRCh38
rs760939387 Chr.15: 38299508-38299508 on Build GRCh38
rs927043607 Chr.15: 38299091-38299091 on Build GRCh38
rs574723299 Chr.15: 38299088-38299088 on Build GRCh38
rs947392765 Chr.15: 38299272-38299272 on Build GRCh38
rs1006222486 Chr.15: 38299299-38299299 on Build GRCh38
rs975930887 Chr.15: 38299133-38299133 on Build GRCh38
rs959758606 Chr.15: 38299510-38299510 on Build GRCh38
rs144515463 Chr.15: 38299323-38299324 on Build GRCh38
rs755788325 Chr.15: 38299409-38299409 on Build GRCh38
rs1056874064 Chr.15: 38299112-38299112 on Build GRCh38
rs760850290 Chr.15: 38299366-38299366 on Build GRCh38
rs758855467 Chr.15: 38299393-38299393 on Build GRCh38
rs929881400 Chr.15: 38299157-38299157 on Build GRCh38
rs749241525 Chr.15: 38299312-38299312 on Build GRCh38
rs769323362 Chr.15: 38299303-38299303 on Build GRCh38
rs757726772 Chr.15: 38299387-38299387 on Build GRCh38
rs778847063 Chr.15: 38299474-38299474 on Build GRCh38
rs10220731 Chr.15: 38299099-38299099 on Build GRCh38
rs148646547 Chr.15: 38299488-38299488 on Build GRCh38
rs1042234072 Chr.15: 38299311-38299311 on Build GRCh38
rs772636310 Chr.15: 38299308-38299308 on Build GRCh38
rs137939714 Chr.15: 38299087-38299087 on Build GRCh38
rs757623945 Chr.15: 38299231-38299231 on Build GRCh38
rs1057518150 Chr.15: 38299411-38299411 on Build GRCh38
rs147204964 Chr.15: 38299464-38299464 on Build GRCh38
rs745859168 Chr.15: 38299481-38299481 on Build GRCh38
rs768775931 Chr.15: 38299512-38299512 on Build GRCh38
rs864622421 Chr.15: 38299391-38299391 on Build GRCh38
rs926984710 Chr.15: 38299472-38299472 on Build GRCh38
rs367745143 Chr.15: 38299400-38299400 on Build GRCh38
rs572710790 Chr.15: 38299259-38299259 on Build GRCh38
rs868471292 Chr.15: 38299114-38299114 on Build GRCh38
rs771825294 Chr.15: 38299483-38299483 on Build GRCh38
rs887742500 Chr.15: 38299249-38299249 on Build GRCh38
rs121434313 Chr.15: 38299410-38299410 on Build GRCh38
rs200789030 Chr.15: 38299331-38299331 on Build GRCh38
rs751895777 Chr.15: 38299219-38299219 on Build GRCh38
rs750777752 Chr.15: 38299392-38299392 on Build GRCh38
rs775671940 Chr.15: 38299362-38299362 on Build GRCh38
rs765788978 Chr.15: 38299521-38299521 on Build GRCh38
rs748759533 Chr.15: 38299463-38299463 on Build GRCh38
rs968982670 Chr.15: 38299396-38299396 on Build GRCh38
rs746572586 Chr.15: 38299307-38299308 on Build GRCh38
rs764234046 Chr.15: 38299382-38299382 on Build GRCh38
rs900576451 Chr.15: 38299341-38299341 on Build GRCh38
rs369708660 Chr.15: 38299186-38299186 on Build GRCh38
rs953257817 Chr.15: 38299162-38299162 on Build GRCh38
rs753924528 Chr.15: 38299383-38299383 on Build GRCh38
rs148520129 Chr.15: 38299324-38299325 on Build GRCh38
rs1057517922 Chr.15: 38299378-38299378 on Build GRCh38
rs767682280 Chr.15: 38299356-38299356 on Build GRCh38
rs1011924858 Chr.15: 38299079-38299079 on Build GRCh38
rs121434318 Chr.15: 38299471-38299471 on Build GRCh38
rs536769726 Chr.15: 38299086-38299086 on Build GRCh38

View Full Product Details