Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18671842-18672294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RS1
Entrez Gene ID
6247
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
UniGene
Hs.715725
Gene Chromosome Location
Chr.X: 18639688-18672103 on Build GRCh38
Transcripts
NM_000330.3

SNP Detail

SNP ID Chromosome Location
rs1014702159 Chr.X: 18671903-18671903 on Build GRCh38
rs766045580 Chr.X: 18672011-18672011 on Build GRCh38
rs975938578 Chr.X: 18672124-18672124 on Build GRCh38
rs755875252 Chr.X: 18672103-18672103 on Build GRCh38
rs920021291 Chr.X: 18672137-18672137 on Build GRCh38
rs763279160 Chr.X: 18671999-18671999 on Build GRCh38
rs281865338 Chr.X: 18672012-18672012 on Build GRCh38
rs104894935 Chr.X: 18672031-18672031 on Build GRCh38
rs62645879 Chr.X: 18672034-18672034 on Build GRCh38
rs931351963 Chr.X: 18672171-18672171 on Build GRCh38
rs763784544 Chr.X: 18672125-18672125 on Build GRCh38
rs553095350 Chr.X: 18671888-18671888 on Build GRCh38
rs281865334 Chr.X: 18672015-18672015 on Build GRCh38
rs756754673 Chr.X: 18672184-18672184 on Build GRCh38
rs765691829 Chr.X: 18671956-18671956 on Build GRCh38
rs281865332 Chr.X: 18672068-18672068 on Build GRCh38
rs189210045 Chr.X: 18671957-18671957 on Build GRCh38
rs947507986 Chr.X: 18672143-18672143 on Build GRCh38
rs202051526 Chr.X: 18672230-18672230 on Build GRCh38
rs766906113 Chr.X: 18672028-18672028 on Build GRCh38
rs281865336 Chr.X: 18672016-18672016 on Build GRCh38
rs571944937 Chr.X: 18672062-18672062 on Build GRCh38
rs1005433341 Chr.X: 18671869-18671869 on Build GRCh38
rs781282917 Chr.X: 18672083-18672083 on Build GRCh38
rs949324623 Chr.X: 18672199-18672199 on Build GRCh38
rs374672514 Chr.X: 18672061-18672061 on Build GRCh38
rs756086692 Chr.X: 18672059-18672059 on Build GRCh38
rs370979911 Chr.X: 18672078-18672078 on Build GRCh38
rs1057389348 Chr.X: 18672220-18672220 on Build GRCh38
rs964065643 Chr.X: 18672084-18672084 on Build GRCh38
rs767155536 Chr.X: 18672032-18672032 on Build GRCh38
rs749064492 Chr.X: 18671889-18671892 on Build GRCh38
rs753313238 Chr.X: 18672128-18672128 on Build GRCh38
rs1028699372 Chr.X: 18671977-18671977 on Build GRCh38
rs370246632 Chr.X: 18671984-18671984 on Build GRCh38
rs761903178 Chr.X: 18671902-18671902 on Build GRCh38
rs376367416 Chr.X: 18672077-18672077 on Build GRCh38
rs752638328 Chr.X: 18672050-18672050 on Build GRCh38
rs281865337 Chr.X: 18672015-18672016 on Build GRCh38
rs193089324 Chr.X: 18671978-18671978 on Build GRCh38
rs913325405 Chr.X: 18672138-18672138 on Build GRCh38
rs908357103 Chr.X: 18672197-18672197 on Build GRCh38
rs771208584 Chr.X: 18672109-18672109 on Build GRCh38
rs904831143 Chr.X: 18671925-18671925 on Build GRCh38
rs780257454 Chr.X: 18672063-18672063 on Build GRCh38
rs879077136 Chr.X: 18672235-18672235 on Build GRCh38
rs893326724 Chr.X: 18672055-18672055 on Build GRCh38
rs770267407 Chr.X: 18672098-18672098 on Build GRCh38
rs769573671 Chr.X: 18671887-18671888 on Build GRCh38
rs281865333 Chr.X: 18672067-18672067 on Build GRCh38
rs281865335 Chr.X: 18672033-18672036 on Build GRCh38
rs766251953 Chr.X: 18672153-18672153 on Build GRCh38

View Full Product Details