Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108877713-108878311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
UniGene
Hs.494738
Gene Chromosome Location
Chr.9: 108867520-108934332 on Build GRCh38
Transcripts
NM_001318360.1 NM_003640.4 XM_011519136.1 XM_011519137.1 XM_017015238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325537 c.3755G>T p.G1252V Chr.9: 108878095-108878095 on Build GRCh38
325538 c.3754G>T p.G1252* Chr.9: 108878096-108878096 on Build GRCh38
6684729 c.3725T>C p.V1242A Chr.9: 108878125-108878125 on Build GRCh38
752135 c.3750A>G p.E1250E Chr.9: 108878100-108878100 on Build GRCh38
1103673 c.3835C>T p.Q1279* Chr.9: 108878015-108878015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008331798 Chr.9: 108877780-108877780 on Build GRCh38
rs755563190 Chr.9: 108878016-108878016 on Build GRCh38
rs149038322 Chr.9: 108878100-108878100 on Build GRCh38
rs574431100 Chr.9: 108878145-108878145 on Build GRCh38
rs199723919 Chr.9: 108878065-108878065 on Build GRCh38
rs781725905 Chr.9: 108877817-108877817 on Build GRCh38
rs746323148 Chr.9: 108877881-108877881 on Build GRCh38
rs201242444 Chr.9: 108878031-108878031 on Build GRCh38
rs774498209 Chr.9: 108878198-108878198 on Build GRCh38
rs767751268 Chr.9: 108878020-108878020 on Build GRCh38
rs990588465 Chr.9: 108878184-108878184 on Build GRCh38
rs560114973 Chr.9: 108878226-108878226 on Build GRCh38
rs749498874 Chr.9: 108878168-108878168 on Build GRCh38
rs763298552 Chr.9: 108878077-108878077 on Build GRCh38
rs934044357 Chr.9: 108878262-108878262 on Build GRCh38
rs557015443 Chr.9: 108877977-108877977 on Build GRCh38
rs3818875 Chr.9: 108878244-108878244 on Build GRCh38
rs926236782 Chr.9: 108878204-108878204 on Build GRCh38
rs778170450 Chr.9: 108878080-108878080 on Build GRCh38
rs760084617 Chr.9: 108878093-108878093 on Build GRCh38
rs774564951 Chr.9: 108878189-108878189 on Build GRCh38
rs773500103 Chr.9: 108878072-108878072 on Build GRCh38
rs934668278 Chr.9: 108878212-108878212 on Build GRCh38
rs749975442 Chr.9: 108878136-108878136 on Build GRCh38
rs138294904 Chr.9: 108877790-108877790 on Build GRCh38
rs781155896 Chr.9: 108878156-108878156 on Build GRCh38
rs141974657 Chr.9: 108877958-108877958 on Build GRCh38
rs917756957 Chr.9: 108877923-108877923 on Build GRCh38
rs558588647 Chr.9: 108878023-108878023 on Build GRCh38
rs753141812 Chr.9: 108878021-108878021 on Build GRCh38
rs1016528229 Chr.9: 108877742-108877742 on Build GRCh38
rs201888676 Chr.9: 108878059-108878059 on Build GRCh38
rs533673443 Chr.9: 108877986-108877986 on Build GRCh38
rs916051427 Chr.9: 108878187-108878187 on Build GRCh38
rs756441006 Chr.9: 108878110-108878110 on Build GRCh38
rs941782750 Chr.9: 108878284-108878284 on Build GRCh38
rs926394717 Chr.9: 108877963-108877963 on Build GRCh38
rs745812438 Chr.9: 108877960-108877960 on Build GRCh38
rs75395135 Chr.9: 108878104-108878104 on Build GRCh38
rs369417060 Chr.9: 108878154-108878154 on Build GRCh38
rs372773763 Chr.9: 108878039-108878039 on Build GRCh38
rs766517046 Chr.9: 108878083-108878083 on Build GRCh38
rs774622315 Chr.9: 108878089-108878089 on Build GRCh38
rs771727737 Chr.9: 108877962-108877962 on Build GRCh38
rs199813856 Chr.9: 108877996-108877996 on Build GRCh38
rs780376545 Chr.9: 108877967-108877967 on Build GRCh38
rs752988049 Chr.9: 108878107-108878107 on Build GRCh38
rs756928248 Chr.9: 108878120-108878120 on Build GRCh38
rs1053555749 Chr.9: 108878243-108878243 on Build GRCh38
rs775118324 Chr.9: 108878064-108878064 on Build GRCh38
rs766569938 Chr.9: 108878005-108878005 on Build GRCh38
rs956568797 Chr.9: 108878214-108878214 on Build GRCh38
rs573025020 Chr.9: 108877900-108877900 on Build GRCh38
rs756667775 Chr.9: 108877792-108877792 on Build GRCh38
rs961343248 Chr.9: 108877767-108877767 on Build GRCh38
rs1032256576 Chr.9: 108877824-108877824 on Build GRCh38
rs201673697 Chr.9: 108878121-108878121 on Build GRCh38
rs543759021 Chr.9: 108878028-108878028 on Build GRCh38
rs916147770 Chr.9: 108877917-108877917 on Build GRCh38
rs368761085 Chr.9: 108878142-108878142 on Build GRCh38
rs112914549 Chr.9: 108877933-108877933 on Build GRCh38
rs757251624 Chr.9: 108877962-108877963 on Build GRCh38
rs200634033 Chr.9: 108878066-108878066 on Build GRCh38
rs753451741 Chr.9: 108878116-108878116 on Build GRCh38
rs748861216 Chr.9: 108877948-108877948 on Build GRCh38
rs768852660 Chr.9: 108878197-108878197 on Build GRCh38
rs1024782852 Chr.9: 108878231-108878231 on Build GRCh38
rs926389639 Chr.9: 108878274-108878274 on Build GRCh38
rs901285079 Chr.9: 108877995-108877995 on Build GRCh38
rs764373685 Chr.9: 108878113-108878113 on Build GRCh38
rs148535504 Chr.9: 108878155-108878155 on Build GRCh38
rs752071786 Chr.9: 108878014-108878014 on Build GRCh38
rs10435855 Chr.9: 108878273-108878273 on Build GRCh38
rs777536646 Chr.9: 108878024-108878024 on Build GRCh38
rs1018343767 Chr.9: 108878183-108878183 on Build GRCh38
rs111634780 Chr.9: 108877947-108877947 on Build GRCh38
rs2275497 Chr.9: 108877804-108877804 on Build GRCh38
rs927744605 Chr.9: 108877794-108877794 on Build GRCh38
rs1018664631 Chr.9: 108877785-108877785 on Build GRCh38
rs374876813 Chr.9: 108877953-108877953 on Build GRCh38
rs145380688 Chr.9: 108878034-108878034 on Build GRCh38
rs775382171 Chr.9: 108877974-108877974 on Build GRCh38
rs576438853 Chr.9: 108878027-108878027 on Build GRCh38
rs749552737 Chr.9: 108878000-108878000 on Build GRCh38
rs771317944 Chr.9: 108878173-108878173 on Build GRCh38
rs778376709 Chr.9: 108878122-108878122 on Build GRCh38
rs113767921 Chr.9: 108877976-108877976 on Build GRCh38

View Full Product Details