Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78002649-78003248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3B1
Entrez Gene ID
8546
Gene Name
adaptor related protein complex 3 beta 1 subunit
Gene Aliases
ADTB3, ADTB3A, HPS, HPS2, PE
UniGene
Hs.532091
Gene Chromosome Location
Chr.5: 78002326-78294755 on Build GRCh38
Transcripts
NM_001271769.1 NM_003664.4 XM_005248618.3 XM_017010001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836528 c.3253C>T p.R1085W Chr.5: 78002934-78002934 on Build GRCh38
4662250 c.3274T>A p.S1092T Chr.5: 78002913-78002913 on Build GRCh38
5759205 c.3165G>A p.L1055L Chr.5: 78003022-78003022 on Build GRCh38
1070334 c.3259C>A p.L1087M Chr.5: 78002928-78002928 on Build GRCh38
5774268 c.3132-4C>A p.? Chr.5: 78003059-78003059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886060769 Chr.5: 78002832-78002833 on Build GRCh38
rs918624052 Chr.5: 78002810-78002810 on Build GRCh38
rs190285646 Chr.5: 78002720-78002720 on Build GRCh38
rs942517218 Chr.5: 78002894-78002894 on Build GRCh38
rs755468074 Chr.5: 78003012-78003012 on Build GRCh38
rs994453591 Chr.5: 78003189-78003189 on Build GRCh38
rs992209236 Chr.5: 78002757-78002757 on Build GRCh38
rs1009638675 Chr.5: 78002878-78002878 on Build GRCh38
rs763841787 Chr.5: 78002866-78002866 on Build GRCh38
rs780686324 Chr.5: 78003096-78003097 on Build GRCh38
rs767072786 Chr.5: 78002997-78002997 on Build GRCh38
rs766029582 Chr.5: 78002801-78002801 on Build GRCh38
rs921887216 Chr.5: 78003135-78003135 on Build GRCh38
rs777824071 Chr.5: 78003029-78003029 on Build GRCh38
rs540703378 Chr.5: 78002782-78002782 on Build GRCh38
rs1021123097 Chr.5: 78002684-78002684 on Build GRCh38
rs753283084 Chr.5: 78002761-78002761 on Build GRCh38
rs751258161 Chr.5: 78002992-78002992 on Build GRCh38
rs765141824 Chr.5: 78002853-78002853 on Build GRCh38
rs369987939 Chr.5: 78002719-78002719 on Build GRCh38
rs34089426 Chr.5: 78002980-78002980 on Build GRCh38
rs778685628 Chr.5: 78003082-78003082 on Build GRCh38
rs764100439 Chr.5: 78002990-78002990 on Build GRCh38
rs776628092 Chr.5: 78003051-78003051 on Build GRCh38
rs11552314 Chr.5: 78002795-78002795 on Build GRCh38
rs6864605 Chr.5: 78002711-78002711 on Build GRCh38
rs576013918 Chr.5: 78003124-78003124 on Build GRCh38
rs772549509 Chr.5: 78003025-78003025 on Build GRCh38
rs755752427 Chr.5: 78002900-78002900 on Build GRCh38
rs558228575 Chr.5: 78003111-78003111 on Build GRCh38
rs767395436 Chr.5: 78003078-78003078 on Build GRCh38
rs929044217 Chr.5: 78002797-78002797 on Build GRCh38
rs142431658 Chr.5: 78003220-78003220 on Build GRCh38
rs200456538 Chr.5: 78002875-78002875 on Build GRCh38
rs772128060 Chr.5: 78002953-78002953 on Build GRCh38
rs182487478 Chr.5: 78002858-78002858 on Build GRCh38
rs769307741 Chr.5: 78002917-78002917 on Build GRCh38
rs773537846 Chr.5: 78002957-78002957 on Build GRCh38
rs748447113 Chr.5: 78003024-78003024 on Build GRCh38
rs760362235 Chr.5: 78003093-78003093 on Build GRCh38
rs538645525 Chr.5: 78003100-78003100 on Build GRCh38
rs761729448 Chr.5: 78002993-78002993 on Build GRCh38
rs535976982 Chr.5: 78003019-78003019 on Build GRCh38
rs778059961 Chr.5: 78003120-78003120 on Build GRCh38
rs538538566 Chr.5: 78003143-78003143 on Build GRCh38
rs760757738 Chr.5: 78002851-78002851 on Build GRCh38
rs895976215 Chr.5: 78003180-78003180 on Build GRCh38
rs750141732 Chr.5: 78003001-78003001 on Build GRCh38
rs780677693 Chr.5: 78002905-78002905 on Build GRCh38
rs760735640 Chr.5: 78002973-78002973 on Build GRCh38
rs753428510 Chr.5: 78003009-78003009 on Build GRCh38
rs146849359 Chr.5: 78003013-78003013 on Build GRCh38
rs771162712 Chr.5: 78003049-78003049 on Build GRCh38
rs779106683 Chr.5: 78002920-78002920 on Build GRCh38
rs375163910 Chr.5: 78002873-78002873 on Build GRCh38
rs759629484 Chr.5: 78003059-78003059 on Build GRCh38
rs755664373 Chr.5: 78003005-78003005 on Build GRCh38
rs570942210 Chr.5: 78002772-78002772 on Build GRCh38
rs780915786 Chr.5: 78002874-78002874 on Build GRCh38
rs976542118 Chr.5: 78002961-78002961 on Build GRCh38
rs546653512 Chr.5: 78003030-78003030 on Build GRCh38
rs748174176 Chr.5: 78003108-78003108 on Build GRCh38
rs759043397 Chr.5: 78003037-78003038 on Build GRCh38
rs746740092 Chr.5: 78003059-78003060 on Build GRCh38
rs749861794 Chr.5: 78002906-78002906 on Build GRCh38
rs959070738 Chr.5: 78002735-78002735 on Build GRCh38
rs753283239 Chr.5: 78003115-78003115 on Build GRCh38
rs758664475 Chr.5: 78003118-78003118 on Build GRCh38
rs991814042 Chr.5: 78003069-78003069 on Build GRCh38
rs768467649 Chr.5: 78003073-78003073 on Build GRCh38
rs893494247 Chr.5: 78003039-78003039 on Build GRCh38
rs137901104 Chr.5: 78002922-78002922 on Build GRCh38
rs779826277 Chr.5: 78003056-78003057 on Build GRCh38
rs1009753859 Chr.5: 78002670-78002670 on Build GRCh38
rs112214336 Chr.5: 78002861-78002861 on Build GRCh38
rs143654049 Chr.5: 78002983-78002983 on Build GRCh38
rs771988084 Chr.5: 78003198-78003198 on Build GRCh38
rs201198522 Chr.5: 78003077-78003077 on Build GRCh38
rs200738086 Chr.5: 78003062-78003062 on Build GRCh38
rs754678823 Chr.5: 78003076-78003076 on Build GRCh38
rs926831596 Chr.5: 78002730-78002730 on Build GRCh38
rs766132650 Chr.5: 78003099-78003099 on Build GRCh38
rs762840206 Chr.5: 78002989-78002989 on Build GRCh38
rs151028592 Chr.5: 78002933-78002933 on Build GRCh38
rs779663529 Chr.5: 78003006-78003006 on Build GRCh38
rs745888313 Chr.5: 78003011-78003011 on Build GRCh38
rs891132448 Chr.5: 78003226-78003226 on Build GRCh38
rs941977192 Chr.5: 78002847-78002847 on Build GRCh38
rs754574333 Chr.5: 78003101-78003101 on Build GRCh38
rs1021165592 Chr.5: 78002901-78002901 on Build GRCh38
rs933221883 Chr.5: 78003165-78003165 on Build GRCh38
rs199521659 Chr.5: 78002892-78002892 on Build GRCh38
rs757415963 Chr.5: 78003125-78003125 on Build GRCh38

View Full Product Details