Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78002200-78002788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3B1
Entrez Gene ID
8546
Gene Name
adaptor related protein complex 3 beta 1 subunit
Gene Aliases
ADTB3, ADTB3A, HPS, HPS2, PE
UniGene
Hs.532091
Gene Chromosome Location
Chr.5: 78002326-78294755 on Build GRCh38
Transcripts
NM_001271769.1 NM_003664.4 XM_005248618.3 XM_017010001.1

SNP Detail

SNP ID Chromosome Location
rs753338653 Chr.5: 78002443-78002443 on Build GRCh38
rs190285646 Chr.5: 78002720-78002720 on Build GRCh38
rs988271636 Chr.5: 78002340-78002340 on Build GRCh38
rs112166406 Chr.5: 78002232-78002232 on Build GRCh38
rs999162849 Chr.5: 78002337-78002337 on Build GRCh38
rs992209236 Chr.5: 78002757-78002757 on Build GRCh38
rs931449825 Chr.5: 78002485-78002485 on Build GRCh38
rs769645790 Chr.5: 78002596-78002596 on Build GRCh38
rs964499296 Chr.5: 78002299-78002299 on Build GRCh38
rs1048945561 Chr.5: 78002521-78002521 on Build GRCh38
rs1005978093 Chr.5: 78002241-78002241 on Build GRCh38
rs970215199 Chr.5: 78002651-78002651 on Build GRCh38
rs960041888 Chr.5: 78002461-78002461 on Build GRCh38
rs900857521 Chr.5: 78002315-78002315 on Build GRCh38
rs972670538 Chr.5: 78002465-78002465 on Build GRCh38
rs1021123097 Chr.5: 78002684-78002684 on Build GRCh38
rs753283084 Chr.5: 78002761-78002761 on Build GRCh38
rs369987939 Chr.5: 78002719-78002719 on Build GRCh38
rs772075302 Chr.5: 78002233-78002233 on Build GRCh38
rs114954951 Chr.5: 78002314-78002314 on Build GRCh38
rs1038816649 Chr.5: 78002615-78002615 on Build GRCh38
rs759104986 Chr.5: 78002444-78002444 on Build GRCh38
rs1010683230 Chr.5: 78002619-78002619 on Build GRCh38
rs115124715 Chr.5: 78002523-78002523 on Build GRCh38
rs886060767 Chr.5: 78002343-78002346 on Build GRCh38
rs567024530 Chr.5: 78002293-78002293 on Build GRCh38
rs955533391 Chr.5: 78002318-78002318 on Build GRCh38
rs887724674 Chr.5: 78002548-78002548 on Build GRCh38
rs926304522 Chr.5: 78002484-78002484 on Build GRCh38
rs6864605 Chr.5: 78002711-78002711 on Build GRCh38
rs886060768 Chr.5: 78002455-78002455 on Build GRCh38
rs918581877 Chr.5: 78002466-78002466 on Build GRCh38
rs1031175064 Chr.5: 78002342-78002342 on Build GRCh38
rs538570213 Chr.5: 78002420-78002420 on Build GRCh38
rs575976353 Chr.5: 78002614-78002614 on Build GRCh38
rs1006841364 Chr.5: 78002308-78002308 on Build GRCh38
rs911081557 Chr.5: 78002522-78002522 on Build GRCh38
rs559226132 Chr.5: 78002560-78002560 on Build GRCh38
rs957771030 Chr.5: 78002345-78002345 on Build GRCh38
rs552733544 Chr.5: 78002423-78002423 on Build GRCh38
rs1056222654 Chr.5: 78002517-78002517 on Build GRCh38
rs781560568 Chr.5: 78002302-78002302 on Build GRCh38
rs914176898 Chr.5: 78002353-78002355 on Build GRCh38
rs865863660 Chr.5: 78002395-78002395 on Build GRCh38
rs867801817 Chr.5: 78002649-78002649 on Build GRCh38
rs889611344 Chr.5: 78002289-78002289 on Build GRCh38
rs1011067836 Chr.5: 78002370-78002370 on Build GRCh38
rs937789738 Chr.5: 78002516-78002516 on Build GRCh38
rs959070738 Chr.5: 78002735-78002735 on Build GRCh38
rs544830395 Chr.5: 78002654-78002654 on Build GRCh38
rs901056728 Chr.5: 78002546-78002546 on Build GRCh38
rs1048264493 Chr.5: 78002283-78002283 on Build GRCh38
rs899824806 Chr.5: 78002621-78002621 on Build GRCh38
rs928864823 Chr.5: 78002436-78002436 on Build GRCh38
rs1040995881 Chr.5: 78002541-78002541 on Build GRCh38
rs1009753859 Chr.5: 78002670-78002670 on Build GRCh38
rs926831596 Chr.5: 78002730-78002730 on Build GRCh38
rs970615539 Chr.5: 78002403-78002403 on Build GRCh38
rs773135696 Chr.5: 78002597-78002597 on Build GRCh38
rs187770688 Chr.5: 78002498-78002498 on Build GRCh38
rs757339302 Chr.5: 78002258-78002258 on Build GRCh38
rs1804790 Chr.5: 78002490-78002490 on Build GRCh38
rs868442378 Chr.5: 78002650-78002650 on Build GRCh38
rs893452770 Chr.5: 78002617-78002617 on Build GRCh38
rs554959862 Chr.5: 78002487-78002487 on Build GRCh38

View Full Product Details