Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113205763-113206305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4902868 c.570C>A p.G190G Chr.11: 113206125-113206125 on Build GRCh38
428423 c.547G>A p.E183K Chr.11: 113206102-113206102 on Build GRCh38
3443435 c.562G>A p.E188K Chr.11: 113206144-113206144 on Build GRCh38
1475042 c.525C>T p.I175I Chr.11: 113206080-113206080 on Build GRCh38
5535236 c.615C>T p.V205V Chr.11: 113206170-113206170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377417619 Chr.11: 113206194-113206194 on Build GRCh38
rs782517960 Chr.11: 113206203-113206203 on Build GRCh38
rs782242910 Chr.11: 113206136-113206136 on Build GRCh38
rs182204242 Chr.11: 113205911-113205911 on Build GRCh38
rs1025519582 Chr.11: 113205891-113205891 on Build GRCh38
rs369100140 Chr.11: 113206219-113206219 on Build GRCh38
rs782725417 Chr.11: 113205998-113205998 on Build GRCh38
rs531203109 Chr.11: 113205857-113205857 on Build GRCh38
rs369920955 Chr.11: 113206193-113206193 on Build GRCh38
rs781993432 Chr.11: 113206258-113206258 on Build GRCh38
rs782494134 Chr.11: 113206056-113206056 on Build GRCh38
rs782579837 Chr.11: 113206039-113206039 on Build GRCh38
rs1018141243 Chr.11: 113205944-113205944 on Build GRCh38
rs782329784 Chr.11: 113206109-113206109 on Build GRCh38
rs929677262 Chr.11: 113205795-113205795 on Build GRCh38
rs782014259 Chr.11: 113206146-113206146 on Build GRCh38
rs976449844 Chr.11: 113205966-113205966 on Build GRCh38
rs1013583443 Chr.11: 113205874-113205874 on Build GRCh38
rs782807183 Chr.11: 113206016-113206016 on Build GRCh38
rs957849343 Chr.11: 113205916-113205916 on Build GRCh38
rs567763856 Chr.11: 113206001-113206001 on Build GRCh38
rs1975259 Chr.11: 113206260-113206260 on Build GRCh38
rs559320851 Chr.11: 113206200-113206200 on Build GRCh38
rs551301385 Chr.11: 113205863-113205863 on Build GRCh38
rs887914114 Chr.11: 113205830-113205830 on Build GRCh38
rs782525915 Chr.11: 113206022-113206022 on Build GRCh38
rs533974441 Chr.11: 113205928-113205928 on Build GRCh38
rs572624276 Chr.11: 113206237-113206237 on Build GRCh38
rs1006494776 Chr.11: 113205856-113205856 on Build GRCh38
rs918215800 Chr.11: 113205987-113205987 on Build GRCh38
rs186642859 Chr.11: 113205953-113205953 on Build GRCh38
rs556599914 Chr.11: 113206097-113206097 on Build GRCh38
rs782184114 Chr.11: 113206105-113206105 on Build GRCh38
rs782683789 Chr.11: 113206183-113206183 on Build GRCh38
rs782161510 Chr.11: 113206165-113206165 on Build GRCh38
rs929752129 Chr.11: 113205991-113205991 on Build GRCh38
rs534017887 Chr.11: 113205945-113205945 on Build GRCh38
rs782813348 Chr.11: 113206206-113206206 on Build GRCh38
rs782632084 Chr.11: 113206081-113206081 on Build GRCh38
rs782535732 Chr.11: 113206091-113206091 on Build GRCh38
rs999640159 Chr.11: 113206222-113206222 on Build GRCh38
rs781929864 Chr.11: 113206188-113206188 on Build GRCh38
rs782125672 Chr.11: 113206006-113206006 on Build GRCh38
rs35576001 Chr.11: 113206082-113206082 on Build GRCh38
rs576548045 Chr.11: 113206138-113206138 on Build GRCh38
rs781912118 Chr.11: 113206229-113206229 on Build GRCh38
rs1032194277 Chr.11: 113205913-113205913 on Build GRCh38
rs949627474 Chr.11: 113206156-113206156 on Build GRCh38
rs202238052 Chr.11: 113205820-113205820 on Build GRCh38
rs376708434 Chr.11: 113206115-113206115 on Build GRCh38
rs781819663 Chr.11: 113206043-113206043 on Build GRCh38

View Full Product Details