Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 22146902-22147402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC15
Entrez Gene ID
401237
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
UniGene
Hs.712707
Gene Chromosome Location
Chr.6: 21666444-22194400 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

Gene Symbol
NBAT1
Entrez Gene ID
729177
Gene Name
neuroblastoma associated transcript 1
Gene Aliases
CASC14, NBAT-1
UniGene
Hs.730673
Gene Chromosome Location
Chr.6: 22134602-22147193 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

SNP Detail

SNP ID Chromosome Location
rs997461816 Chr.6: 22146926-22146926 on Build GRCh38
rs901857243 Chr.6: 22147118-22147118 on Build GRCh38
rs913801696 Chr.6: 22147372-22147373 on Build GRCh38
rs778220461 Chr.6: 22147128-22147128 on Build GRCh38
rs926413966 Chr.6: 22146974-22146974 on Build GRCh38
rs950690295 Chr.6: 22146935-22146935 on Build GRCh38
rs929499157 Chr.6: 22147057-22147057 on Build GRCh38
rs943364874 Chr.6: 22147084-22147084 on Build GRCh38
rs919306827 Chr.6: 22146933-22146933 on Build GRCh38
rs966525854 Chr.6: 22146937-22146937 on Build GRCh38
rs902470222 Chr.6: 22147378-22147378 on Build GRCh38
rs879606052 Chr.6: 22147155-22147155 on Build GRCh38
rs529457154 Chr.6: 22146987-22146987 on Build GRCh38
rs945030348 Chr.6: 22147014-22147014 on Build GRCh38
rs972025776 Chr.6: 22147025-22147025 on Build GRCh38
rs894924068 Chr.6: 22147170-22147170 on Build GRCh38
rs562156523 Chr.6: 22146963-22146963 on Build GRCh38
rs959026429 Chr.6: 22146960-22146960 on Build GRCh38
rs746720949 Chr.6: 22147078-22147078 on Build GRCh38
rs1025024948 Chr.6: 22147181-22147181 on Build GRCh38
rs1026600222 Chr.6: 22147242-22147242 on Build GRCh38
rs989440299 Chr.6: 22147361-22147361 on Build GRCh38
rs775624694 Chr.6: 22147152-22147152 on Build GRCh38
rs753994564 Chr.6: 22147019-22147019 on Build GRCh38
rs150470887 Chr.6: 22147113-22147113 on Build GRCh38
rs879379143 Chr.6: 22147219-22147219 on Build GRCh38
rs570047123 Chr.6: 22147184-22147184 on Build GRCh38
rs1007231095 Chr.6: 22147028-22147028 on Build GRCh38
rs766333591 Chr.6: 22146991-22146991 on Build GRCh38
rs745662573 Chr.6: 22147121-22147121 on Build GRCh38
rs567520723 Chr.6: 22147273-22147273 on Build GRCh38
rs1021178855 Chr.6: 22146927-22146927 on Build GRCh38
rs779499481 Chr.6: 22147133-22147133 on Build GRCh38
rs923227065 Chr.6: 22147322-22147322 on Build GRCh38
rs933377532 Chr.6: 22147323-22147323 on Build GRCh38
rs370685525 Chr.6: 22147252-22147252 on Build GRCh38
rs138344047 Chr.6: 22147179-22147179 on Build GRCh38
rs974864073 Chr.6: 22147305-22147305 on Build GRCh38
rs1042279183 Chr.6: 22147332-22147332 on Build GRCh38
rs1000689914 Chr.6: 22147160-22147160 on Build GRCh38
rs73375049 Chr.6: 22147072-22147072 on Build GRCh38
rs892165271 Chr.6: 22147002-22147002 on Build GRCh38
rs1032041172 Chr.6: 22147166-22147166 on Build GRCh38
rs184457870 Chr.6: 22146949-22146949 on Build GRCh38
rs879144364 Chr.6: 22147066-22147066 on Build GRCh38
rs768822712 Chr.6: 22147224-22147224 on Build GRCh38
rs899327119 Chr.6: 22147042-22147042 on Build GRCh38
rs755461004 Chr.6: 22147054-22147054 on Build GRCh38
rs749702885 Chr.6: 22147002-22147002 on Build GRCh38
rs933200624 Chr.6: 22147124-22147124 on Build GRCh38
rs950720781 Chr.6: 22147094-22147094 on Build GRCh38
rs980641338 Chr.6: 22146965-22146965 on Build GRCh38
rs755577299 Chr.6: 22147127-22147127 on Build GRCh38
rs757988105 Chr.6: 22147033-22147033 on Build GRCh38
rs1032093123 Chr.6: 22146954-22146954 on Build GRCh38
rs909399115 Chr.6: 22147272-22147272 on Build GRCh38
rs142338126 Chr.6: 22147269-22147269 on Build GRCh38

View Full Product Details