Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49253864-49254121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs782018648 Chr.X: 49253990-49253990 on Build GRCh38
rs782505537 Chr.X: 49253943-49253943 on Build GRCh38
rs782731811 Chr.X: 49253958-49253958 on Build GRCh38
rs782118439 Chr.X: 49253897-49253897 on Build GRCh38
rs782679751 Chr.X: 49254077-49254077 on Build GRCh38
rs782371089 Chr.X: 49254011-49254011 on Build GRCh38
rs1052915981 Chr.X: 49254017-49254017 on Build GRCh38
rs782351468 Chr.X: 49253959-49253959 on Build GRCh38
rs782063296 Chr.X: 49254038-49254038 on Build GRCh38
rs782458039 Chr.X: 49254015-49254015 on Build GRCh38
rs781869510 Chr.X: 49253895-49253895 on Build GRCh38
rs781844954 Chr.X: 49254067-49254067 on Build GRCh38
rs200928046 Chr.X: 49253989-49253989 on Build GRCh38
rs369903891 Chr.X: 49253886-49253886 on Build GRCh38
rs781902347 Chr.X: 49254048-49254048 on Build GRCh38
rs782440849 Chr.X: 49254088-49254088 on Build GRCh38
rs74162067 Chr.X: 49254010-49254010 on Build GRCh38
rs781814163 Chr.X: 49254078-49254078 on Build GRCh38
rs782591107 Chr.X: 49253964-49253964 on Build GRCh38
rs782188307 Chr.X: 49253988-49253988 on Build GRCh38
rs782789484 Chr.X: 49254058-49254058 on Build GRCh38
rs782696973 Chr.X: 49254042-49254042 on Build GRCh38
rs782810691 Chr.X: 49253889-49253889 on Build GRCh38
rs782635427 Chr.X: 49253956-49253956 on Build GRCh38
rs782727861 Chr.X: 49253899-49253899 on Build GRCh38
rs781881326 Chr.X: 49253925-49253925 on Build GRCh38
rs782138295 Chr.X: 49254050-49254050 on Build GRCh38
rs782436363 Chr.X: 49253904-49253904 on Build GRCh38
rs143869281 Chr.X: 49253921-49253921 on Build GRCh38
rs781809477 Chr.X: 49253900-49253900 on Build GRCh38

View Full Product Details