Cosmic ID | Mutation | AA | Transcript Location |
---|---|---|---|
6670006 | c.1133C>T | p.A378V | Chr.7: 44145614-44145614 on Build GRCh38 |
4686621 | c.1102C>T | p.R368C | Chr.7: 44145645-44145645 on Build GRCh38 |
4649115 | c.1210C>T | p.R404C | Chr.7: 44145543-44145543 on Build GRCh38 |
6669999 | c.1080G>A | p.S360S | Chr.7: 44145670-44145670 on Build GRCh38 |
6669980 | c.1066C>T | p.L356L | Chr.7: 44145687-44145687 on Build GRCh38 |
SNP ID | Chromosome Location |
---|---|
rs193922269 | Chr.7: 44145575-44145575 on Build GRCh38 |
rs985667823 | Chr.7: 44145564-44145564 on Build GRCh38 |
rs774614309 | Chr.7: 44145610-44145610 on Build GRCh38 |
rs886042015 | Chr.7: 44145586-44145587 on Build GRCh38 |
rs193929375 | Chr.7: 44145560-44145560 on Build GRCh38 |
rs780987378 | Chr.7: 44145589-44145589 on Build GRCh38 |
rs771131882 | Chr.7: 44145609-44145609 on Build GRCh38 |
rs767892728 | Chr.7: 44145529-44145529 on Build GRCh38 |
rs748092387 | Chr.7: 44145486-44145486 on Build GRCh38 |
rs587780343 | Chr.7: 44145638-44145638 on Build GRCh38 |
rs2908274 | Chr.7: 44145489-44145489 on Build GRCh38 |
rs754337045 | Chr.7: 44145645-44145645 on Build GRCh38 |
rs879059455 | Chr.7: 44145490-44145490 on Build GRCh38 |
rs911506580 | Chr.7: 44145494-44145494 on Build GRCh38 |
rs753053384 | Chr.7: 44145535-44145535 on Build GRCh38 |
rs193922264 | Chr.7: 44145620-44145620 on Build GRCh38 |
rs193922267 | Chr.7: 44145597-44145597 on Build GRCh38 |
rs769645457 | Chr.7: 44145488-44145488 on Build GRCh38 |
rs746719398 | Chr.7: 44145482-44145482 on Build GRCh38 |
rs867810018 | Chr.7: 44145542-44145542 on Build GRCh38 |
rs765849566 | Chr.7: 44145661-44145661 on Build GRCh38 |
rs193922268 | Chr.7: 44145593-44145593 on Build GRCh38 |
rs193922263 | Chr.7: 44145626-44145626 on Build GRCh38 |
rs749298368 | Chr.7: 44145603-44145603 on Build GRCh38 |
rs761204125 | Chr.7: 44145549-44145549 on Build GRCh38 |
rs775856595 | Chr.7: 44145634-44145634 on Build GRCh38 |
rs193922272 | Chr.7: 44145510-44145510 on Build GRCh38 |
rs193922266 | Chr.7: 44145608-44145608 on Build GRCh38 |
rs574954919 | Chr.7: 44145687-44145687 on Build GRCh38 |
rs757888672 | Chr.7: 44145555-44145555 on Build GRCh38 |
rs764575801 | Chr.7: 44145642-44145642 on Build GRCh38 |
rs751179138 | Chr.7: 44145566-44145566 on Build GRCh38 |
rs759783300 | Chr.7: 44145613-44145613 on Build GRCh38 |
rs766653778 | Chr.7: 44145502-44145502 on Build GRCh38 |
rs769709550 | Chr.7: 44145598-44145598 on Build GRCh38 |
rs370464857 | Chr.7: 44145561-44145561 on Build GRCh38 |
rs780716926 | Chr.7: 44145678-44145678 on Build GRCh38 |
rs193922265 | Chr.7: 44145614-44145614 on Build GRCh38 |
rs193922262 | Chr.7: 44145636-44145636 on Build GRCh38 |
rs751087372 | Chr.7: 44145664-44145664 on Build GRCh38 |
rs768491249 | Chr.7: 44145485-44145485 on Build GRCh38 |
rs193921338 | Chr.7: 44145590-44145590 on Build GRCh38 |
rs749877032 | Chr.7: 44145551-44145551 on Build GRCh38 |
rs574763474 | Chr.7: 44145632-44145632 on Build GRCh38 |
rs193922271 | Chr.7: 44145543-44145543 on Build GRCh38 |
rs556581174 | Chr.7: 44145637-44145637 on Build GRCh38 |
rs542306878 | Chr.7: 44145568-44145568 on Build GRCh38 |
rs369103069 | Chr.7: 44145682-44145682 on Build GRCh38 |
rs755112715 | Chr.7: 44145517-44145517 on Build GRCh38 |
rs764512775 | Chr.7: 44145550-44145550 on Build GRCh38 |
rs754623359 | Chr.7: 44145670-44145670 on Build GRCh38 |
rs104894016 | Chr.7: 44145618-44145618 on Build GRCh38 |
rs752481898 | Chr.7: 44145679-44145679 on Build GRCh38 |
rs867232360 | Chr.7: 44145552-44145552 on Build GRCh38 |
rs193921340 | Chr.7: 44145581-44145581 on Build GRCh38 |
rs193929374 | Chr.7: 44145617-44145617 on Build GRCh38 |
rs777870079 | Chr.7: 44145602-44145602 on Build GRCh38 |
rs763133840 | Chr.7: 44145492-44145492 on Build GRCh38 |
rs942941310 | Chr.7: 44145506-44145506 on Build GRCh38 |