Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127462219-127462448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001005374.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4598364 c.520A>T p.T174S Chr.9: 127462365-127462365 on Build GRCh38
1105323 c.448C>T p.R150* Chr.9: 127462293-127462293 on Build GRCh38
6704594 c.481G>A p.E161K Chr.9: 127462326-127462326 on Build GRCh38
302593 c.435C>A p.T145T Chr.9: 127462280-127462280 on Build GRCh38
5757312 c.434C>T p.T145I Chr.9: 127462279-127462279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769164021 Chr.9: 127462385-127462385 on Build GRCh38
rs769461961 Chr.9: 127462418-127462418 on Build GRCh38
rs749635888 Chr.9: 127462378-127462378 on Build GRCh38
rs555377969 Chr.9: 127462281-127462281 on Build GRCh38
rs529333601 Chr.9: 127462359-127462359 on Build GRCh38
rs150292099 Chr.9: 127462314-127462314 on Build GRCh38
rs774722158 Chr.9: 127462386-127462386 on Build GRCh38
rs758039190 Chr.9: 127462388-127462388 on Build GRCh38
rs200259701 Chr.9: 127462241-127462241 on Build GRCh38
rs535476934 Chr.9: 127462275-127462275 on Build GRCh38
rs147205387 Chr.9: 127462325-127462325 on Build GRCh38
rs775848557 Chr.9: 127462285-127462285 on Build GRCh38
rs200115008 Chr.9: 127462294-127462294 on Build GRCh38
rs368689811 Chr.9: 127462303-127462303 on Build GRCh38
rs747087615 Chr.9: 127462244-127462244 on Build GRCh38
rs140143823 Chr.9: 127462299-127462299 on Build GRCh38
rs1039329590 Chr.9: 127462329-127462329 on Build GRCh38
rs757426906 Chr.9: 127462358-127462358 on Build GRCh38
rs775318697 Chr.9: 127462423-127462423 on Build GRCh38
rs772739566 Chr.9: 127462302-127462302 on Build GRCh38
rs756516100 Chr.9: 127462363-127462363 on Build GRCh38
rs367955076 Chr.9: 127462400-127462400 on Build GRCh38
rs759239338 Chr.9: 127462415-127462415 on Build GRCh38
rs200356981 Chr.9: 127462306-127462306 on Build GRCh38
rs770348926 Chr.9: 127462283-127462283 on Build GRCh38
rs746159728 Chr.9: 127462362-127462362 on Build GRCh38
rs781577020 Chr.9: 127462280-127462280 on Build GRCh38
rs140809697 Chr.9: 127462343-127462343 on Build GRCh38
rs138001044 Chr.9: 127462318-127462318 on Build GRCh38
rs759326900 Chr.9: 127462319-127462319 on Build GRCh38
rs1014986581 Chr.9: 127462379-127462379 on Build GRCh38
rs916806387 Chr.9: 127462242-127462242 on Build GRCh38
rs946372444 Chr.9: 127462326-127462326 on Build GRCh38
rs1010951527 Chr.9: 127462350-127462350 on Build GRCh38
rs780515350 Chr.9: 127462367-127462367 on Build GRCh38
rs749850642 Chr.9: 127462293-127462293 on Build GRCh38
rs765061936 Chr.9: 127462322-127462322 on Build GRCh38
rs777946282 Chr.9: 127462240-127462240 on Build GRCh38
rs151323851 Chr.9: 127462259-127462259 on Build GRCh38
rs142085060 Chr.9: 127462342-127462342 on Build GRCh38

View Full Product Details