Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226888872-226889107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSEN2
Entrez Gene ID
5664
Gene Name
presenilin 2
Gene Aliases
AD3L, AD4, CMD1V, PS2, STM2
UniGene
Hs.25363
Gene Chromosome Location
Chr.1: 226870572-226903829 on Build GRCh38
Transcripts
NM_000447.2 NM_012486.2 XM_005273199.3 XM_017001835.1 XM_017001836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5215817 c.854C>T p.A285V Chr.1: 226889017-226889017 on Build GRCh38
1320438 c.763A>T p.I255F Chr.1: 226889025-226889025 on Build GRCh38
6282429 c.715C>T p.L239L Chr.1: 226888878-226888878 on Build GRCh38
904774 c.701T>C p.M234T Chr.1: 226888963-226888963 on Build GRCh38
1339321 c.640G>A p.V214M Chr.1: 226888902-226888902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777235854 Chr.1: 226888998-226888998 on Build GRCh38
rs201908554 Chr.1: 226888959-226888959 on Build GRCh38
rs200173519 Chr.1: 226888896-226888896 on Build GRCh38
rs766159571 Chr.1: 226889031-226889031 on Build GRCh38
rs932889540 Chr.1: 226889079-226889079 on Build GRCh38
rs28936379 Chr.1: 226888977-226888977 on Build GRCh38
rs574125890 Chr.1: 226888902-226888902 on Build GRCh38
rs201292728 Chr.1: 226889025-226889025 on Build GRCh38
rs145400391 Chr.1: 226889006-226889006 on Build GRCh38
rs61730652 Chr.1: 226888970-226888970 on Build GRCh38
rs760698293 Chr.1: 226889027-226889027 on Build GRCh38
rs190477948 Chr.1: 226888904-226888904 on Build GRCh38
rs202003390 Chr.1: 226888973-226888973 on Build GRCh38
rs560839429 Chr.1: 226889073-226889073 on Build GRCh38
rs200754713 Chr.1: 226888954-226888954 on Build GRCh38
rs201403206 Chr.1: 226889017-226889017 on Build GRCh38
rs7539017 Chr.1: 226889033-226889033 on Build GRCh38
rs140562931 Chr.1: 226889001-226889001 on Build GRCh38
rs199760083 Chr.1: 226888895-226888895 on Build GRCh38
rs201563376 Chr.1: 226889087-226889087 on Build GRCh38
rs754569211 Chr.1: 226888954-226888955 on Build GRCh38
rs757955974 Chr.1: 226888994-226888994 on Build GRCh38
rs566471708 Chr.1: 226889052-226889052 on Build GRCh38
rs369429245 Chr.1: 226889032-226889032 on Build GRCh38
rs773410250 Chr.1: 226889022-226889022 on Build GRCh38
rs138836272 Chr.1: 226889016-226889016 on Build GRCh38
rs768941393 Chr.1: 226889077-226889077 on Build GRCh38
rs145010538 Chr.1: 226888952-226888952 on Build GRCh38
rs371562367 Chr.1: 226888991-226888991 on Build GRCh38
rs200332829 Chr.1: 226889015-226889015 on Build GRCh38
rs63749884 Chr.1: 226888979-226888979 on Build GRCh38
rs63750880 Chr.1: 226888945-226888945 on Build GRCh38
rs751014475 Chr.1: 226889057-226889057 on Build GRCh38
rs148238688 Chr.1: 226889034-226889034 on Build GRCh38
rs200670135 Chr.1: 226888972-226888972 on Build GRCh38
rs555809980 Chr.1: 226888901-226888901 on Build GRCh38
rs764165210 Chr.1: 226888937-226888937 on Build GRCh38
rs200133314 Chr.1: 226888987-226888987 on Build GRCh38
rs367855127 Chr.1: 226888974-226888974 on Build GRCh38
rs750424014 Chr.1: 226888961-226888961 on Build GRCh38
rs777323657 Chr.1: 226889056-226889056 on Build GRCh38
rs200409502 Chr.1: 226889086-226889086 on Build GRCh38
rs778393774 Chr.1: 226888980-226888980 on Build GRCh38
rs779414075 Chr.1: 226889083-226889083 on Build GRCh38
rs781053654 Chr.1: 226889043-226889043 on Build GRCh38
rs756649903 Chr.1: 226889058-226889058 on Build GRCh38
rs202210060 Chr.1: 226889045-226889045 on Build GRCh38
rs780375882 Chr.1: 226889071-226889071 on Build GRCh38
rs748437025 Chr.1: 226889085-226889085 on Build GRCh38
rs748521246 Chr.1: 226889080-226889081 on Build GRCh38
rs201055590 Chr.1: 226889043-226889043 on Build GRCh38
rs763295042 Chr.1: 226888936-226888936 on Build GRCh38
rs147702142 Chr.1: 226889018-226889018 on Build GRCh38

View Full Product Details