Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10643183-10643420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_005263479.3 XM_017001539.1 XM_017001540.1 XM_017001541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4020087 c.3964C>T p.R1322W Chr.1: 10643216-10643216 on Build GRCh38
4406071 c.3929C>T p.S1310F Chr.1: 10643251-10643251 on Build GRCh38
893277 c.3911G>A p.R1304Q Chr.1: 10643269-10643269 on Build GRCh38
5458165 c.3869-1G>T p.? Chr.1: 10643312-10643312 on Build GRCh38
5776096 c.3922C>G p.Q1308E Chr.1: 10643258-10643258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760839890 Chr.1: 10643337-10643337 on Build GRCh38
rs770575960 Chr.1: 10643264-10643264 on Build GRCh38
rs987725969 Chr.1: 10643272-10643272 on Build GRCh38
rs372548483 Chr.1: 10643218-10643218 on Build GRCh38
rs199630871 Chr.1: 10643204-10643204 on Build GRCh38
rs1005879260 Chr.1: 10643397-10643397 on Build GRCh38
rs548153299 Chr.1: 10643268-10643268 on Build GRCh38
rs377596799 Chr.1: 10643329-10643329 on Build GRCh38
rs771812114 Chr.1: 10643232-10643232 on Build GRCh38
rs755353824 Chr.1: 10643220-10643220 on Build GRCh38
rs768161756 Chr.1: 10643249-10643249 on Build GRCh38
rs1047483937 Chr.1: 10643381-10643381 on Build GRCh38
rs1018748331 Chr.1: 10643289-10643289 on Build GRCh38
rs763883758 Chr.1: 10643310-10643310 on Build GRCh38
rs750585047 Chr.1: 10643203-10643203 on Build GRCh38
rs188045266 Chr.1: 10643217-10643217 on Build GRCh38
rs762653267 Chr.1: 10643286-10643286 on Build GRCh38
rs769832846 Chr.1: 10643269-10643269 on Build GRCh38
rs532843592 Chr.1: 10643247-10643247 on Build GRCh38
rs753170543 Chr.1: 10643362-10643362 on Build GRCh38
rs777283250 Chr.1: 10643238-10643238 on Build GRCh38
rs753123249 Chr.1: 10643363-10643363 on Build GRCh38
rs780053498 Chr.1: 10643216-10643216 on Build GRCh38
rs765714885 Chr.1: 10643356-10643356 on Build GRCh38
rs750626895 Chr.1: 10643311-10643311 on Build GRCh38
rs779286691 Chr.1: 10643219-10643219 on Build GRCh38
rs146914257 Chr.1: 10643288-10643289 on Build GRCh38
rs886151981 Chr.1: 10643382-10643382 on Build GRCh38
rs114268069 Chr.1: 10643346-10643346 on Build GRCh38
rs753888325 Chr.1: 10643350-10643350 on Build GRCh38
rs376782639 Chr.1: 10643243-10643243 on Build GRCh38
rs775413063 Chr.1: 10643270-10643270 on Build GRCh38
rs776231987 Chr.1: 10643267-10643267 on Build GRCh38
rs755524004 Chr.1: 10643355-10643355 on Build GRCh38

View Full Product Details