Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73159234-73159507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2 XM_011518609.1 XM_017014657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6287220 c.184G>A p.E62K Chr.9: 73159337-73159337 on Build GRCh38
5411231 c.191C>T p.T64I Chr.9: 73159344-73159344 on Build GRCh38
4421406 c.277delG p.A93fs*15 Chr.9: 73159397-73159397 on Build GRCh38
5811299 c.269A>T p.Q90L Chr.9: 73159389-73159389 on Build GRCh38
1756213 c.288C>T p.L96L Chr.9: 73159408-73159408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182642121 Chr.9: 73159432-73159432 on Build GRCh38
rs565435361 Chr.9: 73159261-73159261 on Build GRCh38
rs912547303 Chr.9: 73159278-73159278 on Build GRCh38
rs764404494 Chr.9: 73159440-73159440 on Build GRCh38
rs1020609364 Chr.9: 73159478-73159478 on Build GRCh38
rs781132592 Chr.9: 73159306-73159306 on Build GRCh38
rs370059473 Chr.9: 73159462-73159462 on Build GRCh38
rs769885279 Chr.9: 73159315-73159315 on Build GRCh38
rs745959338 Chr.9: 73159418-73159418 on Build GRCh38
rs536986668 Chr.9: 73159382-73159382 on Build GRCh38
rs755924071 Chr.9: 73159392-73159392 on Build GRCh38
rs769562342 Chr.9: 73159309-73159309 on Build GRCh38
rs10114350 Chr.9: 73159322-73159322 on Build GRCh38
rs758013277 Chr.9: 73159375-73159375 on Build GRCh38
rs765972811 Chr.9: 73159380-73159380 on Build GRCh38
rs890867197 Chr.9: 73159436-73159436 on Build GRCh38
rs749513170 Chr.9: 73159317-73159317 on Build GRCh38
rs771311606 Chr.9: 73159402-73159402 on Build GRCh38
rs746986725 Chr.9: 73159298-73159298 on Build GRCh38
rs777604252 Chr.9: 73159394-73159394 on Build GRCh38
rs774669199 Chr.9: 73159407-73159407 on Build GRCh38
rs750091835 Chr.9: 73159368-73159368 on Build GRCh38
rs759650458 Chr.9: 73159332-73159332 on Build GRCh38
rs76459132 Chr.9: 73159367-73159367 on Build GRCh38
rs776007675 Chr.9: 73159351-73159351 on Build GRCh38
rs55652728 Chr.9: 73159290-73159290 on Build GRCh38
rs773376298 Chr.9: 73159314-73159314 on Build GRCh38
rs749564615 Chr.9: 73159397-73159397 on Build GRCh38
rs200599744 Chr.9: 73159422-73159422 on Build GRCh38
rs1023869180 Chr.9: 73159417-73159417 on Build GRCh38
rs762113579 Chr.9: 73159456-73159456 on Build GRCh38
rs75297127 Chr.9: 73159300-73159300 on Build GRCh38
rs774544645 Chr.9: 73159331-73159331 on Build GRCh38
rs116080195 Chr.9: 73159390-73159390 on Build GRCh38
rs942270992 Chr.9: 73159330-73159330 on Build GRCh38
rs11557057 Chr.9: 73159339-73159339 on Build GRCh38
rs144842081 Chr.9: 73159383-73159383 on Build GRCh38
rs376007168 Chr.9: 73159452-73159452 on Build GRCh38
rs757802790 Chr.9: 73159389-73159389 on Build GRCh38
rs760952664 Chr.9: 73159360-73159360 on Build GRCh38
rs767933861 Chr.9: 73159350-73159350 on Build GRCh38
rs777010678 Chr.9: 73159454-73159454 on Build GRCh38
rs780992950 Chr.9: 73159388-73159388 on Build GRCh38

View Full Product Details