Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112961501-112961774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928847
Entrez Gene ID
101928847
Gene Name
uncharacterized LOC101928847
Gene Aliases
-
UniGene
Hs.711235
Gene Chromosome Location
Chr.11: 112959279-112963460 on Build GRCh38
Transcripts
NR_120563.1 NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NR_120563.1 NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

SNP Detail

SNP ID Chromosome Location
rs561424527 Chr.11: 112961564-112961564 on Build GRCh38
rs782768191 Chr.11: 112961556-112961557 on Build GRCh38
rs782225336 Chr.11: 112961617-112961617 on Build GRCh38
rs6589347 Chr.11: 112961636-112961636 on Build GRCh38
rs201206514 Chr.11: 112961577-112961577 on Build GRCh38
rs11381910 Chr.11: 112961585-112961586 on Build GRCh38
rs782527116 Chr.11: 112961531-112961531 on Build GRCh38
rs1048810733 Chr.11: 112961554-112961554 on Build GRCh38
rs112306738 Chr.11: 112961619-112961619 on Build GRCh38
rs782245249 Chr.11: 112961562-112961562 on Build GRCh38
rs199895300 Chr.11: 112961559-112961559 on Build GRCh38
rs781801429 Chr.11: 112961595-112961595 on Build GRCh38
rs782563224 Chr.11: 112961587-112961588 on Build GRCh38
rs572184546 Chr.11: 112961556-112961556 on Build GRCh38
rs543798793 Chr.11: 112961604-112961604 on Build GRCh38
rs764276396 Chr.11: 112961683-112961684 on Build GRCh38
rs782674091 Chr.11: 112961535-112961535 on Build GRCh38
rs782795743 Chr.11: 112961625-112961625 on Build GRCh38
rs967382506 Chr.11: 112961576-112961576 on Build GRCh38
rs782311774 Chr.11: 112961555-112961555 on Build GRCh38
rs781792212 Chr.11: 112961558-112961559 on Build GRCh38
rs782254487 Chr.11: 112961649-112961649 on Build GRCh38
rs781963746 Chr.11: 112961692-112961692 on Build GRCh38
rs782102717 Chr.11: 112961697-112961697 on Build GRCh38
rs559828324 Chr.11: 112961639-112961639 on Build GRCh38
rs782336141 Chr.11: 112961688-112961688 on Build GRCh38
rs782062091 Chr.11: 112961555-112961555 on Build GRCh38
rs782602069 Chr.11: 112961526-112961526 on Build GRCh38
rs782605842 Chr.11: 112961559-112961560 on Build GRCh38
rs781852393 Chr.11: 112961557-112961558 on Build GRCh38
rs782323298 Chr.11: 112961565-112961565 on Build GRCh38
rs782151247 Chr.11: 112961584-112961584 on Build GRCh38
rs374931900 Chr.11: 112961561-112961561 on Build GRCh38
rs781962609 Chr.11: 112961555-112961556 on Build GRCh38
rs7105734 Chr.11: 112961631-112961631 on Build GRCh38
rs1033096683 Chr.11: 112961587-112961587 on Build GRCh38
rs781985812 Chr.11: 112961557-112961557 on Build GRCh38
rs73002342 Chr.11: 112961683-112961683 on Build GRCh38
rs782109341 Chr.11: 112961670-112961670 on Build GRCh38
rs558440771 Chr.11: 112961538-112961538 on Build GRCh38
rs782499528 Chr.11: 112961622-112961622 on Build GRCh38
rs563686839 Chr.11: 112961616-112961616 on Build GRCh38
rs201772924 Chr.11: 112961618-112961618 on Build GRCh38
rs782430578 Chr.11: 112961554-112961554 on Build GRCh38
rs782087726 Chr.11: 112961578-112961578 on Build GRCh38
rs796286007 Chr.11: 112961619-112961619 on Build GRCh38
rs782430131 Chr.11: 112961669-112961670 on Build GRCh38
rs782396666 Chr.11: 112961669-112961669 on Build GRCh38
rs782039043 Chr.11: 112961680-112961680 on Build GRCh38
rs782282074 Chr.11: 112961564-112961565 on Build GRCh38
rs145493999 Chr.11: 112961554-112961555 on Build GRCh38
rs782517564 Chr.11: 112961560-112961561 on Build GRCh38
rs781802202 Chr.11: 112961520-112961520 on Build GRCh38
rs782816995 Chr.11: 112961593-112961593 on Build GRCh38
rs781994961 Chr.11: 112961582-112961582 on Build GRCh38
rs782762564 Chr.11: 112961705-112961705 on Build GRCh38
rs782477221 Chr.11: 112961525-112961525 on Build GRCh38
rs782374331 Chr.11: 112961580-112961580 on Build GRCh38
rs201161826 Chr.11: 112961648-112961648 on Build GRCh38

View Full Product Details