Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44158978-44159215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCK
Entrez Gene ID
2645
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
UniGene
Hs.1270
Gene Chromosome Location
Chr.7: 44143213-44189439 on Build GRCh38
Transcripts
NM_000162.3 NM_033507.1 NM_033508.1 XM_017011966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
166954 c.5C>T p.A2V Chr.7: 44159116-44159116 on Build GRCh38
23395 c.29C>T p.A10V Chr.7: 44159092-44159092 on Build GRCh38
1089928 c.30C>A p.A10A Chr.7: 44159091-44159091 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368776779 Chr.7: 44159103-44159103 on Build GRCh38
rs770144386 Chr.7: 44159077-44159077 on Build GRCh38
rs908956460 Chr.7: 44159189-44159189 on Build GRCh38
rs760138967 Chr.7: 44159109-44159109 on Build GRCh38
rs199605648 Chr.7: 44159042-44159042 on Build GRCh38
rs771198677 Chr.7: 44159065-44159065 on Build GRCh38
rs1005902597 Chr.7: 44159018-44159018 on Build GRCh38
rs372259487 Chr.7: 44159126-44159126 on Build GRCh38
rs75119141 Chr.7: 44159057-44159057 on Build GRCh38
rs773304385 Chr.7: 44159158-44159158 on Build GRCh38
rs748340904 Chr.7: 44159067-44159067 on Build GRCh38
rs778640409 Chr.7: 44159038-44159038 on Build GRCh38
rs540471004 Chr.7: 44159177-44159177 on Build GRCh38
rs886075153 Chr.7: 44159001-44159001 on Build GRCh38
rs1012851036 Chr.7: 44159068-44159068 on Build GRCh38
rs141892872 Chr.7: 44159029-44159029 on Build GRCh38
rs776654918 Chr.7: 44159154-44159154 on Build GRCh38
rs754996159 Chr.7: 44159147-44159147 on Build GRCh38
rs191599719 Chr.7: 44159144-44159144 on Build GRCh38
rs13306393 Chr.7: 44159061-44159061 on Build GRCh38
rs763215868 Chr.7: 44159086-44159086 on Build GRCh38
rs555679654 Chr.7: 44159022-44159022 on Build GRCh38
rs146701164 Chr.7: 44159121-44159121 on Build GRCh38
rs750154659 Chr.7: 44159129-44159129 on Build GRCh38
rs150560724 Chr.7: 44159165-44159165 on Build GRCh38
rs745657165 Chr.7: 44159041-44159041 on Build GRCh38
rs779850352 Chr.7: 44159049-44159049 on Build GRCh38
rs770054616 Chr.7: 44159156-44159156 on Build GRCh38
rs374650442 Chr.7: 44159151-44159151 on Build GRCh38
rs368266661 Chr.7: 44159055-44159055 on Build GRCh38
rs763550503 Chr.7: 44159115-44159115 on Build GRCh38
rs753381934 Chr.7: 44159116-44159116 on Build GRCh38
rs774691156 Chr.7: 44159107-44159107 on Build GRCh38
rs578238909 Chr.7: 44159127-44159127 on Build GRCh38
rs534011553 Chr.7: 44159021-44159021 on Build GRCh38
rs1041341149 Chr.7: 44159190-44159190 on Build GRCh38
rs73314180 Chr.7: 44159164-44159164 on Build GRCh38
rs367774728 Chr.7: 44159155-44159155 on Build GRCh38
rs779944143 Chr.7: 44159139-44159139 on Build GRCh38

View Full Product Details