Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38299188-38299459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2268611 c.66G>A p.M22I Chr.15: 38299406-38299406 on Build GRCh38
6789280 c.93G>A p.W31* Chr.15: 38299433-38299433 on Build GRCh38
2268610 c.43G>T p.A15S Chr.15: 38299383-38299383 on Build GRCh38
3386841 c.66G>T p.M22I Chr.15: 38299406-38299406 on Build GRCh38
5016883 c.58G>C p.V20L Chr.15: 38299398-38299398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781022987 Chr.15: 38299295-38299295 on Build GRCh38
rs780614363 Chr.15: 38299398-38299398 on Build GRCh38
rs757623945 Chr.15: 38299231-38299231 on Build GRCh38
rs1057518150 Chr.15: 38299411-38299411 on Build GRCh38
rs864622421 Chr.15: 38299391-38299391 on Build GRCh38
rs142406055 Chr.15: 38299294-38299294 on Build GRCh38
rs986112492 Chr.15: 38299244-38299244 on Build GRCh38
rs367745143 Chr.15: 38299400-38299400 on Build GRCh38
rs1057517941 Chr.15: 38299386-38299386 on Build GRCh38
rs572710790 Chr.15: 38299259-38299259 on Build GRCh38
rs914544937 Chr.15: 38299258-38299258 on Build GRCh38
rs774071345 Chr.15: 38299325-38299325 on Build GRCh38
rs887742500 Chr.15: 38299249-38299249 on Build GRCh38
rs121434313 Chr.15: 38299410-38299410 on Build GRCh38
rs200789030 Chr.15: 38299331-38299331 on Build GRCh38
rs947392765 Chr.15: 38299272-38299272 on Build GRCh38
rs751895777 Chr.15: 38299219-38299219 on Build GRCh38
rs1006222486 Chr.15: 38299299-38299299 on Build GRCh38
rs750777752 Chr.15: 38299392-38299392 on Build GRCh38
rs144515463 Chr.15: 38299323-38299324 on Build GRCh38
rs755788325 Chr.15: 38299409-38299409 on Build GRCh38
rs760850290 Chr.15: 38299366-38299366 on Build GRCh38
rs758855467 Chr.15: 38299393-38299393 on Build GRCh38
rs775671940 Chr.15: 38299362-38299362 on Build GRCh38
rs769323362 Chr.15: 38299303-38299303 on Build GRCh38
rs749241525 Chr.15: 38299312-38299312 on Build GRCh38
rs757726772 Chr.15: 38299387-38299387 on Build GRCh38
rs968982670 Chr.15: 38299396-38299396 on Build GRCh38
rs746572586 Chr.15: 38299307-38299308 on Build GRCh38
rs764234046 Chr.15: 38299382-38299382 on Build GRCh38
rs900576451 Chr.15: 38299341-38299341 on Build GRCh38
rs1042234072 Chr.15: 38299311-38299311 on Build GRCh38
rs772636310 Chr.15: 38299308-38299308 on Build GRCh38
rs753924528 Chr.15: 38299383-38299383 on Build GRCh38
rs148520129 Chr.15: 38299324-38299325 on Build GRCh38
rs1057517922 Chr.15: 38299378-38299378 on Build GRCh38
rs767682280 Chr.15: 38299356-38299356 on Build GRCh38

View Full Product Details