Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46662842-46663097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6820564 c.1790A>T p.E597V Chr.16: 46663020-46663020 on Build GRCh38
3817994 c.1807G>A p.A603T Chr.16: 46663003-46663003 on Build GRCh38
3509662 c.1759G>A p.G587R Chr.16: 46663051-46663051 on Build GRCh38
3888528 c.1713G>A p.L571L Chr.16: 46663097-46663097 on Build GRCh38
7170399 c.1756C>G p.Q586E Chr.16: 46663054-46663054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762907744 Chr.16: 46662973-46662974 on Build GRCh38
rs142063738 Chr.16: 46663001-46663001 on Build GRCh38
rs148255605 Chr.16: 46663015-46663015 on Build GRCh38
rs201566773 Chr.16: 46663027-46663027 on Build GRCh38
rs145679337 Chr.16: 46663007-46663007 on Build GRCh38
rs765675580 Chr.16: 46662965-46662965 on Build GRCh38
rs113527787 Chr.16: 46662896-46662896 on Build GRCh38
rs750804781 Chr.16: 46662978-46662978 on Build GRCh38
rs945640037 Chr.16: 46662866-46662866 on Build GRCh38
rs755145722 Chr.16: 46663004-46663004 on Build GRCh38
rs572202909 Chr.16: 46662902-46662902 on Build GRCh38
rs749034028 Chr.16: 46662949-46662949 on Build GRCh38
rs770731019 Chr.16: 46662951-46662951 on Build GRCh38
rs903550909 Chr.16: 46662920-46662920 on Build GRCh38
rs770819201 Chr.16: 46663034-46663034 on Build GRCh38
rs34687100 Chr.16: 46663005-46663005 on Build GRCh38
rs745500235 Chr.16: 46663069-46663069 on Build GRCh38
rs752760936 Chr.16: 46663010-46663010 on Build GRCh38
rs750236573 Chr.16: 46662959-46662959 on Build GRCh38
rs762311864 Chr.16: 46662962-46662962 on Build GRCh38
rs541423152 Chr.16: 46662879-46662879 on Build GRCh38
rs777261458 Chr.16: 46662961-46662961 on Build GRCh38
rs892692478 Chr.16: 46662863-46662863 on Build GRCh38
rs769065728 Chr.16: 46662899-46662899 on Build GRCh38
rs771468030 Chr.16: 46662957-46662957 on Build GRCh38
rs763261143 Chr.16: 46662979-46662979 on Build GRCh38
rs77476733 Chr.16: 46662878-46662878 on Build GRCh38
rs773836193 Chr.16: 46662954-46662954 on Build GRCh38
rs562254823 Chr.16: 46662872-46662872 on Build GRCh38
rs868056476 Chr.16: 46663003-46663003 on Build GRCh38
rs749127069 Chr.16: 46663031-46663031 on Build GRCh38
rs896243488 Chr.16: 46663006-46663006 on Build GRCh38
rs956063150 Chr.16: 46662972-46662972 on Build GRCh38
rs778678166 Chr.16: 46663064-46663064 on Build GRCh38
rs759127576 Chr.16: 46662955-46662955 on Build GRCh38

View Full Product Details