Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11956880-11957145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6750269 c.688C>T p.L230F Chr.1: 11957134-11957134 on Build GRCh38
7410769 c.672C>T p.G224G Chr.1: 11956945-11956945 on Build GRCh38
267197 c.676G>A p.V226M Chr.1: 11956949-11956949 on Build GRCh38
6583998 c.828G>A p.R276R Chr.1: 11956960-11956960 on Build GRCh38
5556406 c.816T>C p.H272H Chr.1: 11956948-11956948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375303994 Chr.1: 11956906-11956906 on Build GRCh38
rs776599201 Chr.1: 11956923-11956923 on Build GRCh38
rs777914756 Chr.1: 11956985-11956985 on Build GRCh38
rs563861250 Chr.1: 11956910-11956910 on Build GRCh38
rs535012522 Chr.1: 11956976-11956976 on Build GRCh38
rs372534520 Chr.1: 11956925-11956925 on Build GRCh38
rs749221307 Chr.1: 11957113-11957113 on Build GRCh38
rs756600025 Chr.1: 11956956-11956956 on Build GRCh38
rs762431149 Chr.1: 11957013-11957013 on Build GRCh38
rs772128366 Chr.1: 11956914-11956914 on Build GRCh38
rs376118516 Chr.1: 11957058-11957058 on Build GRCh38
rs766873556 Chr.1: 11957034-11957034 on Build GRCh38
rs566018612 Chr.1: 11956975-11956975 on Build GRCh38
rs138253757 Chr.1: 11957091-11957091 on Build GRCh38
rs747258497 Chr.1: 11957064-11957064 on Build GRCh38
rs369038766 Chr.1: 11956915-11956915 on Build GRCh38
rs373236197 Chr.1: 11957101-11957101 on Build GRCh38
rs927138502 Chr.1: 11957042-11957042 on Build GRCh38
rs769659116 Chr.1: 11957003-11957003 on Build GRCh38
rs778818723 Chr.1: 11956901-11956901 on Build GRCh38
rs747209460 Chr.1: 11956988-11956988 on Build GRCh38
rs748111731 Chr.1: 11956983-11956983 on Build GRCh38
rs1007583680 Chr.1: 11956912-11956912 on Build GRCh38
rs150852515 Chr.1: 11956921-11956921 on Build GRCh38
rs755548816 Chr.1: 11957052-11957052 on Build GRCh38
rs760452720 Chr.1: 11956917-11956917 on Build GRCh38
rs41307745 Chr.1: 11956900-11956900 on Build GRCh38
rs149566879 Chr.1: 11956984-11956984 on Build GRCh38
rs751215041 Chr.1: 11957021-11957021 on Build GRCh38
rs34918895 Chr.1: 11957118-11957119 on Build GRCh38
rs776366598 Chr.1: 11956999-11956999 on Build GRCh38
rs768087078 Chr.1: 11957117-11957117 on Build GRCh38
rs200554697 Chr.1: 11957044-11957044 on Build GRCh38
rs761517804 Chr.1: 11957025-11957025 on Build GRCh38
rs755109500 Chr.1: 11956898-11956899 on Build GRCh38
rs183225496 Chr.1: 11956957-11956957 on Build GRCh38
rs780975714 Chr.1: 11957082-11957082 on Build GRCh38
rs749794366 Chr.1: 11956899-11956899 on Build GRCh38
rs950451933 Chr.1: 11956962-11956962 on Build GRCh38
rs765728620 Chr.1: 11957053-11957053 on Build GRCh38
rs938483658 Chr.1: 11957044-11957044 on Build GRCh38
rs370781041 Chr.1: 11957059-11957059 on Build GRCh38
rs763724588 Chr.1: 11956943-11956943 on Build GRCh38
rs759435283 Chr.1: 11956924-11956924 on Build GRCh38
rs538823621 Chr.1: 11957089-11957089 on Build GRCh38
rs377072582 Chr.1: 11956941-11956941 on Build GRCh38
rs371007185 Chr.1: 11956951-11956951 on Build GRCh38
rs773949948 Chr.1: 11957122-11957122 on Build GRCh38
rs775496572 Chr.1: 11957009-11957009 on Build GRCh38
rs373584054 Chr.1: 11957055-11957055 on Build GRCh38
rs763641520 Chr.1: 11957018-11957018 on Build GRCh38
rs757418080 Chr.1: 11957067-11957067 on Build GRCh38
rs770749842 Chr.1: 11956993-11956993 on Build GRCh38
rs567164584 Chr.1: 11957002-11957002 on Build GRCh38
rs376643174 Chr.1: 11956950-11956950 on Build GRCh38
rs779066341 Chr.1: 11956979-11956979 on Build GRCh38

View Full Product Details