Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108874792-108875049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
UniGene
Hs.494738
Gene Chromosome Location
Chr.9: 108867520-108934332 on Build GRCh38
Transcripts
NM_001318360.1 NM_003640.4 XM_011519136.1 XM_011519137.1 XM_017015238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3168840 c.3887T>C p.I1296T Chr.9: 108874939-108874939 on Build GRCh38
5014867 c.3875C>G p.T1292S Chr.9: 108874951-108874951 on Build GRCh38
4637212 c.3901C>A p.Q1301K Chr.9: 108874925-108874925 on Build GRCh38
4545790 c.3863G>A p.G1288D Chr.9: 108874963-108874963 on Build GRCh38
6504909 c.3896C>T p.S1299F Chr.9: 108874930-108874930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945633667 Chr.9: 108874952-108874952 on Build GRCh38
rs931033803 Chr.9: 108875025-108875025 on Build GRCh38
rs765183062 Chr.9: 108874989-108874989 on Build GRCh38
rs762978201 Chr.9: 108874847-108874847 on Build GRCh38
rs762129560 Chr.9: 108874988-108874988 on Build GRCh38
rs145594625 Chr.9: 108874857-108874857 on Build GRCh38
rs372894892 Chr.9: 108875010-108875010 on Build GRCh38
rs758104586 Chr.9: 108874874-108874875 on Build GRCh38
rs750858926 Chr.9: 108874897-108874897 on Build GRCh38
rs143674809 Chr.9: 108874894-108874894 on Build GRCh38
rs879250768 Chr.9: 108874959-108874959 on Build GRCh38
rs753943910 Chr.9: 108874983-108874983 on Build GRCh38
rs769146127 Chr.9: 108874937-108874937 on Build GRCh38
rs772694065 Chr.9: 108874878-108874878 on Build GRCh38
rs200524123 Chr.9: 108874923-108874923 on Build GRCh38
rs747419554 Chr.9: 108874906-108874906 on Build GRCh38
rs867431578 Chr.9: 108874917-108874917 on Build GRCh38
rs780649768 Chr.9: 108874909-108874909 on Build GRCh38
rs149721955 Chr.9: 108874856-108874856 on Build GRCh38
rs61749202 Chr.9: 108874950-108874950 on Build GRCh38
rs145319352 Chr.9: 108874957-108874957 on Build GRCh38
rs959983712 Chr.9: 108875001-108875001 on Build GRCh38
rs762773957 Chr.9: 108874940-108874940 on Build GRCh38
rs764363951 Chr.9: 108874879-108874879 on Build GRCh38
rs751098040 Chr.9: 108874984-108874997 on Build GRCh38
rs555520875 Chr.9: 108874881-108874881 on Build GRCh38
rs146862778 Chr.9: 108874908-108874908 on Build GRCh38
rs747840196 Chr.9: 108874930-108874930 on Build GRCh38
rs750564744 Chr.9: 108874996-108874996 on Build GRCh38
rs757229823 Chr.9: 108874891-108874891 on Build GRCh38
rs989681376 Chr.9: 108874943-108874943 on Build GRCh38
rs767316416 Chr.9: 108874961-108874961 on Build GRCh38
rs777103049 Chr.9: 108874924-108874926 on Build GRCh38
rs750055987 Chr.9: 108874903-108874903 on Build GRCh38
rs1022019183 Chr.9: 108874925-108874925 on Build GRCh38
rs772800681 Chr.9: 108874938-108874938 on Build GRCh38
rs567612710 Chr.9: 108875005-108875005 on Build GRCh38
rs753943169 Chr.9: 108874883-108874883 on Build GRCh38
rs758997539 Chr.9: 108874901-108874901 on Build GRCh38
rs770891146 Chr.9: 108874946-108874946 on Build GRCh38
rs2027433 Chr.9: 108875024-108875024 on Build GRCh38
rs752124878 Chr.9: 108875015-108875015 on Build GRCh38

View Full Product Details