Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181237550-181237811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817731 c.826A>G p.S276G Chr.5: 181237671-181237671 on Build GRCh38
3013126 c.821T>G p.V274G Chr.5: 181237676-181237676 on Build GRCh38
6580932 c.781T>A p.L261I Chr.5: 181237716-181237716 on Build GRCh38
6672305 c.888G>A p.Q296Q Chr.5: 181237609-181237609 on Build GRCh38
5968122 c.852C>A p.P284P Chr.5: 181237645-181237645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377426248 Chr.5: 181237770-181237770 on Build GRCh38
rs144585710 Chr.5: 181237621-181237621 on Build GRCh38
rs201194486 Chr.5: 181237647-181237647 on Build GRCh38
rs199825760 Chr.5: 181237759-181237761 on Build GRCh38
rs11540195 Chr.5: 181237640-181237640 on Build GRCh38
rs762201797 Chr.5: 181237639-181237639 on Build GRCh38
rs961577153 Chr.5: 181237754-181237754 on Build GRCh38
rs770729712 Chr.5: 181237764-181237764 on Build GRCh38
rs373456604 Chr.5: 181237725-181237725 on Build GRCh38
rs796428056 Chr.5: 181237760-181237760 on Build GRCh38
rs972626674 Chr.5: 181237756-181237756 on Build GRCh38
rs11540194 Chr.5: 181237663-181237663 on Build GRCh38
rs377665647 Chr.5: 181237593-181237593 on Build GRCh38
rs112941138 Chr.5: 181237766-181237766 on Build GRCh38
rs746524968 Chr.5: 181237725-181237726 on Build GRCh38
rs869061550 Chr.5: 181237765-181237766 on Build GRCh38
rs762831199 Chr.5: 181237599-181237599 on Build GRCh38
rs372982968 Chr.5: 181237738-181237738 on Build GRCh38
rs769519658 Chr.5: 181237761-181237762 on Build GRCh38
rs761693579 Chr.5: 181237684-181237684 on Build GRCh38
rs780688483 Chr.5: 181237627-181237627 on Build GRCh38
rs747777388 Chr.5: 181237767-181237767 on Build GRCh38
rs34681418 Chr.5: 181237761-181237761 on Build GRCh38
rs35542217 Chr.5: 181237762-181237762 on Build GRCh38
rs770454166 Chr.5: 181237600-181237600 on Build GRCh38
rs772500913 Chr.5: 181237756-181237760 on Build GRCh38
rs748720438 Chr.5: 181237744-181237744 on Build GRCh38
rs780454789 Chr.5: 181237579-181237579 on Build GRCh38
rs781480812 Chr.5: 181237589-181237589 on Build GRCh38
rs145268208 Chr.5: 181237760-181237762 on Build GRCh38
rs780364981 Chr.5: 181237702-181237702 on Build GRCh38
rs375875847 Chr.5: 181237761-181237763 on Build GRCh38
rs918992829 Chr.5: 181237636-181237636 on Build GRCh38
rs574355393 Chr.5: 181237602-181237602 on Build GRCh38
rs972937702 Chr.5: 181237598-181237598 on Build GRCh38
rs113703732 Chr.5: 181237608-181237608 on Build GRCh38
rs775048852 Chr.5: 181237768-181237783 on Build GRCh38
rs113171370 Chr.5: 181237721-181237721 on Build GRCh38
rs747018485 Chr.5: 181237580-181237580 on Build GRCh38
rs758794696 Chr.5: 181237687-181237687 on Build GRCh38
rs1022596913 Chr.5: 181237642-181237642 on Build GRCh38
rs113238877 Chr.5: 181237607-181237607 on Build GRCh38
rs201737391 Chr.5: 181237768-181237768 on Build GRCh38
rs748310828 Chr.5: 181237729-181237729 on Build GRCh38
rs752241294 Chr.5: 181237633-181237633 on Build GRCh38
rs748974106 Chr.5: 181237744-181237744 on Build GRCh38
rs774000057 Chr.5: 181237601-181237601 on Build GRCh38
rs138107666 Chr.5: 181237660-181237660 on Build GRCh38
rs768870758 Chr.5: 181237585-181237585 on Build GRCh38
rs781391304 Chr.5: 181237728-181237728 on Build GRCh38
rs200634984 Chr.5: 181237701-181237701 on Build GRCh38

View Full Product Details