Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22077273-22077532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs775603606 Chr.X: 22077482-22077482 on Build GRCh38
rs1017946566 Chr.X: 22077409-22077409 on Build GRCh38
rs896008849 Chr.X: 22077354-22077354 on Build GRCh38
rs749081778 Chr.X: 22077479-22077479 on Build GRCh38
rs879069482 Chr.X: 22077324-22077324 on Build GRCh38
rs904755800 Chr.X: 22077407-22077407 on Build GRCh38
rs748086510 Chr.X: 22077459-22077459 on Build GRCh38
rs756115044 Chr.X: 22077460-22077460 on Build GRCh38
rs1000308958 Chr.X: 22077408-22077408 on Build GRCh38
rs748320660 Chr.X: 22077295-22077295 on Build GRCh38
rs754508886 Chr.X: 22077441-22077441 on Build GRCh38
rs970161374 Chr.X: 22077469-22077469 on Build GRCh38
rs1042394018 Chr.X: 22077406-22077406 on Build GRCh38
rs999045469 Chr.X: 22077376-22077376 on Build GRCh38
rs768868675 Chr.X: 22077489-22077489 on Build GRCh38
rs777749067 Chr.X: 22077471-22077471 on Build GRCh38
rs879045425 Chr.X: 22077327-22077327 on Build GRCh38
rs770301827 Chr.X: 22077381-22077381 on Build GRCh38
rs145881500 Chr.X: 22077483-22077483 on Build GRCh38
rs997951924 Chr.X: 22077467-22077467 on Build GRCh38
rs886042025 Chr.X: 22077487-22077487 on Build GRCh38
rs780626060 Chr.X: 22077454-22077454 on Build GRCh38
rs770726332 Chr.X: 22077480-22077480 on Build GRCh38

View Full Product Details