Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16986406-16986661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607287 c.3320C>T p.A1107V Chr.1: 16986548-16986548 on Build GRCh38
899711 c.3352C>A p.L1118M Chr.1: 16986516-16986516 on Build GRCh38
5844514 c.3361_3363delCTG p.L1121delL Chr.1: 16986505-16986507 on Build GRCh38
4622291 c.3283C>A p.L1095I Chr.1: 16986585-16986585 on Build GRCh38
5527091 c.3378C>T p.L1126L Chr.1: 16986490-16986490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765499517 Chr.1: 16986450-16986450 on Build GRCh38
rs761814059 Chr.1: 16986612-16986612 on Build GRCh38
rs775186212 Chr.1: 16986433-16986433 on Build GRCh38
rs34585239 Chr.1: 16986546-16986546 on Build GRCh38
rs537780162 Chr.1: 16986445-16986445 on Build GRCh38
rs750573377 Chr.1: 16986536-16986536 on Build GRCh38
rs765405083 Chr.1: 16986535-16986535 on Build GRCh38
rs137853968 Chr.1: 16986615-16986615 on Build GRCh38
rs867332959 Chr.1: 16986488-16986488 on Build GRCh38
rs373514205 Chr.1: 16986484-16986484 on Build GRCh38
rs112261486 Chr.1: 16986541-16986541 on Build GRCh38
rs755468538 Chr.1: 16986477-16986477 on Build GRCh38
rs369705373 Chr.1: 16986468-16986468 on Build GRCh38
rs763960380 Chr.1: 16986436-16986436 on Build GRCh38
rs778370632 Chr.1: 16986592-16986592 on Build GRCh38
rs115985012 Chr.1: 16986526-16986526 on Build GRCh38
rs781551851 Chr.1: 16986483-16986483 on Build GRCh38
rs770410981 Chr.1: 16986487-16986487 on Build GRCh38
rs541932176 Chr.1: 16986597-16986597 on Build GRCh38
rs145674970 Chr.1: 16986525-16986525 on Build GRCh38
rs769904401 Chr.1: 16986628-16986628 on Build GRCh38
rs375485447 Chr.1: 16986453-16986453 on Build GRCh38
rs755307953 Chr.1: 16986548-16986548 on Build GRCh38
rs771759787 Chr.1: 16986501-16986501 on Build GRCh38
rs757418466 Chr.1: 16986447-16986447 on Build GRCh38
rs751916254 Chr.1: 16986547-16986547 on Build GRCh38
rs781661852 Chr.1: 16986553-16986553 on Build GRCh38
rs377253172 Chr.1: 16986610-16986610 on Build GRCh38
rs201756175 Chr.1: 16986554-16986554 on Build GRCh38
rs771715671 Chr.1: 16986598-16986598 on Build GRCh38
rs780437824 Chr.1: 16986459-16986459 on Build GRCh38
rs756650754 Chr.1: 16986571-16986571 on Build GRCh38
rs758763354 Chr.1: 16986456-16986456 on Build GRCh38
rs370677966 Chr.1: 16986611-16986611 on Build GRCh38
rs760637978 Chr.1: 16986435-16986435 on Build GRCh38
rs374766933 Chr.1: 16986454-16986454 on Build GRCh38
rs773379849 Chr.1: 16986629-16986629 on Build GRCh38
rs927365149 Chr.1: 16986519-16986519 on Build GRCh38
rs372810630 Chr.1: 16986570-16986570 on Build GRCh38
rs773914880 Chr.1: 16986490-16986490 on Build GRCh38
rs764025000 Chr.1: 16986513-16986513 on Build GRCh38
rs758675397 Chr.1: 16986539-16986539 on Build GRCh38
rs745487724 Chr.1: 16986491-16986491 on Build GRCh38
rs886045577 Chr.1: 16986567-16986567 on Build GRCh38
rs375453342 Chr.1: 16986512-16986512 on Build GRCh38
rs913156094 Chr.1: 16986448-16986448 on Build GRCh38
rs775304821 Chr.1: 16986503-16986503 on Build GRCh38

View Full Product Details