Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38349433-38349688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238234 c.633C>T p.Y211Y Chr.15: 38349472-38349472 on Build GRCh38
5610689 c.676C>T p.Q226* Chr.15: 38349515-38349515 on Build GRCh38
6789281 c.641G>A p.R214Q Chr.15: 38349480-38349480 on Build GRCh38
4054221 c.657A>C p.E219D Chr.15: 38349496-38349496 on Build GRCh38
4054220 c.634G>A p.V212I Chr.15: 38349473-38349473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144764225 Chr.15: 38349514-38349514 on Build GRCh38
rs766539664 Chr.15: 38349502-38349502 on Build GRCh38
rs368660900 Chr.15: 38349458-38349458 on Build GRCh38
rs769630742 Chr.15: 38349473-38349473 on Build GRCh38
rs752775519 Chr.15: 38349524-38349524 on Build GRCh38
rs759623047 Chr.15: 38349563-38349564 on Build GRCh38
rs76305448 Chr.15: 38349656-38349656 on Build GRCh38
rs754544760 Chr.15: 38349576-38349578 on Build GRCh38
rs951963194 Chr.15: 38349522-38349522 on Build GRCh38
rs751245187 Chr.15: 38349575-38349577 on Build GRCh38
rs907878287 Chr.15: 38349572-38349572 on Build GRCh38
rs61670754 Chr.15: 38349572-38349573 on Build GRCh38
rs371487006 Chr.15: 38349577-38349577 on Build GRCh38
rs754209977 Chr.15: 38349551-38349551 on Build GRCh38
rs144227155 Chr.15: 38349484-38349484 on Build GRCh38
rs61467691 Chr.15: 38349574-38349575 on Build GRCh38
rs761179655 Chr.15: 38349537-38349537 on Build GRCh38
rs200338097 Chr.15: 38349573-38349573 on Build GRCh38
rs149991304 Chr.15: 38349472-38349472 on Build GRCh38
rs774530752 Chr.15: 38349508-38349508 on Build GRCh38
rs530400140 Chr.15: 38349582-38349582 on Build GRCh38
rs760448920 Chr.15: 38349576-38349576 on Build GRCh38
rs774637929 Chr.15: 38349560-38349561 on Build GRCh38
rs376699107 Chr.15: 38349479-38349479 on Build GRCh38
rs779460733 Chr.15: 38349568-38349568 on Build GRCh38
rs746708059 Chr.15: 38349457-38349457 on Build GRCh38
rs121434316 Chr.15: 38349476-38349476 on Build GRCh38
rs779879921 Chr.15: 38349453-38349453 on Build GRCh38
rs985085386 Chr.15: 38349528-38349528 on Build GRCh38
rs34352434 Chr.15: 38349573-38349574 on Build GRCh38
rs757678569 Chr.15: 38349552-38349552 on Build GRCh38
rs59928364 Chr.15: 38349576-38349577 on Build GRCh38
rs869050671 Chr.15: 38349577-38349578 on Build GRCh38
rs67551443 Chr.15: 38349575-38349576 on Build GRCh38
rs121434314 Chr.15: 38349482-38349482 on Build GRCh38
rs764046710 Chr.15: 38349573-38349573 on Build GRCh38
rs542270695 Chr.15: 38349589-38349589 on Build GRCh38
rs947223730 Chr.15: 38349555-38349555 on Build GRCh38
rs990641041 Chr.15: 38349507-38349507 on Build GRCh38
rs796273851 Chr.15: 38349573-38349576 on Build GRCh38
rs114258866 Chr.15: 38349576-38349576 on Build GRCh38
rs751254307 Chr.15: 38349569-38349569 on Build GRCh38
rs771126418 Chr.15: 38349559-38349560 on Build GRCh38
rs776073446 Chr.15: 38349463-38349463 on Build GRCh38
rs371274407 Chr.15: 38349480-38349480 on Build GRCh38
rs764543358 Chr.15: 38349549-38349549 on Build GRCh38

View Full Product Details