Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226885509-226885766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSEN2
Entrez Gene ID
5664
Gene Name
presenilin 2
Gene Aliases
AD3L, AD4, CMD1V, PS2, STM2
UniGene
Hs.25363
Gene Chromosome Location
Chr.1: 226870572-226903829 on Build GRCh38
Transcripts
NM_000447.2 NM_012486.2 XM_005273199.3 XM_017001835.1 XM_017001836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4978975 c.447C>T p.I149I Chr.1: 226885628-226885628 on Build GRCh38
904771 c.413C>G p.S138C Chr.1: 226885594-226885594 on Build GRCh38
904772 c.475C>A p.L159I Chr.1: 226885656-226885656 on Build GRCh38
4973719 c.415G>A p.V139M Chr.1: 226885596-226885596 on Build GRCh38
904770 c.401G>A p.R134H Chr.1: 226885582-226885582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201196514 Chr.1: 226885610-226885610 on Build GRCh38
rs772893473 Chr.1: 226885696-226885696 on Build GRCh38
rs753615781 Chr.1: 226885709-226885709 on Build GRCh38
rs63750215 Chr.1: 226885603-226885603 on Build GRCh38
rs985104147 Chr.1: 226885661-226885661 on Build GRCh38
rs202178897 Chr.1: 226885596-226885596 on Build GRCh38
rs200931244 Chr.1: 226885668-226885668 on Build GRCh38
rs767829568 Chr.1: 226885686-226885700 on Build GRCh38
rs139573101 Chr.1: 226885641-226885641 on Build GRCh38
rs541495335 Chr.1: 226885747-226885747 on Build GRCh38
rs760299337 Chr.1: 226885600-226885600 on Build GRCh38
rs756296193 Chr.1: 226885648-226885648 on Build GRCh38
rs766702020 Chr.1: 226885734-226885734 on Build GRCh38
rs767181943 Chr.1: 226885534-226885534 on Build GRCh38
rs778936527 Chr.1: 226885669-226885669 on Build GRCh38
rs28936380 Chr.1: 226885546-226885546 on Build GRCh38
rs200981406 Chr.1: 226885535-226885535 on Build GRCh38
rs866044092 Chr.1: 226885629-226885629 on Build GRCh38
rs200924879 Chr.1: 226885592-226885592 on Build GRCh38
rs770519379 Chr.1: 226885707-226885707 on Build GRCh38
rs116003409 Chr.1: 226885604-226885604 on Build GRCh38
rs372616074 Chr.1: 226885532-226885532 on Build GRCh38
rs747738607 Chr.1: 226885595-226885595 on Build GRCh38
rs768523579 Chr.1: 226885527-226885527 on Build GRCh38
rs1004970794 Chr.1: 226885617-226885617 on Build GRCh38
rs375424407 Chr.1: 226885692-226885692 on Build GRCh38
rs758985161 Chr.1: 226885556-226885556 on Build GRCh38
rs148996705 Chr.1: 226885547-226885547 on Build GRCh38
rs566090689 Chr.1: 226885746-226885746 on Build GRCh38
rs370607299 Chr.1: 226885688-226885688 on Build GRCh38
rs200824392 Chr.1: 226885703-226885703 on Build GRCh38
rs61761208 Chr.1: 226885602-226885602 on Build GRCh38
rs547731225 Chr.1: 226885559-226885559 on Build GRCh38
rs566361436 Chr.1: 226885563-226885563 on Build GRCh38
rs199654606 Chr.1: 226885636-226885636 on Build GRCh38
rs903335386 Chr.1: 226885608-226885608 on Build GRCh38
rs761557286 Chr.1: 226885533-226885533 on Build GRCh38
rs373254101 Chr.1: 226885737-226885737 on Build GRCh38
rs756346917 Chr.1: 226885582-226885582 on Build GRCh38
rs780284265 Chr.1: 226885650-226885650 on Build GRCh38
rs750095720 Chr.1: 226885536-226885536 on Build GRCh38
rs149428641 Chr.1: 226885694-226885694 on Build GRCh38
rs200617488 Chr.1: 226885718-226885718 on Build GRCh38
rs748154107 Chr.1: 226885676-226885676 on Build GRCh38
rs1017893609 Chr.1: 226885693-226885693 on Build GRCh38
rs773059327 Chr.1: 226885599-226885599 on Build GRCh38
rs2236910 Chr.1: 226885709-226885709 on Build GRCh38
rs63750812 Chr.1: 226885623-226885623 on Build GRCh38
rs753371172 Chr.1: 226885553-226885553 on Build GRCh38
rs63749851 Chr.1: 226885545-226885545 on Build GRCh38
rs781597971 Chr.1: 226885571-226885571 on Build GRCh38
rs201613477 Chr.1: 226885700-226885700 on Build GRCh38
rs755101354 Chr.1: 226885663-226885663 on Build GRCh38
rs144791481 Chr.1: 226885719-226885719 on Build GRCh38
rs114334281 Chr.1: 226885622-226885622 on Build GRCh38
rs201892898 Chr.1: 226885620-226885620 on Build GRCh38
rs200108534 Chr.1: 226885727-226885727 on Build GRCh38
rs764718172 Chr.1: 226885611-226885611 on Build GRCh38
rs767835970 Chr.1: 226885628-226885628 on Build GRCh38
rs746117072 Chr.1: 226885580-226885580 on Build GRCh38
rs749301595 Chr.1: 226885591-226885591 on Build GRCh38
rs752002301 Chr.1: 226885564-226885564 on Build GRCh38
rs201590095 Chr.1: 226885605-226885605 on Build GRCh38
rs868256785 Chr.1: 226885738-226885738 on Build GRCh38
rs750785455 Chr.1: 226885729-226885729 on Build GRCh38
rs63750197 Chr.1: 226885570-226885570 on Build GRCh38

View Full Product Details