Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62690673-62690898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442056 c.1106C>A p.S369* Chr.11: 62690834-62690834 on Build GRCh38
4165878 c.1154-8G>A p.? Chr.11: 62690700-62690700 on Build GRCh38
6613136 c.1167C>T p.S389S Chr.11: 62690679-62690679 on Build GRCh38
3752583 c.945A>G p.E315E Chr.11: 62690803-62690803 on Build GRCh38
5475550 c.961+7C>T p.? Chr.11: 62690780-62690780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772967591 Chr.11: 62690759-62690759 on Build GRCh38
rs747846786 Chr.11: 62690829-62690829 on Build GRCh38
rs372844289 Chr.11: 62690738-62690738 on Build GRCh38
rs6856 Chr.11: 62690803-62690803 on Build GRCh38
rs991114344 Chr.11: 62690713-62690713 on Build GRCh38
rs771892704 Chr.11: 62690751-62690751 on Build GRCh38
rs776379711 Chr.11: 62690844-62690844 on Build GRCh38
rs778455259 Chr.11: 62690820-62690820 on Build GRCh38
rs149907021 Chr.11: 62690795-62690795 on Build GRCh38
rs768368743 Chr.11: 62690732-62690732 on Build GRCh38
rs758675404 Chr.11: 62690780-62690780 on Build GRCh38
rs202041376 Chr.11: 62690870-62690870 on Build GRCh38
rs759471836 Chr.11: 62690772-62690772 on Build GRCh38
rs762174973 Chr.11: 62690793-62690794 on Build GRCh38
rs144245125 Chr.11: 62690840-62690840 on Build GRCh38
rs757846034 Chr.11: 62690796-62690796 on Build GRCh38
rs372839524 Chr.11: 62690779-62690779 on Build GRCh38
rs1007078676 Chr.11: 62690836-62690836 on Build GRCh38
rs1000536287 Chr.11: 62690867-62690867 on Build GRCh38
rs747967927 Chr.11: 62690746-62690746 on Build GRCh38
rs149990643 Chr.11: 62690838-62690838 on Build GRCh38
rs76184645 Chr.11: 62690761-62690761 on Build GRCh38
rs374170281 Chr.11: 62690810-62690810 on Build GRCh38
rs1010643563 Chr.11: 62690841-62690841 on Build GRCh38
rs564980539 Chr.11: 62690766-62690766 on Build GRCh38
rs369798542 Chr.11: 62690771-62690771 on Build GRCh38
rs765531343 Chr.11: 62690775-62690775 on Build GRCh38
rs748680075 Chr.11: 62690715-62690715 on Build GRCh38
rs368900617 Chr.11: 62690802-62690802 on Build GRCh38
rs771698323 Chr.11: 62690834-62690834 on Build GRCh38
rs772516974 Chr.11: 62690793-62690793 on Build GRCh38
rs150914934 Chr.11: 62690797-62690797 on Build GRCh38
rs746737457 Chr.11: 62690839-62690839 on Build GRCh38
rs764682700 Chr.11: 62690851-62690851 on Build GRCh38
rs199558387 Chr.11: 62690784-62690784 on Build GRCh38

View Full Product Details