Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23988395-23988664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6078800 c.260C>A p.P87H Chr.16: 23988562-23988562 on Build GRCh38
4717694 c.232T>C p.C78R Chr.16: 23988534-23988534 on Build GRCh38
4717696 c.232T>C p.C78R Chr.16: 23988534-23988534 on Build GRCh38
1129022 c.285C>T p.S95S Chr.16: 23988587-23988587 on Build GRCh38
4402091 c.238G>A p.E80K Chr.16: 23988540-23988540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780807794 Chr.16: 23988602-23988602 on Build GRCh38
rs745654273 Chr.16: 23988620-23988620 on Build GRCh38
rs374737151 Chr.16: 23988636-23988636 on Build GRCh38
rs1002043513 Chr.16: 23988508-23988508 on Build GRCh38
rs759454355 Chr.16: 23988521-23988521 on Build GRCh38
rs749204741 Chr.16: 23988609-23988609 on Build GRCh38
rs764380918 Chr.16: 23988533-23988533 on Build GRCh38
rs750901145 Chr.16: 23988571-23988571 on Build GRCh38
rs755399339 Chr.16: 23988589-23988589 on Build GRCh38
rs770813136 Chr.16: 23988481-23988481 on Build GRCh38
rs370311602 Chr.16: 23988511-23988511 on Build GRCh38
rs747765955 Chr.16: 23988604-23988604 on Build GRCh38
rs542028487 Chr.16: 23988472-23988472 on Build GRCh38
rs758927501 Chr.16: 23988581-23988581 on Build GRCh38
rs762805670 Chr.16: 23988476-23988476 on Build GRCh38
rs3729888 Chr.16: 23988607-23988607 on Build GRCh38
rs115750906 Chr.16: 23988566-23988566 on Build GRCh38
rs41276940 Chr.16: 23988632-23988632 on Build GRCh38
rs974969519 Chr.16: 23988610-23988610 on Build GRCh38
rs1033653376 Chr.16: 23988514-23988514 on Build GRCh38
rs892325206 Chr.16: 23988439-23988439 on Build GRCh38
rs754076764 Chr.16: 23988557-23988557 on Build GRCh38
rs371650623 Chr.16: 23988567-23988567 on Build GRCh38
rs371087215 Chr.16: 23988587-23988587 on Build GRCh38
rs751886977 Chr.16: 23988588-23988588 on Build GRCh38
rs776856807 Chr.16: 23988633-23988633 on Build GRCh38
rs963323900 Chr.16: 23988518-23988518 on Build GRCh38
rs3729887 Chr.16: 23988462-23988462 on Build GRCh38
rs987653457 Chr.16: 23988575-23988575 on Build GRCh38
rs183413076 Chr.16: 23988459-23988459 on Build GRCh38
rs770974182 Chr.16: 23988619-23988619 on Build GRCh38

View Full Product Details