Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65974788-65975037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4847174 c.1054G>A p.E352K Chr.7: 65974930-65974930 on Build GRCh38
3833087 c.959A>G p.Y320C Chr.7: 65975025-65975025 on Build GRCh38
4977923 c.1011C>G p.L337L Chr.7: 65974973-65974973 on Build GRCh38
1242337 c.980G>A p.R327H Chr.7: 65975004-65975004 on Build GRCh38
461565 c.950C>T p.S317F Chr.7: 65975034-65975034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764995331 Chr.7: 65974876-65974876 on Build GRCh38
rs144455622 Chr.7: 65974918-65974918 on Build GRCh38
rs970690016 Chr.7: 65974891-65974891 on Build GRCh38
rs753206771 Chr.7: 65974906-65974906 on Build GRCh38
rs763305089 Chr.7: 65974881-65974881 on Build GRCh38
rs141303888 Chr.7: 65974943-65974943 on Build GRCh38
rs775005205 Chr.7: 65974905-65974905 on Build GRCh38
rs147820162 Chr.7: 65974940-65974940 on Build GRCh38
rs1047738535 Chr.7: 65974834-65974834 on Build GRCh38
rs750234091 Chr.7: 65974879-65974879 on Build GRCh38
rs773187021 Chr.7: 65974946-65974946 on Build GRCh38
rs767561248 Chr.7: 65974973-65974973 on Build GRCh38
rs756639453 Chr.7: 65974908-65974908 on Build GRCh38
rs973171845 Chr.7: 65975011-65975011 on Build GRCh38
rs999305370 Chr.7: 65974843-65974843 on Build GRCh38
rs772885108 Chr.7: 65974959-65974959 on Build GRCh38
rs141884647 Chr.7: 65974844-65974844 on Build GRCh38
rs368276036 Chr.7: 65974810-65974810 on Build GRCh38
rs761680481 Chr.7: 65974875-65974875 on Build GRCh38
rs754950198 Chr.7: 65974893-65974893 on Build GRCh38
rs200412277 Chr.7: 65974928-65974928 on Build GRCh38
rs1051843349 Chr.7: 65974819-65974819 on Build GRCh38
rs1021888229 Chr.7: 65974889-65974889 on Build GRCh38
rs150782923 Chr.7: 65974863-65974863 on Build GRCh38
rs191153460 Chr.7: 65974933-65974933 on Build GRCh38
rs890225813 Chr.7: 65974826-65974826 on Build GRCh38
rs766682710 Chr.7: 65974888-65974888 on Build GRCh38
rs781730954 Chr.7: 65974897-65974897 on Build GRCh38
rs185307365 Chr.7: 65974861-65974861 on Build GRCh38
rs561880652 Chr.7: 65974996-65974996 on Build GRCh38
rs752792335 Chr.7: 65974976-65974976 on Build GRCh38
rs767992971 Chr.7: 65974931-65974931 on Build GRCh38
rs758070679 Chr.7: 65974919-65974919 on Build GRCh38
rs762798994 Chr.7: 65974953-65974953 on Build GRCh38
rs121918182 Chr.7: 65974934-65974934 on Build GRCh38
rs774381911 Chr.7: 65974961-65974961 on Build GRCh38
rs531607427 Chr.7: 65974942-65974942 on Build GRCh38
rs1052167702 Chr.7: 65974862-65974862 on Build GRCh38
rs368189360 Chr.7: 65974959-65974959 on Build GRCh38
rs554759648 Chr.7: 65974922-65974922 on Build GRCh38
rs766716968 Chr.7: 65974960-65974960 on Build GRCh38
rs781143 Chr.7: 65974892-65974892 on Build GRCh38
rs121918175 Chr.7: 65974923-65974923 on Build GRCh38
rs776365387 Chr.7: 65974874-65974874 on Build GRCh38
rs778151820 Chr.7: 65974915-65974915 on Build GRCh38
rs886412828 Chr.7: 65974860-65974860 on Build GRCh38
rs372851133 Chr.7: 65974964-65974964 on Build GRCh38

View Full Product Details