Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151027870-151028103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX3
Entrez Gene ID
2878
Gene Name
glutathione peroxidase 3
Gene Aliases
GPx-P, GSHPx-3, GSHPx-P
UniGene
Hs.386793
Gene Chromosome Location
Chr.5: 151020438-151028993 on Build GRCh38
Transcripts
NM_002084.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223036 c.575G>A p.G192D Chr.5: 151028024-151028024 on Build GRCh38
6996528 c.502C>T p.R168C Chr.5: 151027951-151027951 on Build GRCh38
4688357 c.538C>T p.R180C Chr.5: 151027987-151027987 on Build GRCh38
288952 c.539G>A p.R180H Chr.5: 151027988-151027988 on Build GRCh38
7104250 c.484C>T p.L162F Chr.5: 151027933-151027933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746734937 Chr.5: 151027951-151027951 on Build GRCh38
rs750182118 Chr.5: 151027907-151027907 on Build GRCh38
rs374193246 Chr.5: 151027952-151027952 on Build GRCh38
rs368970475 Chr.5: 151028064-151028064 on Build GRCh38
rs755316250 Chr.5: 151028024-151028038 on Build GRCh38
rs751139605 Chr.5: 151028063-151028063 on Build GRCh38
rs1040271400 Chr.5: 151028050-151028050 on Build GRCh38
rs779157665 Chr.5: 151027949-151027949 on Build GRCh38
rs574927554 Chr.5: 151027992-151027992 on Build GRCh38
rs758219351 Chr.5: 151027914-151027914 on Build GRCh38
rs560735748 Chr.5: 151028057-151028057 on Build GRCh38
rs755686829 Chr.5: 151028073-151028073 on Build GRCh38
rs1047991777 Chr.5: 151028079-151028079 on Build GRCh38
rs780644613 Chr.5: 151027991-151027991 on Build GRCh38
rs375837316 Chr.5: 151028058-151028058 on Build GRCh38
rs886805020 Chr.5: 151028081-151028081 on Build GRCh38
rs934659851 Chr.5: 151027930-151027930 on Build GRCh38
rs752213053 Chr.5: 151028068-151028068 on Build GRCh38
rs748643419 Chr.5: 151027953-151027953 on Build GRCh38
rs765184725 Chr.5: 151027896-151027896 on Build GRCh38
rs772121564 Chr.5: 151028034-151028034 on Build GRCh38
rs372652345 Chr.5: 151028039-151028039 on Build GRCh38
rs542081520 Chr.5: 151028049-151028049 on Build GRCh38
rs1003872944 Chr.5: 151028082-151028082 on Build GRCh38
rs888180765 Chr.5: 151027944-151027944 on Build GRCh38
rs779516716 Chr.5: 151027929-151027929 on Build GRCh38
rs771337658 Chr.5: 151027988-151027988 on Build GRCh38
rs949325290 Chr.5: 151027893-151027893 on Build GRCh38
rs200470998 Chr.5: 151027915-151027915 on Build GRCh38
rs986650820 Chr.5: 151027905-151027905 on Build GRCh38
rs184186742 Chr.5: 151027966-151027966 on Build GRCh38
rs556389247 Chr.5: 151027981-151027981 on Build GRCh38
rs770375282 Chr.5: 151027956-151027956 on Build GRCh38
rs8177447 Chr.5: 151027895-151027895 on Build GRCh38
rs759865306 Chr.5: 151028024-151028024 on Build GRCh38
rs77994288 Chr.5: 151027906-151027906 on Build GRCh38
rs929597263 Chr.5: 151027933-151027933 on Build GRCh38
rs757168447 Chr.5: 151027980-151027980 on Build GRCh38
rs370891492 Chr.5: 151028051-151028051 on Build GRCh38
rs1048188676 Chr.5: 151027935-151027935 on Build GRCh38
rs371559625 Chr.5: 151028067-151028067 on Build GRCh38

View Full Product Details