Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196998237-196998478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR5
Entrez Gene ID
81494
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
UniGene
Hs.282594
Gene Chromosome Location
Chr.1: 196975022-197009725 on Build GRCh38
Transcripts
NM_030787.3 XM_011510020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229666 c.1124G>A p.W375* Chr.1: 196998281-196998281 on Build GRCh38
1748045 c.1204G>A p.E402K Chr.1: 196998289-196998289 on Build GRCh38
1721694 c.1219G>A p.E407K Chr.1: 196998304-196998304 on Build GRCh38
5813164 c.1142G>C p.C381S Chr.1: 196998299-196998299 on Build GRCh38
3480964 c.1129C>T p.P377S Chr.1: 196998286-196998286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047249338 Chr.1: 196998401-196998401 on Build GRCh38
rs761669618 Chr.1: 196998341-196998341 on Build GRCh38
rs149179493 Chr.1: 196998416-196998416 on Build GRCh38
rs531979314 Chr.1: 196998372-196998372 on Build GRCh38
rs748127696 Chr.1: 196998274-196998274 on Build GRCh38
rs543369125 Chr.1: 196998357-196998357 on Build GRCh38
rs1031528727 Chr.1: 196998325-196998325 on Build GRCh38
rs1001577669 Chr.1: 196998442-196998442 on Build GRCh38
rs748631825 Chr.1: 196998293-196998293 on Build GRCh38
rs963558853 Chr.1: 196998431-196998431 on Build GRCh38
rs905537751 Chr.1: 196998425-196998425 on Build GRCh38
rs111327589 Chr.1: 196998292-196998292 on Build GRCh38
rs777962172 Chr.1: 196998298-196998298 on Build GRCh38
rs1045686922 Chr.1: 196998423-196998423 on Build GRCh38
rs750077898 Chr.1: 196998263-196998263 on Build GRCh38
rs544997795 Chr.1: 196998297-196998297 on Build GRCh38
rs749031519 Chr.1: 196998324-196998324 on Build GRCh38
rs888736775 Chr.1: 196998413-196998413 on Build GRCh38
rs779126931 Chr.1: 196998294-196998294 on Build GRCh38
rs756599015 Chr.1: 196998324-196998324 on Build GRCh38
rs778320306 Chr.1: 196998350-196998350 on Build GRCh38
rs771598432 Chr.1: 196998351-196998351 on Build GRCh38
rs116891819 Chr.1: 196998398-196998398 on Build GRCh38
rs747255817 Chr.1: 196998302-196998302 on Build GRCh38
rs922657988 Chr.1: 196998392-196998392 on Build GRCh38
rs373303829 Chr.1: 196998349-196998349 on Build GRCh38
rs561546276 Chr.1: 196998358-196998358 on Build GRCh38
rs771254388 Chr.1: 196998309-196998309 on Build GRCh38
rs755579894 Chr.1: 196998289-196998289 on Build GRCh38
rs376742710 Chr.1: 196998275-196998275 on Build GRCh38
rs1332664 Chr.1: 196998328-196998328 on Build GRCh38
rs553453505 Chr.1: 196998322-196998322 on Build GRCh38
rs774165075 Chr.1: 196998340-196998340 on Build GRCh38
rs556270179 Chr.1: 196998273-196998273 on Build GRCh38

View Full Product Details