Gene Transcripts

Gene Symbol
Usp30
Entrez Gene ID
100756
Gene Name
ubiquitin specific peptidase 30
Gene Aliases
6330590F17Rik, AI851327, D5Ertd483e
Chromosome Location
Chr. 5: 114100328 - 114123510 on Build GRCm38
UniGene ID
Mm.240421
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033202.3 NP_001028374.1 7 748
GenBank mRNA AK078164.1 - 3 100
AK144578.1 - 7 749
AK216289.1 - 3 114
BC038606.1 - 7 770

View Full Product Details