Gene Transcripts

Gene Symbol
Psme4
Entrez Gene ID
103554
Gene Name
proteasome (prosome, macropain) activator subunit 4
Gene Aliases
AA409398, AU041366, TEMO, mKIAA0077
Chromosome Location
Chr. 11: 30771328 - 30881150 on Build GRCm38
UniGene ID
Mm.240066
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134013.3 NP_598774.2 10 1421
GenBank mRNA AF296169.1 - 10 1132
AK052279.1 - 1 2463
BC141381.1 - 10 1134
CV557303.1 - 3 290

View Full Product Details