Gene Transcripts

Gene Symbol
Osbpl11
Entrez Gene ID
106326
Gene Name
oxysterol binding protein-like 11
Gene Aliases
9430097N02Rik, AI132306, AU019795, ORP-11
Chromosome Location
Chr. 16: 33169763 - 33243312 on Build GRCm38
UniGene ID
Mm.486405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176840.3 NP_789810.2 6 1557
GenBank mRNA BC035278.1 - 6 1557

View Full Product Details