Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178143.2 | NP_835279.2 | 4 | 551 | |
GenBank mRNA | AK044030.1 | - | 4 | 387 | |
AK134573.1 | - | 4 | 409 | ||
BC138565.1 | - | 4 | 560 | 8861585 | |
BC138566.1 | - | 4 | 560 | 8861586 | |
BY718361.1 | - | 4 | 406 | ||
FJ588504.1 | - | 3 | 309 |