Gene Transcripts
- Gene Symbol
- Atad3a
- Entrez Gene ID
- 108888
- Gene Name
- ATPase family, AAA domain containing 3A
- Gene Aliases
- 2400004H09Rik, Atad3, Tob3, mKIAA1273
- Chromosome Location
- Chr. 4: 155740640 - 155761108 on Build GRCm38
- UniGene ID
- Mm.241152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_179203.3 |
NP_849534.2
|
4, 5 |
543 |
|
GenBank mRNA |
AF343079.1 |
- |
4, 5 |
589 |
|
AK075921.1 |
- |
4, 5 |
539 |
|
AK148974.1 |
- |
4, 5 |
542 |
|
AK149702.1 |
- |
4, 5 |
542 |
|
AK150037.1 |
- |
4, 5 |
542 |
|
AK165724.1 |
- |
4, 5 |
541 |
|
AK166099.1 |
- |
4, 5 |
539 |
|
AK172038.1 |
- |
4, 5 |
539 |
|
AK172350.1 |
- |
4, 5 |
544 |
|
AK173141.1 |
- |
4, 5 |
491 |
|
BC023301.1 |
- |
4, 5 |
480 |
3671787 |
BC058373.1 |
- |
4, 5 |
527 |
6828037 |
BC060036.1 |
- |
4, 5 |
475 |
6530155 |
View Full Product Details